Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86809082-86809627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS23
Entrez Gene ID
11098
Gene Name
protease, serine 23
Gene Aliases
SIG13, SPUVE, ZSIG13
UniGene
Hs.25338
Gene Chromosome Location
Chr.11: 86791059-86952910 on Build GRCh38
Transcripts
NM_001293178.1 NM_001293179.1 NM_001293180.1 NM_007173.5 NR_120591.1 NR_120592.1 NR_120593.1

SNP Detail

SNP ID Chromosome Location
rs936556039 Chr.11: 86809596-86809596 on Build GRCh38
rs1038875180 Chr.11: 86809472-86809472 on Build GRCh38
rs75414855 Chr.11: 86809593-86809593 on Build GRCh38
rs942821351 Chr.11: 86809328-86809328 on Build GRCh38
rs761356738 Chr.11: 86809265-86809265 on Build GRCh38
rs1015813034 Chr.11: 86809261-86809261 on Build GRCh38
rs10219261 Chr.11: 86809239-86809239 on Build GRCh38
rs937748919 Chr.11: 86809338-86809338 on Build GRCh38
rs867169152 Chr.11: 86809533-86809533 on Build GRCh38
rs891128897 Chr.11: 86809492-86809492 on Build GRCh38
rs77287545 Chr.11: 86809553-86809553 on Build GRCh38
rs1042703272 Chr.11: 86809381-86809381 on Build GRCh38
rs1051138243 Chr.11: 86809481-86809481 on Build GRCh38
rs963110736 Chr.11: 86809163-86809163 on Build GRCh38
rs926387406 Chr.11: 86809314-86809314 on Build GRCh38
rs924078458 Chr.11: 86809567-86809567 on Build GRCh38
rs974190031 Chr.11: 86809289-86809289 on Build GRCh38
rs983081220 Chr.11: 86809108-86809108 on Build GRCh38
rs1042260940 Chr.11: 86809114-86809114 on Build GRCh38
rs982424953 Chr.11: 86809238-86809238 on Build GRCh38
rs997340278 Chr.11: 86809476-86809476 on Build GRCh38
rs925221129 Chr.11: 86809595-86809595 on Build GRCh38
rs936505502 Chr.11: 86809405-86809405 on Build GRCh38
rs899890952 Chr.11: 86809136-86809136 on Build GRCh38
rs1015818842 Chr.11: 86809508-86809508 on Build GRCh38
rs893790420 Chr.11: 86809282-86809282 on Build GRCh38
rs962855156 Chr.11: 86809280-86809280 on Build GRCh38
rs546405526 Chr.11: 86809447-86809447 on Build GRCh38
rs959153505 Chr.11: 86809540-86809540 on Build GRCh38
rs143938670 Chr.11: 86809532-86809532 on Build GRCh38
rs1000664911 Chr.11: 86809120-86809120 on Build GRCh38
rs186958192 Chr.11: 86809227-86809227 on Build GRCh38
rs576333161 Chr.11: 86809294-86809294 on Build GRCh38
rs369197729 Chr.11: 86809499-86809499 on Build GRCh38
rs996887760 Chr.11: 86809170-86809170 on Build GRCh38
rs900365557 Chr.11: 86809362-86809362 on Build GRCh38
rs917570765 Chr.11: 86809368-86809368 on Build GRCh38
rs146137272 Chr.11: 86809193-86809193 on Build GRCh38
rs777355034 Chr.11: 86809536-86809536 on Build GRCh38
rs1015847019 Chr.11: 86809111-86809111 on Build GRCh38
rs1033095342 Chr.11: 86809131-86809131 on Build GRCh38
rs190388914 Chr.11: 86809557-86809557 on Build GRCh38
rs1040178202 Chr.11: 86809336-86809336 on Build GRCh38
rs1015901528 Chr.11: 86809458-86809458 on Build GRCh38
rs1004726597 Chr.11: 86809423-86809423 on Build GRCh38
rs903678576 Chr.11: 86809117-86809117 on Build GRCh38
rs773190568 Chr.11: 86809144-86809148 on Build GRCh38
rs545269111 Chr.11: 86809374-86809374 on Build GRCh38
rs560188443 Chr.11: 86809475-86809475 on Build GRCh38
rs917638589 Chr.11: 86809583-86809583 on Build GRCh38
rs568428789 Chr.11: 86809493-86809493 on Build GRCh38
rs962905561 Chr.11: 86809513-86809513 on Build GRCh38
rs935537364 Chr.11: 86809244-86809244 on Build GRCh38
rs2155080 Chr.11: 86809590-86809590 on Build GRCh38
rs990958820 Chr.11: 86809165-86809165 on Build GRCh38
rs532925833 Chr.11: 86809409-86809409 on Build GRCh38
rs758038291 Chr.11: 86809465-86809465 on Build GRCh38
rs970466018 Chr.11: 86809535-86809535 on Build GRCh38
rs191448390 Chr.11: 86809351-86809351 on Build GRCh38
rs949575750 Chr.11: 86809237-86809238 on Build GRCh38
rs1004177322 Chr.11: 86809494-86809494 on Build GRCh38
rs183606759 Chr.11: 86809403-86809403 on Build GRCh38
rs991862260 Chr.11: 86809360-86809360 on Build GRCh38

View Full Product Details