Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 88504437-88504939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRM5
Entrez Gene ID
2915
Gene Name
glutamate metabotropic receptor 5
Gene Aliases
GPRC1E, MGLUR5, PPP1R86, mGlu5
UniGene
Hs.147361
Gene Chromosome Location
Chr.11: 88504576-89065971 on Build GRCh38
Transcripts
NM_000842.4 NM_001143831.2 XM_006718828.3 XM_011542792.1 XM_017017627.1 NR_049724.1

Gene Symbol
GRM5-AS1
Entrez Gene ID
100873989
Gene Name
GRM5 antisense RNA 1
Gene Aliases
-
UniGene
Hs.616625
Gene Chromosome Location
Chr.11: 88504576-88524054 on Build GRCh38
Transcripts
NM_000842.4 NM_001143831.2 XM_006718828.3 XM_011542792.1 XM_017017627.1 NR_049724.1

SNP Detail

SNP ID Chromosome Location
rs999806383 Chr.11: 88504647-88504647 on Build GRCh38
rs117570066 Chr.11: 88504584-88504584 on Build GRCh38
rs972646379 Chr.11: 88504782-88504782 on Build GRCh38
rs992588960 Chr.11: 88504672-88504672 on Build GRCh38
rs1031408874 Chr.11: 88504662-88504662 on Build GRCh38
rs183613675 Chr.11: 88504706-88504706 on Build GRCh38
rs193229181 Chr.11: 88504912-88504912 on Build GRCh38
rs1037086011 Chr.11: 88504495-88504495 on Build GRCh38
rs1023689297 Chr.11: 88504673-88504673 on Build GRCh38
rs904112033 Chr.11: 88504636-88504636 on Build GRCh38
rs1022454564 Chr.11: 88504631-88504631 on Build GRCh38
rs578034467 Chr.11: 88504772-88504772 on Build GRCh38
rs941261165 Chr.11: 88504774-88504774 on Build GRCh38
rs952467502 Chr.11: 88504745-88504745 on Build GRCh38
rs992450803 Chr.11: 88504808-88504808 on Build GRCh38
rs558202556 Chr.11: 88504748-88504748 on Build GRCh38
rs556120841 Chr.11: 88504694-88504694 on Build GRCh38
rs933663342 Chr.11: 88504549-88504549 on Build GRCh38
rs959721147 Chr.11: 88504804-88504804 on Build GRCh38
rs536149834 Chr.11: 88504671-88504671 on Build GRCh38
rs1050726070 Chr.11: 88504828-88504828 on Build GRCh38
rs369226335 Chr.11: 88504492-88504492 on Build GRCh38
rs925449527 Chr.11: 88504892-88504892 on Build GRCh38
rs567116729 Chr.11: 88504642-88504642 on Build GRCh38
rs933631868 Chr.11: 88504815-88504815 on Build GRCh38
rs889535048 Chr.11: 88504593-88504593 on Build GRCh38
rs941325449 Chr.11: 88504494-88504494 on Build GRCh38
rs979926989 Chr.11: 88504756-88504756 on Build GRCh38
rs1042951498 Chr.11: 88504840-88504840 on Build GRCh38
rs1019974156 Chr.11: 88504693-88504694 on Build GRCh38
rs538252869 Chr.11: 88504747-88504747 on Build GRCh38
rs16914108 Chr.11: 88504813-88504813 on Build GRCh38
rs1043856380 Chr.11: 88504869-88504869 on Build GRCh38
rs542047299 Chr.11: 88504519-88504519 on Build GRCh38
rs942493771 Chr.11: 88504841-88504841 on Build GRCh38
rs1042552893 Chr.11: 88504532-88504532 on Build GRCh38
rs16914106 Chr.11: 88504712-88504712 on Build GRCh38
rs910964105 Chr.11: 88504829-88504829 on Build GRCh38
rs1006979375 Chr.11: 88504630-88504630 on Build GRCh38
rs1049774121 Chr.11: 88504604-88504604 on Build GRCh38
rs1000910241 Chr.11: 88504559-88504559 on Build GRCh38
rs188552689 Chr.11: 88504908-88504908 on Build GRCh38
rs960713301 Chr.11: 88504666-88504666 on Build GRCh38
rs1026785646 Chr.11: 88504718-88504718 on Build GRCh38
rs11020139 Chr.11: 88504481-88504481 on Build GRCh38
rs979798954 Chr.11: 88504746-88504747 on Build GRCh38
rs904050925 Chr.11: 88504878-88504878 on Build GRCh38
rs540580217 Chr.11: 88504865-88504865 on Build GRCh38
rs918517763 Chr.11: 88504793-88504793 on Build GRCh38
rs927032347 Chr.11: 88504777-88504777 on Build GRCh38

View Full Product Details