Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 93153965-93154506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC36A4
Entrez Gene ID
120103
Gene Name
solute carrier family 36 member 4
Gene Aliases
PAT4
UniGene
Hs.148766
Gene Chromosome Location
Chr.11: 93144171-93197983 on Build GRCh38
Transcripts
NM_001286139.1 NM_152313.3 XM_017017174.1 XM_017017175.1 XM_017017176.1 XM_017017177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3935679 c.1171G>T p.E391* Chr.11: 93154144-93154144 on Build GRCh38
3953977 c.1088A>G p.Y363C Chr.11: 93154227-93154227 on Build GRCh38
5991911 c.1038-7delT p.? Chr.11: 93154284-93154284 on Build GRCh38
1322077 c.1194G>T p.L398F Chr.11: 93154121-93154121 on Build GRCh38
6781883 c.1164A>G p.Q388Q Chr.11: 93154151-93154151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893401866 Chr.11: 93154340-93154340 on Build GRCh38
rs990752462 Chr.11: 93154014-93154014 on Build GRCh38
rs570439171 Chr.11: 93154063-93154063 on Build GRCh38
rs776977549 Chr.11: 93154277-93154277 on Build GRCh38
rs940469900 Chr.11: 93154206-93154206 on Build GRCh38
rs760918592 Chr.11: 93154079-93154079 on Build GRCh38
rs771185943 Chr.11: 93154272-93154272 on Build GRCh38
rs182082275 Chr.11: 93154130-93154130 on Build GRCh38
rs143575651 Chr.11: 93154217-93154217 on Build GRCh38
rs747261948 Chr.11: 93154175-93154175 on Build GRCh38
rs541371387 Chr.11: 93154011-93154011 on Build GRCh38
rs780602714 Chr.11: 93154155-93154155 on Build GRCh38
rs774253433 Chr.11: 93154298-93154298 on Build GRCh38
rs367750102 Chr.11: 93154240-93154240 on Build GRCh38
rs775412558 Chr.11: 93154201-93154201 on Build GRCh38
rs1035776323 Chr.11: 93154471-93154471 on Build GRCh38
rs374712275 Chr.11: 93154220-93154220 on Build GRCh38
rs139534791 Chr.11: 93154212-93154212 on Build GRCh38
rs963804014 Chr.11: 93154429-93154429 on Build GRCh38
rs899179427 Chr.11: 93154290-93154290 on Build GRCh38
rs996563979 Chr.11: 93154440-93154440 on Build GRCh38
rs995442517 Chr.11: 93154007-93154007 on Build GRCh38
rs747449159 Chr.11: 93154264-93154264 on Build GRCh38
rs577970283 Chr.11: 93154075-93154077 on Build GRCh38
rs991505407 Chr.11: 93154116-93154116 on Build GRCh38
rs78317190 Chr.11: 93154039-93154039 on Build GRCh38
rs775500413 Chr.11: 93154287-93154287 on Build GRCh38
rs144566090 Chr.11: 93154183-93154183 on Build GRCh38
rs1019869576 Chr.11: 93154382-93154382 on Build GRCh38
rs368158946 Chr.11: 93154132-93154132 on Build GRCh38
rs10765587 Chr.11: 93154475-93154475 on Build GRCh38
rs373793634 Chr.11: 93154161-93154161 on Build GRCh38
rs377056465 Chr.11: 93154167-93154167 on Build GRCh38
rs571852565 Chr.11: 93154390-93154390 on Build GRCh38
rs780415599 Chr.11: 93154460-93154461 on Build GRCh38
rs779218019 Chr.11: 93154376-93154376 on Build GRCh38
rs745427392 Chr.11: 93154157-93154157 on Build GRCh38
rs767252295 Chr.11: 93154222-93154222 on Build GRCh38
rs759502530 Chr.11: 93154110-93154110 on Build GRCh38
rs200947849 Chr.11: 93154279-93154279 on Build GRCh38
rs17854445 Chr.11: 93154188-93154188 on Build GRCh38
rs1011848928 Chr.11: 93154354-93154354 on Build GRCh38
rs769215034 Chr.11: 93154061-93154061 on Build GRCh38
rs762821688 Chr.11: 93154211-93154211 on Build GRCh38
rs776694196 Chr.11: 93154190-93154190 on Build GRCh38
rs758633779 Chr.11: 93154070-93154070 on Build GRCh38
rs759718601 Chr.11: 93154195-93154195 on Build GRCh38
rs765267539 Chr.11: 93154121-93154121 on Build GRCh38
rs768624002 Chr.11: 93154297-93154297 on Build GRCh38
rs766388493 Chr.11: 93154085-93154085 on Build GRCh38
rs186669390 Chr.11: 93154452-93154452 on Build GRCh38
rs765226022 Chr.11: 93154197-93154197 on Build GRCh38
rs199948847 Chr.11: 93154065-93154065 on Build GRCh38
rs147137520 Chr.11: 93154255-93154255 on Build GRCh38
rs967527761 Chr.11: 93154417-93154417 on Build GRCh38
rs191011114 Chr.11: 93154096-93154096 on Build GRCh38
rs1020927214 Chr.11: 93154358-93154358 on Build GRCh38
rs756885560 Chr.11: 93154146-93154146 on Build GRCh38
rs1025455045 Chr.11: 93154012-93154012 on Build GRCh38
rs751228107 Chr.11: 93154135-93154135 on Build GRCh38
rs17854444 Chr.11: 93154247-93154247 on Build GRCh38
rs968377842 Chr.11: 93154456-93154456 on Build GRCh38
rs761729198 Chr.11: 93154300-93154300 on Build GRCh38
rs975064018 Chr.11: 93154089-93154089 on Build GRCh38
rs755718601 Chr.11: 93154234-93154234 on Build GRCh38
rs762910285 Chr.11: 93154291-93154291 on Build GRCh38
rs546765758 Chr.11: 93154329-93154329 on Build GRCh38
rs748684438 Chr.11: 93154245-93154245 on Build GRCh38
rs375961741 Chr.11: 93154231-93154231 on Build GRCh38
rs540658503 Chr.11: 93154138-93154138 on Build GRCh38
rs772554735 Chr.11: 93154077-93154077 on Build GRCh38
rs371565740 Chr.11: 93154173-93154173 on Build GRCh38
rs758930081 Chr.11: 93154253-93154253 on Build GRCh38
rs767214107 Chr.11: 93154308-93154308 on Build GRCh38

View Full Product Details