Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 93479174-93479682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMCO4
Entrez Gene ID
56935
Gene Name
single-pass membrane protein with coiled-coil domains 4
Gene Aliases
C11orf75, FN5
UniGene
Hs.438064
Gene Chromosome Location
Chr.11: 93478472-93586856 on Build GRCh38
Transcripts
NM_020179.2 XM_011542907.1 XM_011542908.2 XM_011542909.1 XM_011542910.2 XM_011542911.2 XM_017018019.1 XM_017018020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5789518 c.5G>C p.R2P Chr.11: 93479185-93479185 on Build GRCh38
6866350 c.16G>T p.G6W Chr.11: 93479174-93479174 on Build GRCh38
3454689 c.12C>T p.L4L Chr.11: 93479178-93479178 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs557617596 Chr.11: 93479490-93479490 on Build GRCh38
rs760940483 Chr.11: 93479201-93479201 on Build GRCh38
rs964103123 Chr.11: 93479583-93479583 on Build GRCh38
rs918873487 Chr.11: 93479227-93479227 on Build GRCh38
rs929611991 Chr.11: 93479287-93479287 on Build GRCh38
rs947237048 Chr.11: 93479401-93479401 on Build GRCh38
rs117972535 Chr.11: 93479314-93479314 on Build GRCh38
rs188451881 Chr.11: 93479577-93479577 on Build GRCh38
rs763585208 Chr.11: 93479213-93479214 on Build GRCh38
rs903146761 Chr.11: 93479442-93479442 on Build GRCh38
rs987348105 Chr.11: 93479390-93479390 on Build GRCh38
rs1026194640 Chr.11: 93479360-93479360 on Build GRCh38
rs975927201 Chr.11: 93479431-93479431 on Build GRCh38
rs942705735 Chr.11: 93479419-93479419 on Build GRCh38
rs1002390736 Chr.11: 93479256-93479256 on Build GRCh38
rs912598993 Chr.11: 93479418-93479418 on Build GRCh38
rs754036634 Chr.11: 93479204-93479204 on Build GRCh38
rs954590912 Chr.11: 93479380-93479380 on Build GRCh38
rs947238309 Chr.11: 93479596-93479596 on Build GRCh38
rs996103203 Chr.11: 93479479-93479479 on Build GRCh38
rs528664953 Chr.11: 93479595-93479595 on Build GRCh38
rs892635803 Chr.11: 93479474-93479474 on Build GRCh38
rs80217712 Chr.11: 93479291-93479291 on Build GRCh38
rs1025900266 Chr.11: 93479487-93479487 on Build GRCh38
rs949649683 Chr.11: 93479548-93479548 on Build GRCh38
rs975176260 Chr.11: 93479585-93479585 on Build GRCh38
rs986424144 Chr.11: 93479507-93479507 on Build GRCh38
rs549244127 Chr.11: 93479213-93479213 on Build GRCh38
rs904851628 Chr.11: 93479211-93479211 on Build GRCh38
rs554750308 Chr.11: 93479578-93479578 on Build GRCh38
rs537227026 Chr.11: 93479209-93479209 on Build GRCh38
rs924497920 Chr.11: 93479642-93479642 on Build GRCh38
rs773626137 Chr.11: 93479198-93479198 on Build GRCh38
rs999474214 Chr.11: 93479453-93479453 on Build GRCh38
rs925368149 Chr.11: 93479450-93479450 on Build GRCh38
rs567754866 Chr.11: 93479285-93479285 on Build GRCh38
rs770664954 Chr.11: 93479514-93479514 on Build GRCh38
rs538930560 Chr.11: 93479456-93479456 on Build GRCh38
rs1041616852 Chr.11: 93479422-93479422 on Build GRCh38
rs962899968 Chr.11: 93479478-93479478 on Build GRCh38
rs765331872 Chr.11: 93479233-93479233 on Build GRCh38
rs938604187 Chr.11: 93479643-93479643 on Build GRCh38
rs182895693 Chr.11: 93479574-93479574 on Build GRCh38
rs540748286 Chr.11: 93479623-93479623 on Build GRCh38
rs7950036 Chr.11: 93479609-93479609 on Build GRCh38
rs1015994948 Chr.11: 93479282-93479282 on Build GRCh38
rs577989447 Chr.11: 93479373-93479373 on Build GRCh38
rs1052582728 Chr.11: 93479473-93479473 on Build GRCh38
rs558564789 Chr.11: 93479592-93479592 on Build GRCh38
rs912279818 Chr.11: 93479293-93479293 on Build GRCh38
rs1034993234 Chr.11: 93479455-93479455 on Build GRCh38
rs779314812 Chr.11: 93479203-93479203 on Build GRCh38

View Full Product Details