Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113334846-113335344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC12
Entrez Gene ID
54970
Gene Name
tetratricopeptide repeat domain 12
Gene Aliases
TPARM
UniGene
Hs.288772
Gene Chromosome Location
Chr.11: 113314529-113385952 on Build GRCh38
Transcripts
NM_001318533.1 NM_017868.3 XM_005271604.3 XM_011542890.2 XM_017017943.1 XM_017017944.1 XM_017017945.1 XM_017017946.1 XM_017017947.1 XM_017017948.1 XM_017017949.1 XM_017017950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3359016 c.513A>C p.E171D Chr.11: 113334974-113334974 on Build GRCh38
6552675 c.540G>T p.M180I Chr.11: 113335001-113335001 on Build GRCh38
7131129 c.559C>A p.L187M Chr.11: 113335020-113335020 on Build GRCh38
3359015 c.513A>C p.E171D Chr.11: 113334974-113334974 on Build GRCh38
6552674 c.540G>T p.M180I Chr.11: 113335001-113335001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977866006 Chr.11: 113335185-113335185 on Build GRCh38
rs782572665 Chr.11: 113335183-113335183 on Build GRCh38
rs190512713 Chr.11: 113335207-113335207 on Build GRCh38
rs782495751 Chr.11: 113334999-113334999 on Build GRCh38
rs889466444 Chr.11: 113335239-113335239 on Build GRCh38
rs782099906 Chr.11: 113334901-113334901 on Build GRCh38
rs782642907 Chr.11: 113334991-113334991 on Build GRCh38
rs782048390 Chr.11: 113334981-113334981 on Build GRCh38
rs564110226 Chr.11: 113335174-113335174 on Build GRCh38
rs76717283 Chr.11: 113334873-113334873 on Build GRCh38
rs782662099 Chr.11: 113335054-113335054 on Build GRCh38
rs111319463 Chr.11: 113335119-113335119 on Build GRCh38
rs567414850 Chr.11: 113335245-113335245 on Build GRCh38
rs993554770 Chr.11: 113334889-113334889 on Build GRCh38
rs943566851 Chr.11: 113335273-113335273 on Build GRCh38
rs952037652 Chr.11: 113335012-113335012 on Build GRCh38
rs782473431 Chr.11: 113335027-113335027 on Build GRCh38
rs782171368 Chr.11: 113335035-113335035 on Build GRCh38
rs782641338 Chr.11: 113334902-113334902 on Build GRCh38
rs529620893 Chr.11: 113335317-113335317 on Build GRCh38
rs148098336 Chr.11: 113335313-113335313 on Build GRCh38
rs544401862 Chr.11: 113335092-113335092 on Build GRCh38
rs747929536 Chr.11: 113334989-113334989 on Build GRCh38
rs782407631 Chr.11: 113334899-113334899 on Build GRCh38
rs902016365 Chr.11: 113334880-113334880 on Build GRCh38
rs782572401 Chr.11: 113335032-113335032 on Build GRCh38
rs782222623 Chr.11: 113334932-113334932 on Build GRCh38
rs782595119 Chr.11: 113335038-113335039 on Build GRCh38
rs781895156 Chr.11: 113334987-113334987 on Build GRCh38
rs1049399515 Chr.11: 113335226-113335226 on Build GRCh38
rs1026851990 Chr.11: 113334962-113334962 on Build GRCh38
rs925060091 Chr.11: 113335195-113335195 on Build GRCh38
rs371074508 Chr.11: 113334955-113334955 on Build GRCh38
rs936309162 Chr.11: 113335223-113335223 on Build GRCh38
rs1019082588 Chr.11: 113334876-113334876 on Build GRCh38
rs782003267 Chr.11: 113334900-113334900 on Build GRCh38
rs375480655 Chr.11: 113335086-113335086 on Build GRCh38
rs35559406 Chr.11: 113335060-113335061 on Build GRCh38
rs782420349 Chr.11: 113335052-113335052 on Build GRCh38
rs782815786 Chr.11: 113334983-113334983 on Build GRCh38
rs782721452 Chr.11: 113334943-113334943 on Build GRCh38
rs113759490 Chr.11: 113334877-113334877 on Build GRCh38
rs782374599 Chr.11: 113335083-113335083 on Build GRCh38
rs533254703 Chr.11: 113335163-113335163 on Build GRCh38
rs781830424 Chr.11: 113334999-113334999 on Build GRCh38
rs369095177 Chr.11: 113335065-113335065 on Build GRCh38

View Full Product Details