Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124116275-124116776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWA5A
Entrez Gene ID
4013
Gene Name
von Willebrand factor A domain containing 5A
Gene Aliases
BCSC-1, BCSC1, LOH11CR2A
UniGene
Hs.152944
Gene Chromosome Location
Chr.11: 124115404-124146912 on Build GRCh38
Transcripts
NM_001130142.1 NM_014622.4 NM_198315.2 XM_011542828.2

SNP Detail

SNP ID Chromosome Location
rs559953160 Chr.11: 124116711-124116711 on Build GRCh38
rs544948897 Chr.11: 124116585-124116585 on Build GRCh38
rs116322247 Chr.11: 124116574-124116574 on Build GRCh38
rs990550527 Chr.11: 124116753-124116753 on Build GRCh38
rs978196694 Chr.11: 124116723-124116723 on Build GRCh38
rs745596049 Chr.11: 124116312-124116312 on Build GRCh38
rs375460181 Chr.11: 124116693-124116693 on Build GRCh38
rs541749735 Chr.11: 124116689-124116689 on Build GRCh38
rs998213246 Chr.11: 124116554-124116554 on Build GRCh38
rs773124070 Chr.11: 124116749-124116749 on Build GRCh38
rs774850683 Chr.11: 124116515-124116515 on Build GRCh38
rs1056302728 Chr.11: 124116442-124116442 on Build GRCh38
rs113429696 Chr.11: 124116619-124116619 on Build GRCh38
rs992079671 Chr.11: 124116599-124116599 on Build GRCh38
rs61632268 Chr.11: 124116424-124116424 on Build GRCh38
rs977767632 Chr.11: 124116439-124116439 on Build GRCh38
rs1030229273 Chr.11: 124116705-124116705 on Build GRCh38
rs975939679 Chr.11: 124116388-124116388 on Build GRCh38
rs192988788 Chr.11: 124116643-124116643 on Build GRCh38
rs1039136326 Chr.11: 124116537-124116537 on Build GRCh38
rs552599421 Chr.11: 124116507-124116507 on Build GRCh38
rs925053805 Chr.11: 124116446-124116446 on Build GRCh38
rs567935935 Chr.11: 124116718-124116718 on Build GRCh38
rs918050931 Chr.11: 124116648-124116648 on Build GRCh38
rs1055740204 Chr.11: 124116535-124116535 on Build GRCh38
rs937964669 Chr.11: 124116425-124116426 on Build GRCh38
rs1032003180 Chr.11: 124116747-124116747 on Build GRCh38
rs534597376 Chr.11: 124116502-124116502 on Build GRCh38
rs1017865146 Chr.11: 124116675-124116675 on Build GRCh38
rs531260359 Chr.11: 124116543-124116543 on Build GRCh38
rs1047530857 Chr.11: 124116513-124116513 on Build GRCh38
rs900317158 Chr.11: 124116694-124116694 on Build GRCh38
rs536870743 Chr.11: 124116381-124116381 on Build GRCh38
rs988856394 Chr.11: 124116324-124116324 on Build GRCh38
rs551569663 Chr.11: 124116328-124116328 on Build GRCh38
rs567259020 Chr.11: 124116498-124116498 on Build GRCh38
rs188611477 Chr.11: 124116609-124116609 on Build GRCh38
rs372147587 Chr.11: 124116563-124116563 on Build GRCh38
rs1047625354 Chr.11: 124116714-124116714 on Build GRCh38
rs562256334 Chr.11: 124116640-124116640 on Build GRCh38
rs775402287 Chr.11: 124116359-124116359 on Build GRCh38
rs551721615 Chr.11: 124116729-124116729 on Build GRCh38
rs1030415792 Chr.11: 124116562-124116562 on Build GRCh38
rs760282081 Chr.11: 124116592-124116592 on Build GRCh38
rs916709468 Chr.11: 124116550-124116550 on Build GRCh38
rs929001325 Chr.11: 124116453-124116453 on Build GRCh38
rs563698286 Chr.11: 124116400-124116400 on Build GRCh38
rs763626842 Chr.11: 124116666-124116666 on Build GRCh38
rs549441148 Chr.11: 124116583-124116583 on Build GRCh38
rs997409742 Chr.11: 124116695-124116695 on Build GRCh38
rs956119234 Chr.11: 124116321-124116321 on Build GRCh38
rs888560025 Chr.11: 124116634-124116634 on Build GRCh38
rs929981887 Chr.11: 124116593-124116593 on Build GRCh38
rs1030497761 Chr.11: 124116303-124116303 on Build GRCh38
rs138233531 Chr.11: 124116591-124116591 on Build GRCh38
rs929137368 Chr.11: 124116699-124116699 on Build GRCh38

View Full Product Details