Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 948389-948897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOC2L
Entrez Gene ID
26155
Gene Name
NOC2 like nucleolar associated transcriptional repressor
Gene Aliases
NET15, NET7, NIR, PPP1R112
UniGene
Hs.405987
Gene Chromosome Location
Chr.1: 944203-959299 on Build GRCh38
Transcripts
NM_015658.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7086149 c.1528A>C p.N510H Chr.1: 948519-948519 on Build GRCh38
5353776 c.1464C>G p.F488L Chr.1: 948583-948583 on Build GRCh38
4010382 c.1531G>A p.V511I Chr.1: 948516-948516 on Build GRCh38
7100273 c.1524G>C p.L508L Chr.1: 948523-948523 on Build GRCh38
5847431 c.1510G>A p.V504M Chr.1: 948537-948537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74336521 Chr.1: 948722-948722 on Build GRCh38
rs376363823 Chr.1: 948731-948735 on Build GRCh38
rs72631890 Chr.1: 948519-948519 on Build GRCh38
rs190217509 Chr.1: 948640-948640 on Build GRCh38
rs960846925 Chr.1: 948432-948432 on Build GRCh38
rs764049727 Chr.1: 948535-948535 on Build GRCh38
rs765008445 Chr.1: 948448-948448 on Build GRCh38
rs897371797 Chr.1: 948706-948706 on Build GRCh38
rs751920109 Chr.1: 948527-948527 on Build GRCh38
rs7522415 Chr.1: 948711-948711 on Build GRCh38
rs978467584 Chr.1: 948833-948833 on Build GRCh38
rs746824727 Chr.1: 948559-948559 on Build GRCh38
rs764516117 Chr.1: 948469-948469 on Build GRCh38
rs375141658 Chr.1: 948480-948480 on Build GRCh38
rs750870295 Chr.1: 948539-948539 on Build GRCh38
rs755202760 Chr.1: 948551-948551 on Build GRCh38
rs887540874 Chr.1: 948741-948741 on Build GRCh38
rs375064108 Chr.1: 948497-948497 on Build GRCh38
rs142735174 Chr.1: 948712-948713 on Build GRCh38
rs74472614 Chr.1: 948764-948764 on Build GRCh38
rs1032202216 Chr.1: 948811-948811 on Build GRCh38
rs551347219 Chr.1: 948866-948866 on Build GRCh38
rs777608108 Chr.1: 948491-948491 on Build GRCh38
rs937035887 Chr.1: 948634-948634 on Build GRCh38
rs982074696 Chr.1: 948858-948858 on Build GRCh38
rs778965967 Chr.1: 948557-948557 on Build GRCh38
rs753985624 Chr.1: 948638-948638 on Build GRCh38
rs763232295 Chr.1: 948463-948463 on Build GRCh38
rs762655000 Chr.1: 948512-948512 on Build GRCh38
rs533482839 Chr.1: 948735-948735 on Build GRCh38
rs978350545 Chr.1: 948798-948798 on Build GRCh38
rs201792380 Chr.1: 948442-948442 on Build GRCh38
rs201640609 Chr.1: 948660-948660 on Build GRCh38
rs762123766 Chr.1: 948537-948537 on Build GRCh38
rs369566860 Chr.1: 948618-948618 on Build GRCh38
rs60085302 Chr.1: 948720-948721 on Build GRCh38
rs565042969 Chr.1: 948632-948632 on Build GRCh38
rs1026484897 Chr.1: 948435-948435 on Build GRCh38
rs767802924 Chr.1: 948612-948612 on Build GRCh38
rs775044682 Chr.1: 948588-948588 on Build GRCh38
rs1005990931 Chr.1: 948744-948744 on Build GRCh38
rs985495108 Chr.1: 948763-948763 on Build GRCh38
rs954015958 Chr.1: 948760-948760 on Build GRCh38
rs780173891 Chr.1: 948549-948549 on Build GRCh38
rs912514080 Chr.1: 948572-948572 on Build GRCh38
rs547000248 Chr.1: 948759-948759 on Build GRCh38
rs759079420 Chr.1: 948446-948446 on Build GRCh38
rs930188040 Chr.1: 948868-948868 on Build GRCh38
rs376660163 Chr.1: 948647-948657 on Build GRCh38
rs374853608 Chr.1: 948589-948589 on Build GRCh38
rs766417042 Chr.1: 948633-948633 on Build GRCh38
rs912942005 Chr.1: 948594-948594 on Build GRCh38
rs780152066 Chr.1: 948483-948483 on Build GRCh38
rs749455382 Chr.1: 948487-948487 on Build GRCh38
rs200896050 Chr.1: 948541-948541 on Build GRCh38
rs994336566 Chr.1: 948425-948425 on Build GRCh38
rs900209517 Chr.1: 948750-948750 on Build GRCh38
rs771825911 Chr.1: 948489-948489 on Build GRCh38
rs201749198 Chr.1: 948452-948452 on Build GRCh38
rs1006313366 Chr.1: 948468-948468 on Build GRCh38
rs183088415 Chr.1: 948745-948745 on Build GRCh38
rs757156333 Chr.1: 948562-948562 on Build GRCh38
rs958098149 Chr.1: 948817-948817 on Build GRCh38
rs112515528 Chr.1: 948478-948478 on Build GRCh38
rs544681080 Chr.1: 948714-948714 on Build GRCh38
rs4970455 Chr.1: 948721-948721 on Build GRCh38
rs965677348 Chr.1: 948804-948804 on Build GRCh38
rs988628541 Chr.1: 948818-948818 on Build GRCh38
rs966571518 Chr.1: 948775-948775 on Build GRCh38
rs139116730 Chr.1: 948538-948538 on Build GRCh38
rs765513018 Chr.1: 948648-948648 on Build GRCh38
rs559197331 Chr.1: 948566-948566 on Build GRCh38
rs143278708 Chr.1: 948503-948503 on Build GRCh38
rs140441976 Chr.1: 948614-948614 on Build GRCh38
rs970721451 Chr.1: 948854-948854 on Build GRCh38
rs1049103050 Chr.1: 948871-948871 on Build GRCh38
rs1019541936 Chr.1: 948772-948772 on Build GRCh38
rs371695354 Chr.1: 948580-948580 on Build GRCh38
rs773943074 Chr.1: 948597-948597 on Build GRCh38
rs775427240 Chr.1: 948711-948712 on Build GRCh38
rs190915384 Chr.1: 948864-948864 on Build GRCh38
rs372726801 Chr.1: 948496-948496 on Build GRCh38
rs896781922 Chr.1: 948656-948656 on Build GRCh38
rs111993795 Chr.1: 948670-948670 on Build GRCh38
rs199834703 Chr.1: 948567-948567 on Build GRCh38
rs1027097512 Chr.1: 948752-948752 on Build GRCh38
rs771025619 Chr.1: 948492-948492 on Build GRCh38
rs551786029 Chr.1: 948826-948826 on Build GRCh38
rs762179047 Chr.1: 948608-948608 on Build GRCh38
rs977862909 Chr.1: 948532-948532 on Build GRCh38
rs887499152 Chr.1: 948460-948460 on Build GRCh38
rs752905674 Chr.1: 948651-948651 on Build GRCh38
rs930135480 Chr.1: 948733-948733 on Build GRCh38
rs187594576 Chr.1: 948825-948825 on Build GRCh38
rs768385039 Chr.1: 948595-948595 on Build GRCh38
rs757741373 Chr.1: 948477-948477 on Build GRCh38
rs927078336 Chr.1: 948863-948863 on Build GRCh38

View Full Product Details