Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1442360-1442855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWA1
Entrez Gene ID
64856
Gene Name
von Willebrand factor A domain containing 1
Gene Aliases
WARP
UniGene
Hs.449009
Gene Chromosome Location
Chr.1: 1435523-1442882 on Build GRCh38
Transcripts
NM_022834.4 NM_199121.2

SNP Detail

SNP ID Chromosome Location
rs868761215 Chr.1: 1442596-1442596 on Build GRCh38
rs767737651 Chr.1: 1442785-1442786 on Build GRCh38
rs975571645 Chr.1: 1442745-1442745 on Build GRCh38
rs188333330 Chr.1: 1442628-1442628 on Build GRCh38
rs866139300 Chr.1: 1442478-1442478 on Build GRCh38
rs977437049 Chr.1: 1442792-1442792 on Build GRCh38
rs558408833 Chr.1: 1442495-1442495 on Build GRCh38
rs141752169 Chr.1: 1442589-1442589 on Build GRCh38
rs982632765 Chr.1: 1442558-1442558 on Build GRCh38
rs535182806 Chr.1: 1442447-1442447 on Build GRCh38
rs981748805 Chr.1: 1442548-1442548 on Build GRCh38
rs986882535 Chr.1: 1442597-1442597 on Build GRCh38
rs1045023912 Chr.1: 1442667-1442667 on Build GRCh38
rs974374501 Chr.1: 1442567-1442567 on Build GRCh38
rs962844268 Chr.1: 1442776-1442776 on Build GRCh38
rs962671278 Chr.1: 1442560-1442560 on Build GRCh38
rs751615593 Chr.1: 1442476-1442476 on Build GRCh38
rs566389076 Chr.1: 1442436-1442436 on Build GRCh38
rs1053667632 Chr.1: 1442799-1442799 on Build GRCh38
rs867732675 Chr.1: 1442559-1442559 on Build GRCh38
rs570528584 Chr.1: 1442501-1442501 on Build GRCh38
rs904887481 Chr.1: 1442707-1442707 on Build GRCh38
rs570064331 Chr.1: 1442414-1442414 on Build GRCh38
rs970326209 Chr.1: 1442543-1442543 on Build GRCh38
rs371328806 Chr.1: 1442568-1442568 on Build GRCh38
rs764118211 Chr.1: 1442423-1442423 on Build GRCh38
rs769518287 Chr.1: 1442444-1442444 on Build GRCh38
rs574030859 Chr.1: 1442593-1442593 on Build GRCh38
rs893263110 Chr.1: 1442625-1442625 on Build GRCh38
rs150280246 Chr.1: 1442419-1442419 on Build GRCh38
rs1004341262 Chr.1: 1442742-1442742 on Build GRCh38
rs545361013 Chr.1: 1442803-1442803 on Build GRCh38
rs183674208 Chr.1: 1442604-1442604 on Build GRCh38
rs533250540 Chr.1: 1442729-1442729 on Build GRCh38
rs893393173 Chr.1: 1442442-1442442 on Build GRCh38
rs1001861205 Chr.1: 1442708-1442708 on Build GRCh38
rs932757036 Chr.1: 1442415-1442415 on Build GRCh38
rs555202281 Chr.1: 1442616-1442616 on Build GRCh38
rs975563196 Chr.1: 1442590-1442590 on Build GRCh38
rs549870854 Chr.1: 1442389-1442389 on Build GRCh38
rs922709555 Chr.1: 1442765-1442765 on Build GRCh38
rs999739516 Chr.1: 1442446-1442446 on Build GRCh38
rs711180 Chr.1: 1442793-1442793 on Build GRCh38
rs138874345 Chr.1: 1442422-1442422 on Build GRCh38
rs934824025 Chr.1: 1442433-1442433 on Build GRCh38
rs755965625 Chr.1: 1442614-1442614 on Build GRCh38
rs35653771 Chr.1: 1442457-1442457 on Build GRCh38
rs759437279 Chr.1: 1442808-1442808 on Build GRCh38
rs1054017406 Chr.1: 1442607-1442607 on Build GRCh38
rs374272210 Chr.1: 1442388-1442388 on Build GRCh38
rs909931872 Chr.1: 1442395-1442395 on Build GRCh38
rs1015446398 Chr.1: 1442746-1442746 on Build GRCh38
rs147051399 Chr.1: 1442617-1442617 on Build GRCh38
rs758950480 Chr.1: 1442393-1442393 on Build GRCh38
rs935251595 Chr.1: 1442794-1442794 on Build GRCh38
rs934043718 Chr.1: 1442779-1442779 on Build GRCh38
rs896239981 Chr.1: 1442731-1442731 on Build GRCh38
rs1021971817 Chr.1: 1442494-1442494 on Build GRCh38
rs772744562 Chr.1: 1442482-1442482 on Build GRCh38
rs542921350 Chr.1: 1442661-1442661 on Build GRCh38
rs1023252831 Chr.1: 1442542-1442542 on Build GRCh38
rs902409432 Chr.1: 1442602-1442602 on Build GRCh38
rs942821602 Chr.1: 1442398-1442398 on Build GRCh38
rs539556081 Chr.1: 1442551-1442551 on Build GRCh38
rs1025286676 Chr.1: 1442681-1442681 on Build GRCh38
rs986768424 Chr.1: 1442828-1442828 on Build GRCh38
rs951732017 Chr.1: 1442697-1442697 on Build GRCh38
rs995466534 Chr.1: 1442784-1442784 on Build GRCh38
rs1034753344 Chr.1: 1442546-1442546 on Build GRCh38
rs112270823 Chr.1: 1442722-1442722 on Build GRCh38
rs541748357 Chr.1: 1442606-1442606 on Build GRCh38
rs569791493 Chr.1: 1442396-1442396 on Build GRCh38
rs1001975802 Chr.1: 1442515-1442515 on Build GRCh38

View Full Product Details