Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 130123492-130123989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APLP2
Entrez Gene ID
334
Gene Name
amyloid beta precursor like protein 2
Gene Aliases
APLP-2, APPH, APPL2, CDEBP
UniGene
Hs.370247
Gene Chromosome Location
Chr.11: 130069821-130144811 on Build GRCh38
Transcripts
NM_001142276.1 NM_001142277.1 NM_001142278.1 NM_001243299.1 NM_001642.2 NR_024515.1 NR_024516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1475263 c.1005C>T p.C335C Chr.11: 130123694-130123694 on Build GRCh38
5925547 c.1090+8C>T p.? Chr.11: 130123787-130123787 on Build GRCh38
4019334 c.941C>T p.A314V Chr.11: 130123630-130123630 on Build GRCh38
329143 c.967A>T p.M323L Chr.11: 130123656-130123656 on Build GRCh38
7197134 c.978G>C p.W326C Chr.11: 130123667-130123667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527689416 Chr.11: 130123851-130123851 on Build GRCh38
rs758568357 Chr.11: 130123795-130123795 on Build GRCh38
rs546391563 Chr.11: 130123793-130123793 on Build GRCh38
rs758428295 Chr.11: 130123648-130123648 on Build GRCh38
rs752399273 Chr.11: 130123827-130123827 on Build GRCh38
rs1006301004 Chr.11: 130123888-130123888 on Build GRCh38
rs930593511 Chr.11: 130123527-130123527 on Build GRCh38
rs747515478 Chr.11: 130123800-130123800 on Build GRCh38
rs754779373 Chr.11: 130123700-130123700 on Build GRCh38
rs748257586 Chr.11: 130123618-130123618 on Build GRCh38
rs945259538 Chr.11: 130123574-130123574 on Build GRCh38
rs117994299 Chr.11: 130123658-130123658 on Build GRCh38
rs772688189 Chr.11: 130123677-130123677 on Build GRCh38
rs759443645 Chr.11: 130123822-130123822 on Build GRCh38
rs764649597 Chr.11: 130123695-130123695 on Build GRCh38
rs3886744 Chr.11: 130123529-130123529 on Build GRCh38
rs372558282 Chr.11: 130123818-130123818 on Build GRCh38
rs763775520 Chr.11: 130123686-130123686 on Build GRCh38
rs774730310 Chr.11: 130123609-130123609 on Build GRCh38
rs760384969 Chr.11: 130123636-130123636 on Build GRCh38
rs774461685 Chr.11: 130123963-130123963 on Build GRCh38
rs756725992 Chr.11: 130123645-130123645 on Build GRCh38
rs983607664 Chr.11: 130123514-130123514 on Build GRCh38
rs376424694 Chr.11: 130123808-130123808 on Build GRCh38
rs539623328 Chr.11: 130123567-130123567 on Build GRCh38
rs913415950 Chr.11: 130123968-130123968 on Build GRCh38
rs766585889 Chr.11: 130123805-130123805 on Build GRCh38
rs141638912 Chr.11: 130123663-130123663 on Build GRCh38
rs542688342 Chr.11: 130123699-130123699 on Build GRCh38
rs773227254 Chr.11: 130123599-130123599 on Build GRCh38
rs776149533 Chr.11: 130123730-130123730 on Build GRCh38
rs541085939 Chr.11: 130123721-130123721 on Build GRCh38
rs771147161 Chr.11: 130123719-130123719 on Build GRCh38
rs1019399725 Chr.11: 130123571-130123571 on Build GRCh38
rs771399225 Chr.11: 130123600-130123600 on Build GRCh38
rs900550488 Chr.11: 130123629-130123629 on Build GRCh38
rs577335648 Chr.11: 130123758-130123758 on Build GRCh38
rs761155845 Chr.11: 130123683-130123683 on Build GRCh38
rs773887845 Chr.11: 130123821-130123821 on Build GRCh38
rs767468024 Chr.11: 130123823-130123823 on Build GRCh38
rs949724035 Chr.11: 130123772-130123772 on Build GRCh38
rs374272849 Chr.11: 130123568-130123568 on Build GRCh38
rs375624657 Chr.11: 130123775-130123775 on Build GRCh38
rs959303108 Chr.11: 130123546-130123546 on Build GRCh38
rs752582093 Chr.11: 130123777-130123777 on Build GRCh38
rs748366525 Chr.11: 130123593-130123593 on Build GRCh38
rs375730738 Chr.11: 130123674-130123674 on Build GRCh38
rs113352584 Chr.11: 130123780-130123780 on Build GRCh38
rs935786924 Chr.11: 130123954-130123954 on Build GRCh38
rs761341617 Chr.11: 130123749-130123749 on Build GRCh38
rs758218836 Chr.11: 130123697-130123697 on Build GRCh38
rs373556814 Chr.11: 130123794-130123794 on Build GRCh38
rs369336732 Chr.11: 130123698-130123698 on Build GRCh38
rs757376095 Chr.11: 130123785-130123785 on Build GRCh38
rs753157155 Chr.11: 130123631-130123631 on Build GRCh38
rs777927148 Chr.11: 130123673-130123673 on Build GRCh38
rs760143269 Chr.11: 130123623-130123623 on Build GRCh38
rs1002151768 Chr.11: 130123868-130123868 on Build GRCh38
rs753361132 Chr.11: 130123641-130123641 on Build GRCh38
rs753559526 Chr.11: 130123687-130123687 on Build GRCh38
rs767265271 Chr.11: 130123776-130123776 on Build GRCh38
rs966370547 Chr.11: 130123913-130123913 on Build GRCh38
rs751481974 Chr.11: 130123768-130123768 on Build GRCh38
rs775542519 Chr.11: 130123517-130123517 on Build GRCh38
rs796527462 Chr.11: 130123850-130123850 on Build GRCh38
rs777973673 Chr.11: 130123659-130123659 on Build GRCh38
rs766245157 Chr.11: 130123763-130123763 on Build GRCh38
rs1380078 Chr.11: 130123541-130123541 on Build GRCh38
rs778749333 Chr.11: 130123589-130123589 on Build GRCh38
rs769264332 Chr.11: 130123814-130123814 on Build GRCh38
rs763863958 Chr.11: 130123637-130123637 on Build GRCh38
rs3886745 Chr.11: 130123537-130123537 on Build GRCh38
rs201772983 Chr.11: 130123804-130123804 on Build GRCh38
rs369687086 Chr.11: 130123816-130123816 on Build GRCh38
rs372840531 Chr.11: 130123932-130123932 on Build GRCh38
rs753416938 Chr.11: 130123673-130123673 on Build GRCh38
rs557699633 Chr.11: 130123538-130123538 on Build GRCh38
rs944500682 Chr.11: 130123616-130123616 on Build GRCh38
rs762569937 Chr.11: 130123817-130123817 on Build GRCh38
rs369478296 Chr.11: 130123787-130123787 on Build GRCh38
rs779691092 Chr.11: 130123652-130123652 on Build GRCh38
rs759981266 Chr.11: 130123586-130123587 on Build GRCh38
rs747389828 Chr.11: 130123676-130123676 on Build GRCh38
rs146199947 Chr.11: 130123740-130123740 on Build GRCh38
rs576304274 Chr.11: 130123662-130123662 on Build GRCh38
rs961217477 Chr.11: 130123898-130123898 on Build GRCh38
rs989155736 Chr.11: 130123550-130123550 on Build GRCh38
rs761586315 Chr.11: 130123694-130123694 on Build GRCh38
rs76370121 Chr.11: 130123718-130123718 on Build GRCh38
rs911773425 Chr.11: 130123743-130123743 on Build GRCh38
rs748860332 Chr.11: 130123578-130123578 on Build GRCh38
rs771778673 Chr.11: 130123591-130123591 on Build GRCh38
rs777059296 Chr.11: 130123747-130123747 on Build GRCh38
rs185801556 Chr.11: 130123563-130123563 on Build GRCh38
rs764964482 Chr.11: 130123757-130123757 on Build GRCh38
rs1001674902 Chr.11: 130123672-130123672 on Build GRCh38
rs775951729 Chr.11: 130123682-130123682 on Build GRCh38
rs367821777 Chr.11: 130123630-130123630 on Build GRCh38
rs750314819 Chr.11: 130123647-130123647 on Build GRCh38
rs757214865 Chr.11: 130123582-130123582 on Build GRCh38
rs916924531 Chr.11: 130123761-130123761 on Build GRCh38
rs78611322 Chr.11: 130123928-130123928 on Build GRCh38
rs1010670260 Chr.11: 130123895-130123895 on Build GRCh38
rs989847502 Chr.11: 130123964-130123964 on Build GRCh38
rs779958252 Chr.11: 130123592-130123592 on Build GRCh38
rs768001672 Chr.11: 130123592-130123593 on Build GRCh38
rs749503671 Chr.11: 130123713-130123713 on Build GRCh38
rs772421840 Chr.11: 130123722-130123722 on Build GRCh38
rs115641832 Chr.11: 130123876-130123876 on Build GRCh38
rs959414642 Chr.11: 130123883-130123883 on Build GRCh38
rs777390678 Chr.11: 130123575-130123575 on Build GRCh38
rs745612718 Chr.11: 130123590-130123590 on Build GRCh38
rs1022031022 Chr.11: 130123902-130123902 on Build GRCh38
rs376905429 Chr.11: 130123830-130123830 on Build GRCh38
rs1044107320 Chr.11: 130123536-130123536 on Build GRCh38
rs372001669 Chr.11: 130123735-130123735 on Build GRCh38
rs891956762 Chr.11: 130123809-130123809 on Build GRCh38
rs144814874 Chr.11: 130123710-130123710 on Build GRCh38
rs1016805200 Chr.11: 130123897-130123897 on Build GRCh38
rs371634970 Chr.11: 130123653-130123653 on Build GRCh38
rs561786130 Chr.11: 130123833-130123833 on Build GRCh38

View Full Product Details