Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134276549-134277046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1L3
Entrez Gene ID
112937
Gene Name
galactosidase beta 1 like 3
Gene Aliases
-
UniGene
Hs.721382
Gene Chromosome Location
Chr.11: 134275447-134321364 on Build GRCh38
Transcripts
NM_001080407.2 XM_011542566.2 XM_011542567.2 XM_011542568.2 XM_011542569.2 XM_011542570.2 XM_011542571.2 XM_017017155.1 XM_017017156.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6418649 c.16C>T p.L6F Chr.11: 134276756-134276756 on Build GRCh38
6418648 c.16C>T p.L6F Chr.11: 134276756-134276756 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs938114375 Chr.11: 134276574-134276574 on Build GRCh38
rs573342818 Chr.11: 134276932-134276932 on Build GRCh38
rs902492520 Chr.11: 134276824-134276824 on Build GRCh38
rs990873191 Chr.11: 134276850-134276850 on Build GRCh38
rs867913990 Chr.11: 134276782-134276782 on Build GRCh38
rs936849073 Chr.11: 134276653-134276653 on Build GRCh38
rs995848475 Chr.11: 134276720-134276720 on Build GRCh38
rs1019504013 Chr.11: 134276623-134276623 on Build GRCh38
rs927004569 Chr.11: 134276921-134276922 on Build GRCh38
rs965303731 Chr.11: 134276886-134276886 on Build GRCh38
rs983789311 Chr.11: 134276917-134276917 on Build GRCh38
rs924216279 Chr.11: 134276889-134276889 on Build GRCh38
rs900682307 Chr.11: 134276716-134276716 on Build GRCh38
rs865797612 Chr.11: 134276650-134276650 on Build GRCh38
rs145395751 Chr.11: 134276586-134276586 on Build GRCh38
rs949681933 Chr.11: 134276805-134276805 on Build GRCh38
rs949601804 Chr.11: 134276998-134276998 on Build GRCh38
rs932171493 Chr.11: 134276922-134276922 on Build GRCh38
rs530949285 Chr.11: 134276651-134276651 on Build GRCh38
rs910604262 Chr.11: 134276826-134276826 on Build GRCh38
rs890105114 Chr.11: 134276748-134276748 on Build GRCh38
rs759972510 Chr.11: 134276698-134276698 on Build GRCh38
rs970741974 Chr.11: 134276881-134276881 on Build GRCh38
rs936881982 Chr.11: 134276896-134276896 on Build GRCh38
rs994721045 Chr.11: 134277024-134277024 on Build GRCh38
rs1001374942 Chr.11: 134276568-134276568 on Build GRCh38
rs889877368 Chr.11: 134276851-134276851 on Build GRCh38
rs898832332 Chr.11: 134277022-134277022 on Build GRCh38
rs978533431 Chr.11: 134276882-134276882 on Build GRCh38
rs918267890 Chr.11: 134276669-134276669 on Build GRCh38
rs1036476864 Chr.11: 134276996-134276996 on Build GRCh38
rs994677366 Chr.11: 134276803-134276803 on Build GRCh38
rs958532971 Chr.11: 134276849-134276849 on Build GRCh38
rs181828569 Chr.11: 134276589-134276589 on Build GRCh38
rs918022654 Chr.11: 134276873-134276873 on Build GRCh38
rs1004544291 Chr.11: 134276995-134276995 on Build GRCh38
rs190388497 Chr.11: 134276939-134276939 on Build GRCh38
rs768069662 Chr.11: 134276752-134276752 on Build GRCh38
rs949426022 Chr.11: 134276625-134276625 on Build GRCh38
rs1053973086 Chr.11: 134276914-134276914 on Build GRCh38
rs753251266 Chr.11: 134276802-134276802 on Build GRCh38
rs563422070 Chr.11: 134276649-134276649 on Build GRCh38
rs890229935 Chr.11: 134276966-134276966 on Build GRCh38
rs1042628276 Chr.11: 134276643-134276643 on Build GRCh38
rs1030052488 Chr.11: 134276722-134276722 on Build GRCh38
rs1042367267 Chr.11: 134276809-134276809 on Build GRCh38
rs370437749 Chr.11: 134276936-134276936 on Build GRCh38
rs990919736 Chr.11: 134276988-134276988 on Build GRCh38
rs1043025914 Chr.11: 134277001-134277001 on Build GRCh38
rs185973625 Chr.11: 134276915-134276915 on Build GRCh38
rs1000909204 Chr.11: 134276825-134276825 on Build GRCh38
rs896708427 Chr.11: 134276784-134276784 on Build GRCh38
rs933670844 Chr.11: 134276845-134276873 on Build GRCh38
rs1025591145 Chr.11: 134276900-134276904 on Build GRCh38
rs1053922111 Chr.11: 134276665-134276665 on Build GRCh38
rs985331263 Chr.11: 134276608-134276608 on Build GRCh38
rs1034143382 Chr.11: 134276834-134276834 on Build GRCh38
rs1004978893 Chr.11: 134276773-134276773 on Build GRCh38

View Full Product Details