Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4548609-4549121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD51AP1
Entrez Gene ID
10635
Gene Name
RAD51 associated protein 1
Gene Aliases
PIR51
UniGene
Hs.730696
Gene Chromosome Location
Chr.12: 4538784-4560047 on Build GRCh38
Transcripts
NM_001130862.1 NM_006479.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6136951 c.538G>C p.E180Q Chr.12: 4548818-4548818 on Build GRCh38
4719969 c.98G>A p.G33E Chr.12: 4548732-4548732 on Build GRCh38
6136954 c.508G>T p.G170C Chr.12: 4548788-4548788 on Build GRCh38
5361949 c.463G>A p.A155T Chr.12: 4548743-4548743 on Build GRCh38
6136950 c.589G>C p.E197Q Chr.12: 4548818-4548818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952354013 Chr.12: 4548980-4548980 on Build GRCh38
rs998737044 Chr.12: 4548894-4548894 on Build GRCh38
rs746369712 Chr.12: 4548888-4548888 on Build GRCh38
rs753513632 Chr.12: 4548689-4548689 on Build GRCh38
rs750561718 Chr.12: 4548969-4548969 on Build GRCh38
rs750415596 Chr.12: 4548701-4548701 on Build GRCh38
rs200919420 Chr.12: 4548830-4548830 on Build GRCh38
rs965597677 Chr.12: 4549087-4549090 on Build GRCh38
rs774592035 Chr.12: 4548866-4548866 on Build GRCh38
rs193272333 Chr.12: 4548882-4548882 on Build GRCh38
rs761684492 Chr.12: 4548692-4548692 on Build GRCh38
rs912625690 Chr.12: 4549070-4549070 on Build GRCh38
rs763949301 Chr.12: 4548688-4548688 on Build GRCh38
rs762064267 Chr.12: 4548938-4548938 on Build GRCh38
rs772127456 Chr.12: 4548767-4548767 on Build GRCh38
rs768465622 Chr.12: 4548838-4548839 on Build GRCh38
rs773258503 Chr.12: 4548826-4548826 on Build GRCh38
rs754529148 Chr.12: 4548842-4548842 on Build GRCh38
rs147167939 Chr.12: 4548710-4548710 on Build GRCh38
rs201976110 Chr.12: 4548758-4548758 on Build GRCh38
rs144906332 Chr.12: 4548798-4548798 on Build GRCh38
rs116552921 Chr.12: 4548760-4548760 on Build GRCh38
rs765219194 Chr.12: 4548696-4548696 on Build GRCh38
rs542362221 Chr.12: 4548729-4548729 on Build GRCh38
rs771305807 Chr.12: 4548755-4548755 on Build GRCh38
rs754653104 Chr.12: 4548717-4548717 on Build GRCh38
rs983746626 Chr.12: 4548995-4548995 on Build GRCh38
rs201787958 Chr.12: 4548850-4548850 on Build GRCh38
rs991240814 Chr.12: 4548968-4548968 on Build GRCh38
rs375466989 Chr.12: 4548995-4548996 on Build GRCh38
rs532505251 Chr.12: 4548812-4548812 on Build GRCh38
rs765982514 Chr.12: 4548833-4548833 on Build GRCh38
rs141200406 Chr.12: 4549006-4549006 on Build GRCh38
rs977429771 Chr.12: 4549024-4549024 on Build GRCh38
rs376241140 Chr.12: 4548680-4548680 on Build GRCh38
rs752436723 Chr.12: 4548855-4548855 on Build GRCh38
rs779755530 Chr.12: 4548713-4548713 on Build GRCh38
rs116277003 Chr.12: 4549007-4549007 on Build GRCh38
rs775499438 Chr.12: 4548769-4548769 on Build GRCh38
rs527550915 Chr.12: 4548987-4548987 on Build GRCh38
rs114193736 Chr.12: 4548716-4548716 on Build GRCh38
rs7968517 Chr.12: 4549044-4549044 on Build GRCh38
rs374106347 Chr.12: 4548663-4548663 on Build GRCh38
rs141058015 Chr.12: 4548787-4548787 on Build GRCh38
rs75806977 Chr.12: 4548952-4548952 on Build GRCh38
rs932843875 Chr.12: 4549053-4549053 on Build GRCh38
rs757596580 Chr.12: 4548856-4548856 on Build GRCh38
rs765161532 Chr.12: 4548815-4548815 on Build GRCh38
rs1049830264 Chr.12: 4549063-4549063 on Build GRCh38
rs1021495474 Chr.12: 4548860-4548860 on Build GRCh38
rs780901436 Chr.12: 4548720-4548720 on Build GRCh38
rs768706534 Chr.12: 4548783-4548783 on Build GRCh38
rs750967468 Chr.12: 4548837-4548837 on Build GRCh38
rs760818542 Chr.12: 4548772-4548772 on Build GRCh38
rs951461262 Chr.12: 4549045-4549045 on Build GRCh38

View Full Product Details