Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4556379-4556880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD51AP1
Entrez Gene ID
10635
Gene Name
RAD51 associated protein 1
Gene Aliases
PIR51
UniGene
Hs.730696
Gene Chromosome Location
Chr.12: 4538784-4560047 on Build GRCh38
Transcripts
NM_001130862.1 NM_006479.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6072620 c.858T>C p.T286T Chr.12: 4556438-4556438 on Build GRCh38
4207744 c.839C>A p.S280Y Chr.12: 4556419-4556419 on Build GRCh38
2068073 c.827G>A p.R276H Chr.12: 4556458-4556458 on Build GRCh38
6761908 c.863T>C p.V288A Chr.12: 4556494-4556494 on Build GRCh38
6072622 c.453T>C p.T151T Chr.12: 4556438-4556438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532396364 Chr.12: 4556660-4556660 on Build GRCh38
rs931315977 Chr.12: 4556682-4556682 on Build GRCh38
rs565128425 Chr.12: 4556838-4556838 on Build GRCh38
rs750887868 Chr.12: 4556405-4556405 on Build GRCh38
rs776117003 Chr.12: 4556488-4556488 on Build GRCh38
rs761089612 Chr.12: 4556496-4556498 on Build GRCh38
rs978422174 Chr.12: 4556483-4556483 on Build GRCh38
rs772760833 Chr.12: 4556479-4556479 on Build GRCh38
rs750656319 Chr.12: 4556555-4556555 on Build GRCh38
rs776188063 Chr.12: 4556462-4556462 on Build GRCh38
rs1015885446 Chr.12: 4556444-4556444 on Build GRCh38
rs768138700 Chr.12: 4556460-4556460 on Build GRCh38
rs747612735 Chr.12: 4556464-4556464 on Build GRCh38
rs778902378 Chr.12: 4556779-4556779 on Build GRCh38
rs915216543 Chr.12: 4556709-4556709 on Build GRCh38
rs954105723 Chr.12: 4556615-4556615 on Build GRCh38
rs917116827 Chr.12: 4556536-4556536 on Build GRCh38
rs761332048 Chr.12: 4556493-4556493 on Build GRCh38
rs757307589 Chr.12: 4556544-4556544 on Build GRCh38
rs948563245 Chr.12: 4556612-4556612 on Build GRCh38
rs756707430 Chr.12: 4556429-4556429 on Build GRCh38
rs753897389 Chr.12: 4556506-4556506 on Build GRCh38
rs968111729 Chr.12: 4556723-4556723 on Build GRCh38
rs780440734 Chr.12: 4556710-4556710 on Build GRCh38
rs758170511 Chr.12: 4556564-4556564 on Build GRCh38
rs1048345389 Chr.12: 4556780-4556780 on Build GRCh38
rs138705865 Chr.12: 4556458-4556458 on Build GRCh38
rs764576072 Chr.12: 4556598-4556600 on Build GRCh38
rs776047640 Chr.12: 4556472-4556473 on Build GRCh38
rs979086183 Chr.12: 4556819-4556819 on Build GRCh38
rs755117064 Chr.12: 4556773-4556773 on Build GRCh38
rs552653953 Chr.12: 4556676-4556676 on Build GRCh38
rs368957431 Chr.12: 4556457-4556457 on Build GRCh38
rs759904187 Chr.12: 4556482-4556482 on Build GRCh38
rs756342451 Chr.12: 4556607-4556607 on Build GRCh38
rs745631261 Chr.12: 4556424-4556424 on Build GRCh38
rs779885571 Chr.12: 4556571-4556571 on Build GRCh38
rs769425735 Chr.12: 4556474-4556474 on Build GRCh38
rs887108825 Chr.12: 4556806-4556806 on Build GRCh38
rs201243183 Chr.12: 4556667-4556668 on Build GRCh38
rs1044724332 Chr.12: 4556624-4556624 on Build GRCh38
rs765463201 Chr.12: 4556551-4556551 on Build GRCh38
rs976855388 Chr.12: 4556534-4556534 on Build GRCh38
rs375762547 Chr.12: 4556415-4556415 on Build GRCh38
rs771799015 Chr.12: 4556455-4556455 on Build GRCh38
rs753813759 Chr.12: 4556738-4556738 on Build GRCh38
rs768031221 Chr.12: 4556484-4556484 on Build GRCh38
rs371887719 Chr.12: 4556503-4556503 on Build GRCh38

View Full Product Details