Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6848742-6849249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA3
Entrez Gene ID
83461
Gene Name
cell division cycle associated 3
Gene Aliases
GRCC8, TOME-1
UniGene
Hs.524216
Gene Chromosome Location
Chr.12: 6844796-6852066 on Build GRCh38
Transcripts
NM_001297602.1 NM_001297603.1 NM_001297604.1 NM_031299.5 NR_123727.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5987175 c.713G>T p.G238V Chr.12: 6849137-6849137 on Build GRCh38
7360428 c.667C>G p.P223A Chr.12: 6849183-6849183 on Build GRCh38
4518730 c.774_775GG>AA p.E259K Chr.12: 6849075-6849076 on Build GRCh38
4559355 c.801G>C p.E267D Chr.12: 6849049-6849049 on Build GRCh38
1628801 c.798G>A p.V266V Chr.12: 6849052-6849052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782721788 Chr.12: 6849123-6849123 on Build GRCh38
rs1042440931 Chr.12: 6848892-6848892 on Build GRCh38
rs782573750 Chr.12: 6848909-6848909 on Build GRCh38
rs115017035 Chr.12: 6848833-6848833 on Build GRCh38
rs782761210 Chr.12: 6848992-6848992 on Build GRCh38
rs782265791 Chr.12: 6849049-6849049 on Build GRCh38
rs149041953 Chr.12: 6849092-6849092 on Build GRCh38
rs1003545766 Chr.12: 6848991-6848991 on Build GRCh38
rs375691603 Chr.12: 6848803-6848803 on Build GRCh38
rs782020608 Chr.12: 6849180-6849180 on Build GRCh38
rs61733172 Chr.12: 6849093-6849093 on Build GRCh38
rs201026595 Chr.12: 6849014-6849014 on Build GRCh38
rs782375921 Chr.12: 6849052-6849052 on Build GRCh38
rs908320581 Chr.12: 6848787-6848787 on Build GRCh38
rs906635837 Chr.12: 6848918-6848918 on Build GRCh38
rs782164674 Chr.12: 6849109-6849109 on Build GRCh38
rs11064428 Chr.12: 6849097-6849097 on Build GRCh38
rs539746123 Chr.12: 6848940-6848941 on Build GRCh38
rs782671696 Chr.12: 6849032-6849032 on Build GRCh38
rs781909731 Chr.12: 6849000-6849000 on Build GRCh38
rs782515898 Chr.12: 6849134-6849134 on Build GRCh38
rs782335077 Chr.12: 6849014-6849015 on Build GRCh38
rs782469981 Chr.12: 6849148-6849148 on Build GRCh38
rs782713955 Chr.12: 6849183-6849183 on Build GRCh38
rs781828454 Chr.12: 6848993-6848993 on Build GRCh38
rs782285838 Chr.12: 6848807-6848807 on Build GRCh38
rs782328318 Chr.12: 6849089-6849089 on Build GRCh38
rs781814047 Chr.12: 6849222-6849222 on Build GRCh38
rs148172476 Chr.12: 6849192-6849192 on Build GRCh38
rs760802476 Chr.12: 6848942-6848942 on Build GRCh38
rs781943073 Chr.12: 6848972-6848972 on Build GRCh38
rs782072618 Chr.12: 6848995-6848995 on Build GRCh38
rs781965657 Chr.12: 6849061-6849061 on Build GRCh38
rs782638944 Chr.12: 6849140-6849140 on Build GRCh38
rs782173096 Chr.12: 6849025-6849025 on Build GRCh38
rs3180416 Chr.12: 6848827-6848827 on Build GRCh38
rs782067953 Chr.12: 6849193-6849193 on Build GRCh38
rs943698607 Chr.12: 6848788-6848788 on Build GRCh38
rs782233142 Chr.12: 6849144-6849144 on Build GRCh38
rs369606193 Chr.12: 6849191-6849191 on Build GRCh38
rs781885050 Chr.12: 6849133-6849133 on Build GRCh38
rs782593230 Chr.12: 6849019-6849019 on Build GRCh38
rs367669653 Chr.12: 6849008-6849008 on Build GRCh38
rs113760877 Chr.12: 6848998-6848998 on Build GRCh38
rs995062589 Chr.12: 6849028-6849028 on Build GRCh38
rs782182693 Chr.12: 6849168-6849168 on Build GRCh38
rs781857226 Chr.12: 6849207-6849207 on Build GRCh38
rs781881385 Chr.12: 6848786-6848786 on Build GRCh38
rs782030784 Chr.12: 6849197-6849197 on Build GRCh38
rs149921153 Chr.12: 6849113-6849113 on Build GRCh38
rs945196003 Chr.12: 6849209-6849209 on Build GRCh38
rs781931240 Chr.12: 6849179-6849179 on Build GRCh38
rs201794409 Chr.12: 6849013-6849013 on Build GRCh38
rs1042243704 Chr.12: 6849210-6849210 on Build GRCh38
rs782627537 Chr.12: 6849012-6849012 on Build GRCh38
rs200884660 Chr.12: 6849128-6849128 on Build GRCh38
rs782594919 Chr.12: 6849149-6849149 on Build GRCh38
rs782699279 Chr.12: 6849194-6849194 on Build GRCh38
rs782385664 Chr.12: 6849027-6849027 on Build GRCh38
rs893619064 Chr.12: 6848860-6848860 on Build GRCh38
rs372805520 Chr.12: 6849112-6849112 on Build GRCh38
rs376989216 Chr.12: 6849116-6849116 on Build GRCh38
rs982498333 Chr.12: 6848776-6848776 on Build GRCh38
rs782239942 Chr.12: 6848879-6848879 on Build GRCh38
rs782089102 Chr.12: 6849066-6849066 on Build GRCh38
rs1053634078 Chr.12: 6848856-6848856 on Build GRCh38
rs782497054 Chr.12: 6849219-6849219 on Build GRCh38

View Full Product Details