Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7081931-7082332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1R
Entrez Gene ID
715
Gene Name
complement C1r subcomponent
Gene Aliases
-
UniGene
Hs.524224
Gene Chromosome Location
Chr.12: 7080209-7092447 on Build GRCh38
Transcripts
NM_001733.4 XM_017019915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3359924 c.1187G>A p.G396D Chr.12: 7082082-7082082 on Build GRCh38
5728222 c.1164G>A p.R388R Chr.12: 7082105-7082105 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780697431 Chr.12: 7082013-7082013 on Build GRCh38
rs747703059 Chr.12: 7082020-7082020 on Build GRCh38
rs948846954 Chr.12: 7081980-7081980 on Build GRCh38
rs977745613 Chr.12: 7082180-7082180 on Build GRCh38
rs915640581 Chr.12: 7082275-7082275 on Build GRCh38
rs900775092 Chr.12: 7082120-7082120 on Build GRCh38
rs775580498 Chr.12: 7082147-7082147 on Build GRCh38
rs944405050 Chr.12: 7081982-7081982 on Build GRCh38
rs773720903 Chr.12: 7082043-7082043 on Build GRCh38
rs182085700 Chr.12: 7082111-7082111 on Build GRCh38
rs968631264 Chr.12: 7082153-7082153 on Build GRCh38
rs1048611911 Chr.12: 7082000-7082000 on Build GRCh38
rs1036994049 Chr.12: 7082104-7082104 on Build GRCh38
rs770371130 Chr.12: 7082007-7082007 on Build GRCh38
rs902566595 Chr.12: 7082146-7082146 on Build GRCh38
rs888671162 Chr.12: 7082072-7082072 on Build GRCh38
rs762964953 Chr.12: 7082044-7082044 on Build GRCh38
rs1004877964 Chr.12: 7082073-7082073 on Build GRCh38
rs948464082 Chr.12: 7082286-7082286 on Build GRCh38
rs367741977 Chr.12: 7082123-7082123 on Build GRCh38
rs901164255 Chr.12: 7082119-7082119 on Build GRCh38
rs1021428433 Chr.12: 7082141-7082141 on Build GRCh38
rs999572492 Chr.12: 7082176-7082176 on Build GRCh38
rs1012683640 Chr.12: 7082297-7082297 on Build GRCh38
rs1028354322 Chr.12: 7082129-7082129 on Build GRCh38
rs959966862 Chr.12: 7082223-7082223 on Build GRCh38
rs767514366 Chr.12: 7082137-7082137 on Build GRCh38
rs992518120 Chr.12: 7082251-7082251 on Build GRCh38
rs1031627439 Chr.12: 7082211-7082211 on Build GRCh38
rs896480759 Chr.12: 7082233-7082233 on Build GRCh38
rs187620491 Chr.12: 7082274-7082274 on Build GRCh38
rs759775152 Chr.12: 7082131-7082131 on Build GRCh38

View Full Product Details