Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12117993-12118520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRP6
Entrez Gene ID
4040
Gene Name
LDL receptor related protein 6
Gene Aliases
ADCAD2, STHAG7
UniGene
Hs.584775
Gene Chromosome Location
Chr.12: 12116025-12267021 on Build GRCh38
Transcripts
NM_002336.2 XM_006719078.3 XM_011520671.2 XM_017019304.1

SNP Detail

SNP ID Chromosome Location
rs994537899 Chr.12: 12118345-12118345 on Build GRCh38
rs1007135950 Chr.12: 12118369-12118369 on Build GRCh38
rs946136879 Chr.12: 12118360-12118360 on Build GRCh38
rs904590133 Chr.12: 12118374-12118374 on Build GRCh38
rs1023973502 Chr.12: 12118200-12118200 on Build GRCh38
rs1055966674 Chr.12: 12118400-12118400 on Build GRCh38
rs562136208 Chr.12: 12118333-12118333 on Build GRCh38
rs975201223 Chr.12: 12118314-12118314 on Build GRCh38
rs921214393 Chr.12: 12118324-12118324 on Build GRCh38
rs951209001 Chr.12: 12118267-12118267 on Build GRCh38
rs886751904 Chr.12: 12118490-12118490 on Build GRCh38
rs1014471254 Chr.12: 12118420-12118420 on Build GRCh38
rs34069601 Chr.12: 12118140-12118141 on Build GRCh38
rs933907216 Chr.12: 12118336-12118336 on Build GRCh38
rs772034790 Chr.12: 12118484-12118484 on Build GRCh38
rs965551804 Chr.12: 12118412-12118412 on Build GRCh38
rs529383516 Chr.12: 12118211-12118214 on Build GRCh38
rs937258506 Chr.12: 12118242-12118242 on Build GRCh38
rs1052200798 Chr.12: 12118343-12118343 on Build GRCh38
rs571965595 Chr.12: 12118144-12118144 on Build GRCh38
rs936186160 Chr.12: 12118378-12118378 on Build GRCh38
rs188775539 Chr.12: 12118297-12118297 on Build GRCh38
rs559786268 Chr.12: 12118361-12118361 on Build GRCh38
rs756239619 Chr.12: 12118052-12118052 on Build GRCh38
rs777803203 Chr.12: 12118201-12118201 on Build GRCh38
rs968768291 Chr.12: 12118462-12118462 on Build GRCh38
rs1057385022 Chr.12: 12118287-12118287 on Build GRCh38
rs564448608 Chr.12: 12118259-12118259 on Build GRCh38
rs75937324 Chr.12: 12118049-12118049 on Build GRCh38
rs772739348 Chr.12: 12118185-12118188 on Build GRCh38
rs572144642 Chr.12: 12118335-12118335 on Build GRCh38
rs1036071143 Chr.12: 12118321-12118321 on Build GRCh38
rs989304906 Chr.12: 12118436-12118436 on Build GRCh38
rs187028420 Chr.12: 12118367-12118367 on Build GRCh38
rs1013966177 Chr.12: 12118186-12118186 on Build GRCh38
rs115201126 Chr.12: 12118155-12118155 on Build GRCh38
rs958933640 Chr.12: 12118182-12118182 on Build GRCh38
rs912712982 Chr.12: 12118105-12118105 on Build GRCh38
rs762480275 Chr.12: 12118176-12118176 on Build GRCh38
rs1020336689 Chr.12: 12118455-12118455 on Build GRCh38
rs111310175 Chr.12: 12118149-12118149 on Build GRCh38
rs535053198 Chr.12: 12118027-12118027 on Build GRCh38
rs947352418 Chr.12: 12118205-12118205 on Build GRCh38
rs576144622 Chr.12: 12118296-12118296 on Build GRCh38
rs181563497 Chr.12: 12118357-12118357 on Build GRCh38
rs927250334 Chr.12: 12118213-12118214 on Build GRCh38
rs545780740 Chr.12: 12118481-12118481 on Build GRCh38
rs551777337 Chr.12: 12118406-12118406 on Build GRCh38
rs982521716 Chr.12: 12118286-12118286 on Build GRCh38
rs772250055 Chr.12: 12118280-12118280 on Build GRCh38

View Full Product Details