Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 21814270-21814770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC9
Entrez Gene ID
10060
Gene Name
ATP binding cassette subfamily C member 9
Gene Aliases
ABC37, ATFB12, CANTU, CMD1O, SUR2
UniGene
Hs.732701
Gene Chromosome Location
Chr.12: 21797389-21941869 on Build GRCh38
Transcripts
NM_005691.3 NM_020297.3 XM_005253284.3 XM_005253286.3 XM_005253287.4 XM_005253288.3 XM_005253289.3 XM_005253290.3 XM_006719025.3 XM_011520545.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5638619 c.4090G>A p.D1364N Chr.12: 21814656-21814656 on Build GRCh38
6590531 c.4040G>A p.R1347H Chr.12: 21814706-21814706 on Build GRCh38
5638618 c.4090G>A p.D1364N Chr.12: 21814656-21814656 on Build GRCh38
5051507 c.4067C>A p.S1356Y Chr.12: 21814679-21814679 on Build GRCh38
3459272 c.4053G>A p.G1351G Chr.12: 21814693-21814693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565989226 Chr.12: 21814530-21814530 on Build GRCh38
rs1026323755 Chr.12: 21814398-21814398 on Build GRCh38
rs1042563557 Chr.12: 21814531-21814531 on Build GRCh38
rs537511747 Chr.12: 21814617-21814617 on Build GRCh38
rs1022669974 Chr.12: 21814353-21814353 on Build GRCh38
rs763715321 Chr.12: 21814595-21814595 on Build GRCh38
rs774500228 Chr.12: 21814625-21814625 on Build GRCh38
rs755341471 Chr.12: 21814654-21814654 on Build GRCh38
rs756560119 Chr.12: 21814648-21814648 on Build GRCh38
rs965764832 Chr.12: 21814502-21814502 on Build GRCh38
rs578145919 Chr.12: 21814608-21814608 on Build GRCh38
rs982134285 Chr.12: 21814457-21814457 on Build GRCh38
rs11046194 Chr.12: 21814419-21814419 on Build GRCh38
rs926902304 Chr.12: 21814459-21814459 on Build GRCh38
rs878897512 Chr.12: 21814607-21814607 on Build GRCh38
rs796151820 Chr.12: 21814739-21814739 on Build GRCh38
rs777578143 Chr.12: 21814660-21814660 on Build GRCh38
rs972520654 Chr.12: 21814352-21814352 on Build GRCh38
rs145005748 Chr.12: 21814684-21814684 on Build GRCh38
rs767492149 Chr.12: 21814631-21814631 on Build GRCh38
rs535009862 Chr.12: 21814572-21814572 on Build GRCh38
rs745787969 Chr.12: 21814594-21814594 on Build GRCh38
rs1005415927 Chr.12: 21814306-21814306 on Build GRCh38
rs557601519 Chr.12: 21814717-21814717 on Build GRCh38
rs778634668 Chr.12: 21814653-21814653 on Build GRCh38
rs201129154 Chr.12: 21814605-21814605 on Build GRCh38
rs112918103 Chr.12: 21814489-21814489 on Build GRCh38
rs906857986 Chr.12: 21814380-21814380 on Build GRCh38
rs112846331 Chr.12: 21814473-21814473 on Build GRCh38
rs900917228 Chr.12: 21814578-21814578 on Build GRCh38
rs113882813 Chr.12: 21814589-21814589 on Build GRCh38
rs775449737 Chr.12: 21814737-21814737 on Build GRCh38
rs751606135 Chr.12: 21814421-21814421 on Build GRCh38
rs761888147 Chr.12: 21814627-21814627 on Build GRCh38
rs895414316 Chr.12: 21814345-21814345 on Build GRCh38
rs963511472 Chr.12: 21814334-21814334 on Build GRCh38
rs1002511136 Chr.12: 21814393-21814393 on Build GRCh38
rs764224298 Chr.12: 21814309-21814309 on Build GRCh38
rs182689954 Chr.12: 21814507-21814507 on Build GRCh38
rs745866389 Chr.12: 21814728-21814728 on Build GRCh38
rs150563694 Chr.12: 21814467-21814467 on Build GRCh38
rs933674604 Chr.12: 21814583-21814583 on Build GRCh38
rs865858522 Chr.12: 21814688-21814688 on Build GRCh38
rs756557011 Chr.12: 21814619-21814620 on Build GRCh38
rs868834502 Chr.12: 21814706-21814706 on Build GRCh38
rs200058602 Chr.12: 21814349-21814349 on Build GRCh38
rs912598385 Chr.12: 21814499-21814499 on Build GRCh38
rs376991991 Chr.12: 21814645-21814645 on Build GRCh38
rs916746571 Chr.12: 21814589-21814591 on Build GRCh38
rs767871542 Chr.12: 21814615-21814615 on Build GRCh38
rs756856961 Chr.12: 21814702-21814702 on Build GRCh38
rs536178922 Chr.12: 21814515-21814515 on Build GRCh38
rs989996843 Chr.12: 21814432-21814432 on Build GRCh38
rs953924340 Chr.12: 21814299-21814299 on Build GRCh38
rs769587879 Chr.12: 21814734-21814734 on Build GRCh38
rs989622075 Chr.12: 21814471-21814471 on Build GRCh38
rs546852989 Chr.12: 21814526-21814526 on Build GRCh38
rs868023533 Chr.12: 21814656-21814656 on Build GRCh38
rs766810801 Chr.12: 21814651-21814651 on Build GRCh38
rs1005447267 Chr.12: 21814646-21814646 on Build GRCh38

View Full Product Details