Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 22660945-22661450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETNK1
Entrez Gene ID
55500
Gene Name
ethanolamine kinase 1
Gene Aliases
EKI, EKI 1, EKI1, Nbla10396
UniGene
Hs.29464
Gene Chromosome Location
Chr.12: 22625142-22690674 on Build GRCh38
Transcripts
NM_018638.4 XM_017019580.1 XM_017019581.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6441361 c.893_894CC>TT p.S298F Chr.12: 22661131-22661132 on Build GRCh38
5012507 c.956A>G p.N319S Chr.12: 22661194-22661194 on Build GRCh38
7121583 c.923A>G p.H308R Chr.12: 22661161-22661161 on Build GRCh38
4186501 c.958G>A p.E320K Chr.12: 22661196-22661196 on Build GRCh38
6136107 c.825-1_825GG>TT p.? Chr.12: 22661062-22661063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769607611 Chr.12: 22661226-22661226 on Build GRCh38
rs770093854 Chr.12: 22661058-22661059 on Build GRCh38
rs764510405 Chr.12: 22661298-22661298 on Build GRCh38
rs528695758 Chr.12: 22661331-22661331 on Build GRCh38
rs748475461 Chr.12: 22661018-22661018 on Build GRCh38
rs1001479800 Chr.12: 22661311-22661311 on Build GRCh38
rs529230783 Chr.12: 22661392-22661392 on Build GRCh38
rs979149187 Chr.12: 22661412-22661412 on Build GRCh38
rs564152157 Chr.12: 22661105-22661105 on Build GRCh38
rs746458212 Chr.12: 22661184-22661184 on Build GRCh38
rs764201342 Chr.12: 22661082-22661082 on Build GRCh38
rs752911248 Chr.12: 22661064-22661064 on Build GRCh38
rs549004680 Chr.12: 22661408-22661408 on Build GRCh38
rs184076882 Chr.12: 22661351-22661351 on Build GRCh38
rs770992839 Chr.12: 22661194-22661194 on Build GRCh38
rs767948882 Chr.12: 22661091-22661091 on Build GRCh38
rs773738762 Chr.12: 22661240-22661244 on Build GRCh38
rs777406621 Chr.12: 22661172-22661172 on Build GRCh38
rs61921420 Chr.12: 22661036-22661036 on Build GRCh38
rs747740695 Chr.12: 22661152-22661152 on Build GRCh38
rs773064963 Chr.12: 22661025-22661025 on Build GRCh38
rs971786934 Chr.12: 22661310-22661310 on Build GRCh38
rs989910138 Chr.12: 22660978-22660978 on Build GRCh38
rs1034236895 Chr.12: 22661312-22661312 on Build GRCh38
rs976234285 Chr.12: 22661410-22661410 on Build GRCh38
rs989941112 Chr.12: 22661341-22661341 on Build GRCh38
rs765405801 Chr.12: 22661047-22661047 on Build GRCh38
rs750817176 Chr.12: 22661094-22661094 on Build GRCh38
rs945703697 Chr.12: 22661029-22661029 on Build GRCh38
rs1033598099 Chr.12: 22661252-22661252 on Build GRCh38
rs370901209 Chr.12: 22661038-22661038 on Build GRCh38
rs372374763 Chr.12: 22661020-22661020 on Build GRCh38
rs577787968 Chr.12: 22661296-22661296 on Build GRCh38
rs967187615 Chr.12: 22661382-22661382 on Build GRCh38
rs897205878 Chr.12: 22661193-22661193 on Build GRCh38
rs762349594 Chr.12: 22661355-22661355 on Build GRCh38
rs898321575 Chr.12: 22661238-22661238 on Build GRCh38
rs752120627 Chr.12: 22661086-22661086 on Build GRCh38
rs934283495 Chr.12: 22661069-22661069 on Build GRCh38
rs769389633 Chr.12: 22661362-22661363 on Build GRCh38
rs762989356 Chr.12: 22661070-22661070 on Build GRCh38
rs1002001970 Chr.12: 22661241-22661241 on Build GRCh38
rs1024350415 Chr.12: 22661308-22661308 on Build GRCh38
rs4963793 Chr.12: 22661102-22661102 on Build GRCh38
rs757914471 Chr.12: 22661126-22661126 on Build GRCh38
rs745679082 Chr.12: 22661223-22661223 on Build GRCh38
rs776666269 Chr.12: 22661213-22661213 on Build GRCh38
rs760683361 Chr.12: 22661030-22661030 on Build GRCh38
rs756457134 Chr.12: 22661099-22661099 on Build GRCh38
rs775613608 Chr.12: 22661236-22661236 on Build GRCh38
rs75025746 Chr.12: 22661089-22661089 on Build GRCh38

View Full Product Details