Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 22673562-22674075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETNK1
Entrez Gene ID
55500
Gene Name
ethanolamine kinase 1
Gene Aliases
EKI, EKI 1, EKI1, Nbla10396
UniGene
Hs.29464
Gene Chromosome Location
Chr.12: 22625142-22690674 on Build GRCh38
Transcripts
NM_018638.4 XM_017019580.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7366678 c.1133A>G p.K378R Chr.12: 22673581-22673581 on Build GRCh38
692731 c.1165G>A p.E389K Chr.12: 22673613-22673613 on Build GRCh38
7214057 c.1171G>T p.E391* Chr.12: 22673619-22673619 on Build GRCh38
1236021 c.1134A>T p.K378N Chr.12: 22673582-22673582 on Build GRCh38
6656650 c.1135G>T p.E379* Chr.12: 22673583-22673583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565596037 Chr.12: 22674023-22674023 on Build GRCh38
rs191438505 Chr.12: 22673992-22673992 on Build GRCh38
rs1029176910 Chr.12: 22673775-22673775 on Build GRCh38
rs370944369 Chr.12: 22673966-22673966 on Build GRCh38
rs561463572 Chr.12: 22673787-22673787 on Build GRCh38
rs369325062 Chr.12: 22673734-22673734 on Build GRCh38
rs1040744335 Chr.12: 22673996-22673996 on Build GRCh38
rs760027138 Chr.12: 22673684-22673684 on Build GRCh38
rs948025593 Chr.12: 22673864-22673864 on Build GRCh38
rs905265260 Chr.12: 22673896-22673896 on Build GRCh38
rs913689858 Chr.12: 22673988-22673988 on Build GRCh38
rs771116039 Chr.12: 22673731-22673732 on Build GRCh38
rs547588129 Chr.12: 22673964-22673964 on Build GRCh38
rs772199457 Chr.12: 22673630-22673630 on Build GRCh38
rs647446 Chr.12: 22673875-22673875 on Build GRCh38
rs902115882 Chr.12: 22673753-22673753 on Build GRCh38
rs1035923664 Chr.12: 22673929-22673929 on Build GRCh38
rs988535580 Chr.12: 22673733-22673733 on Build GRCh38
rs968381846 Chr.12: 22673769-22673769 on Build GRCh38
rs1001351297 Chr.12: 22673923-22673923 on Build GRCh38
rs370798726 Chr.12: 22673825-22673825 on Build GRCh38
rs917894810 Chr.12: 22673848-22673848 on Build GRCh38
rs1052211032 Chr.12: 22673948-22673948 on Build GRCh38
rs773352173 Chr.12: 22673642-22673642 on Build GRCh38
rs773977042 Chr.12: 22673627-22673627 on Build GRCh38
rs374176525 Chr.12: 22673622-22673622 on Build GRCh38
rs942870149 Chr.12: 22673801-22673801 on Build GRCh38
rs573261265 Chr.12: 22673917-22673917 on Build GRCh38
rs892130307 Chr.12: 22673689-22673689 on Build GRCh38
rs1052111149 Chr.12: 22673680-22673680 on Build GRCh38
rs960213947 Chr.12: 22673731-22673731 on Build GRCh38
rs1001297832 Chr.12: 22673725-22673725 on Build GRCh38
rs186243437 Chr.12: 22673890-22673890 on Build GRCh38
rs543141235 Chr.12: 22673882-22673882 on Build GRCh38
rs771496948 Chr.12: 22673678-22673678 on Build GRCh38
rs543525729 Chr.12: 22674014-22674014 on Build GRCh38
rs17349043 Chr.12: 22673817-22673817 on Build GRCh38
rs369949913 Chr.12: 22673699-22673699 on Build GRCh38
rs753477128 Chr.12: 22673681-22673681 on Build GRCh38
rs925137801 Chr.12: 22673916-22673916 on Build GRCh38
rs180977703 Chr.12: 22673768-22673768 on Build GRCh38
rs529699973 Chr.12: 22673726-22673726 on Build GRCh38
rs747295452 Chr.12: 22673729-22673730 on Build GRCh38
rs76887157 Chr.12: 22673839-22673839 on Build GRCh38
rs996415927 Chr.12: 22673767-22673767 on Build GRCh38
rs756790376 Chr.12: 22673675-22673675 on Build GRCh38
rs952165267 Chr.12: 22673792-22673792 on Build GRCh38
rs760602109 Chr.12: 22673655-22673655 on Build GRCh38
rs895612625 Chr.12: 22673955-22673955 on Build GRCh38
rs776949956 Chr.12: 22673679-22673679 on Build GRCh38
rs1007926319 Chr.12: 22673701-22673701 on Build GRCh38
rs558797835 Chr.12: 22673797-22673797 on Build GRCh38
rs984874288 Chr.12: 22673813-22673813 on Build GRCh38
rs538582464 Chr.12: 22673795-22673795 on Build GRCh38
rs375707259 Chr.12: 22673640-22673640 on Build GRCh38
rs930218026 Chr.12: 22673830-22673830 on Build GRCh38
rs748368675 Chr.12: 22673606-22673606 on Build GRCh38
rs763661599 Chr.12: 22673678-22673679 on Build GRCh38
rs374960062 Chr.12: 22673866-22673866 on Build GRCh38
rs34032941 Chr.12: 22673614-22673614 on Build GRCh38
rs569020172 Chr.12: 22673803-22673803 on Build GRCh38
rs943766960 Chr.12: 22673989-22673989 on Build GRCh38
rs968810235 Chr.12: 22674027-22674027 on Build GRCh38

View Full Product Details