Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49066041-49066552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHEBL1
Entrez Gene ID
121268
Gene Name
Ras homolog enriched in brain like 1
Gene Aliases
RHEBL1c
UniGene
Hs.159013
Gene Chromosome Location
Chr.12: 49064676-49070682 on Build GRCh38
Transcripts
NM_001303126.1 NM_144593.2 NR_130123.1 XM_017018794.1 XM_017018795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3986982 c.349G>T p.V117L Chr.12: 49066262-49066262 on Build GRCh38
5987439 c.367C>T p.L123F Chr.12: 49066244-49066244 on Build GRCh38
6137213 c.332G>A p.R111Q Chr.12: 49066476-49066476 on Build GRCh38
6766200 c.285C>T p.V95V Chr.12: 49066523-49066523 on Build GRCh38
6425586 c.362C>A p.A121E Chr.12: 49066249-49066249 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs77472342 Chr.12: 49066352-49066352 on Build GRCh38
rs536373755 Chr.12: 49066468-49066468 on Build GRCh38
rs761140168 Chr.12: 49066457-49066457 on Build GRCh38
rs997151804 Chr.12: 49066386-49066386 on Build GRCh38
rs373786554 Chr.12: 49066498-49066498 on Build GRCh38
rs984571222 Chr.12: 49066068-49066068 on Build GRCh38
rs1016456698 Chr.12: 49066105-49066105 on Build GRCh38
rs753276277 Chr.12: 49066462-49066462 on Build GRCh38
rs750601845 Chr.12: 49066502-49066502 on Build GRCh38
rs781253789 Chr.12: 49066487-49066487 on Build GRCh38
rs181745558 Chr.12: 49066433-49066433 on Build GRCh38
rs1041071947 Chr.12: 49066370-49066370 on Build GRCh38
rs994441577 Chr.12: 49066381-49066381 on Build GRCh38
rs542688904 Chr.12: 49066312-49066312 on Build GRCh38
rs113813611 Chr.12: 49066391-49066391 on Build GRCh38
rs763157208 Chr.12: 49066385-49066385 on Build GRCh38
rs375870579 Chr.12: 49066285-49066285 on Build GRCh38
rs755706621 Chr.12: 49066392-49066392 on Build GRCh38
rs765835216 Chr.12: 49066298-49066298 on Build GRCh38
rs116459786 Chr.12: 49066365-49066365 on Build GRCh38
rs755544251 Chr.12: 49066446-49066447 on Build GRCh38
rs1055722463 Chr.12: 49066308-49066308 on Build GRCh38
rs750870160 Chr.12: 49066281-49066281 on Build GRCh38
rs370921880 Chr.12: 49066435-49066435 on Build GRCh38
rs142176077 Chr.12: 49066479-49066479 on Build GRCh38
rs377588652 Chr.12: 49066318-49066318 on Build GRCh38
rs763050719 Chr.12: 49066228-49066228 on Build GRCh38
rs888609901 Chr.12: 49066431-49066431 on Build GRCh38
rs774562669 Chr.12: 49066245-49066245 on Build GRCh38
rs564458570 Chr.12: 49066432-49066432 on Build GRCh38
rs962545950 Chr.12: 49066144-49066144 on Build GRCh38
rs529017203 Chr.12: 49066304-49066304 on Build GRCh38
rs200725647 Chr.12: 49066453-49066453 on Build GRCh38
rs373731837 Chr.12: 49066311-49066311 on Build GRCh38
rs745796190 Chr.12: 49066496-49066496 on Build GRCh38
rs752028469 Chr.12: 49066313-49066321 on Build GRCh38
rs777676008 Chr.12: 49066477-49066477 on Build GRCh38
rs928912703 Chr.12: 49066234-49066234 on Build GRCh38
rs201327675 Chr.12: 49066319-49066319 on Build GRCh38
rs1053855900 Chr.12: 49066340-49066340 on Build GRCh38
rs769941020 Chr.12: 49066210-49066210 on Build GRCh38
rs370160928 Chr.12: 49066187-49066187 on Build GRCh38
rs148047837 Chr.12: 49066476-49066476 on Build GRCh38
rs990486615 Chr.12: 49066253-49066253 on Build GRCh38
rs767467419 Chr.12: 49066475-49066475 on Build GRCh38
rs769492355 Chr.12: 49066223-49066223 on Build GRCh38
rs752340547 Chr.12: 49066469-49066469 on Build GRCh38
rs773510023 Chr.12: 49066459-49066459 on Build GRCh38
rs768722325 Chr.12: 49066510-49066510 on Build GRCh38
rs148140221 Chr.12: 49066418-49066418 on Build GRCh38
rs773597911 Chr.12: 49066446-49066446 on Build GRCh38
rs866263648 Chr.12: 49066240-49066240 on Build GRCh38
rs746951158 Chr.12: 49066192-49066192 on Build GRCh38
rs369525595 Chr.12: 49066320-49066320 on Build GRCh38
rs553453687 Chr.12: 49066474-49066474 on Build GRCh38
rs935197735 Chr.12: 49066360-49066360 on Build GRCh38
rs186774244 Chr.12: 49066460-49066460 on Build GRCh38
rs762230005 Chr.12: 49066250-49066250 on Build GRCh38
rs762035223 Chr.12: 49066445-49066445 on Build GRCh38
rs759154146 Chr.12: 49066465-49066465 on Build GRCh38
rs563464646 Chr.12: 49066093-49066093 on Build GRCh38
rs768967826 Chr.12: 49066244-49066244 on Build GRCh38
rs949306632 Chr.12: 49066313-49066313 on Build GRCh38
rs369102432 Chr.12: 49066302-49066302 on Build GRCh38
rs371246457 Chr.12: 49066277-49066277 on Build GRCh38
rs771027543 Chr.12: 49066427-49066427 on Build GRCh38

View Full Product Details