Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2400767-2401186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RER1
Entrez Gene ID
11079
Gene Name
retention in endoplasmic reticulum sorting receptor 1
Gene Aliases
-
UniGene
Hs.525527
Gene Chromosome Location
Chr.1: 2391314-2405444 on Build GRCh38
Transcripts
NM_007033.4 XM_005244713.4 XM_006710306.2 XM_011540542.1 XM_011540543.1 XM_017000131.1 XM_017000132.1 XM_017000133.1 XM_017000134.1 XM_017000135.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7198239 c.340C>G p.R114G Chr.1: 2400910-2400910 on Build GRCh38
5544179 c.359A>C p.K120T Chr.1: 2400929-2400929 on Build GRCh38
5954998 c.304C>T p.P102S Chr.1: 2400874-2400874 on Build GRCh38
3984844 c.299C>G p.S100W Chr.1: 2400869-2400869 on Build GRCh38
3485303 c.305C>T p.P102L Chr.1: 2400875-2400875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777310477 Chr.1: 2401061-2401061 on Build GRCh38
rs768843089 Chr.1: 2400869-2400869 on Build GRCh38
rs987569319 Chr.1: 2401052-2401052 on Build GRCh38
rs746907390 Chr.1: 2400857-2400857 on Build GRCh38
rs1003539722 Chr.1: 2400947-2400947 on Build GRCh38
rs945777478 Chr.1: 2400977-2400977 on Build GRCh38
rs983363369 Chr.1: 2401002-2401002 on Build GRCh38
rs866875767 Chr.1: 2400871-2400871 on Build GRCh38
rs906662558 Chr.1: 2401137-2401137 on Build GRCh38
rs762623581 Chr.1: 2400965-2400965 on Build GRCh38
rs772519977 Chr.1: 2400882-2400882 on Build GRCh38
rs915404592 Chr.1: 2401098-2401098 on Build GRCh38
rs767852176 Chr.1: 2401125-2401125 on Build GRCh38
rs1016523780 Chr.1: 2401004-2401004 on Build GRCh38
rs375264274 Chr.1: 2400861-2400861 on Build GRCh38
rs761909194 Chr.1: 2400827-2400828 on Build GRCh38
rs778969831 Chr.1: 2400870-2400870 on Build GRCh38
rs1016142228 Chr.1: 2401164-2401164 on Build GRCh38
rs370517114 Chr.1: 2400845-2400845 on Build GRCh38
rs371428165 Chr.1: 2400819-2400819 on Build GRCh38
rs376395746 Chr.1: 2400816-2400816 on Build GRCh38
rs1003570703 Chr.1: 2401158-2401158 on Build GRCh38
rs948185734 Chr.1: 2401104-2401104 on Build GRCh38
rs758160454 Chr.1: 2400834-2400834 on Build GRCh38
rs540537116 Chr.1: 2401069-2401069 on Build GRCh38
rs1041076389 Chr.1: 2400885-2400885 on Build GRCh38
rs555736280 Chr.1: 2400900-2400900 on Build GRCh38
rs777594923 Chr.1: 2400853-2400853 on Build GRCh38
rs752325044 Chr.1: 2400954-2400954 on Build GRCh38
rs557727556 Chr.1: 2401038-2401038 on Build GRCh38
rs1054015121 Chr.1: 2401088-2401088 on Build GRCh38
rs373549265 Chr.1: 2400806-2400806 on Build GRCh38
rs377673649 Chr.1: 2400888-2400888 on Build GRCh38
rs577526206 Chr.1: 2401156-2401156 on Build GRCh38
rs1001425317 Chr.1: 2401017-2401017 on Build GRCh38
rs758964465 Chr.1: 2400941-2400941 on Build GRCh38
rs777253701 Chr.1: 2400932-2400932 on Build GRCh38
rs541077842 Chr.1: 2401029-2401029 on Build GRCh38
rs921716471 Chr.1: 2401036-2401036 on Build GRCh38
rs923610126 Chr.1: 2401066-2401066 on Build GRCh38
rs961471074 Chr.1: 2401087-2401087 on Build GRCh38
rs1023643910 Chr.1: 2400839-2400839 on Build GRCh38
rs935602986 Chr.1: 2401070-2401070 on Build GRCh38
rs952546300 Chr.1: 2401127-2401127 on Build GRCh38
rs538575110 Chr.1: 2401164-2401164 on Build GRCh38
rs897834796 Chr.1: 2401154-2401166 on Build GRCh38
rs12037485 Chr.1: 2400952-2400952 on Build GRCh38
rs950721532 Chr.1: 2400976-2400976 on Build GRCh38
rs376882772 Chr.1: 2400833-2400833 on Build GRCh38
rs372657483 Chr.1: 2400831-2400831 on Build GRCh38
rs1005215704 Chr.1: 2401163-2401163 on Build GRCh38
rs1025354448 Chr.1: 2401068-2401068 on Build GRCh38
rs1047531164 Chr.1: 2401159-2401159 on Build GRCh38
rs1042547988 Chr.1: 2400985-2400985 on Build GRCh38
rs963172578 Chr.1: 2401014-2401014 on Build GRCh38
rs887086912 Chr.1: 2401162-2401162 on Build GRCh38
rs764784280 Chr.1: 2400825-2400825 on Build GRCh38
rs749313704 Chr.1: 2400862-2400862 on Build GRCh38

View Full Product Details