Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6085116-6085608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNAB2
Entrez Gene ID
8514
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 2
Gene Aliases
AKR6A5, HKvbeta2, HKvbeta2.1, HKvbeta2.2, KCNA2B, KV-BETA-2
UniGene
Hs.440497
Gene Chromosome Location
Chr.1: 5992298-6101193 on Build GRCh38
Transcripts
NM_001199860.1 NM_001199861.1 NM_001199862.1 NM_001199863.1 NM_003636.3 NM_172130.2 XM_005263514.2 XM_011542321.2 XM_011542322.2 XM_017002618.1 XM_017002619.1 XM_017002620.1 XM_017002621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1185283 c.286G>T p.E96* Chr.1: 6085208-6085208 on Build GRCh38
4009154 c.417A>G p.K139K Chr.1: 6085240-6085240 on Build GRCh38
5938686 c.425+7G>A p.? Chr.1: 6085255-6085255 on Build GRCh38
4547475 c.423G>A p.W141* Chr.1: 6085246-6085246 on Build GRCh38
4934952 c.391G>A p.V131I Chr.1: 6085214-6085214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771037177 Chr.1: 6085236-6085236 on Build GRCh38
rs896043055 Chr.1: 6085225-6085225 on Build GRCh38
rs904554620 Chr.1: 6085432-6085432 on Build GRCh38
rs976993422 Chr.1: 6085473-6085473 on Build GRCh38
rs985924292 Chr.1: 6085489-6085489 on Build GRCh38
rs985424919 Chr.1: 6085496-6085496 on Build GRCh38
rs776029092 Chr.1: 6085258-6085258 on Build GRCh38
rs370400010 Chr.1: 6085384-6085384 on Build GRCh38
rs569979142 Chr.1: 6085446-6085446 on Build GRCh38
rs765682344 Chr.1: 6085181-6085181 on Build GRCh38
rs1057268627 Chr.1: 6085373-6085373 on Build GRCh38
rs1013343048 Chr.1: 6085245-6085245 on Build GRCh38
rs553287281 Chr.1: 6085483-6085483 on Build GRCh38
rs577288968 Chr.1: 6085200-6085200 on Build GRCh38
rs371993946 Chr.1: 6085154-6085154 on Build GRCh38
rs140923168 Chr.1: 6085542-6085542 on Build GRCh38
rs918847303 Chr.1: 6085526-6085526 on Build GRCh38
rs922729250 Chr.1: 6085293-6085293 on Build GRCh38
rs373174603 Chr.1: 6085183-6085183 on Build GRCh38
rs762708527 Chr.1: 6085295-6085295 on Build GRCh38
rs1022648632 Chr.1: 6085402-6085402 on Build GRCh38
rs771069169 Chr.1: 6085157-6085157 on Build GRCh38
rs750076076 Chr.1: 6085195-6085195 on Build GRCh38
rs1056660464 Chr.1: 6085549-6085550 on Build GRCh38
rs757329311 Chr.1: 6085294-6085294 on Build GRCh38
rs896060635 Chr.1: 6085382-6085382 on Build GRCh38
rs753642030 Chr.1: 6085287-6085287 on Build GRCh38
rs375700932 Chr.1: 6085328-6085328 on Build GRCh38
rs200963488 Chr.1: 6085283-6085283 on Build GRCh38
rs567438833 Chr.1: 6085478-6085478 on Build GRCh38
rs764523207 Chr.1: 6085261-6085261 on Build GRCh38
rs200151183 Chr.1: 6085256-6085256 on Build GRCh38
rs918349614 Chr.1: 6085564-6085564 on Build GRCh38
rs767083225 Chr.1: 6085188-6085188 on Build GRCh38
rs577251480 Chr.1: 6085468-6085468 on Build GRCh38
rs114763101 Chr.1: 6085466-6085466 on Build GRCh38
rs1013985805 Chr.1: 6085399-6085399 on Build GRCh38
rs541553661 Chr.1: 6085469-6085469 on Build GRCh38
rs941024756 Chr.1: 6085523-6085523 on Build GRCh38
rs36032813 Chr.1: 6085320-6085320 on Build GRCh38
rs765115783 Chr.1: 6085234-6085234 on Build GRCh38
rs776844923 Chr.1: 6085161-6085161 on Build GRCh38
rs767045580 Chr.1: 6085281-6085281 on Build GRCh38
rs778747851 Chr.1: 6085288-6085288 on Build GRCh38
rs968689873 Chr.1: 6085255-6085255 on Build GRCh38
rs540985324 Chr.1: 6085502-6085502 on Build GRCh38
rs201665803 Chr.1: 6085254-6085254 on Build GRCh38
rs1033376545 Chr.1: 6085429-6085429 on Build GRCh38
rs755965564 Chr.1: 6085198-6085198 on Build GRCh38
rs563605401 Chr.1: 6085572-6085572 on Build GRCh38
rs779789708 Chr.1: 6085203-6085203 on Build GRCh38
rs537411186 Chr.1: 6085461-6085461 on Build GRCh38
rs1038371826 Chr.1: 6085521-6085521 on Build GRCh38
rs547260720 Chr.1: 6085338-6085338 on Build GRCh38
rs909783593 Chr.1: 6085517-6085517 on Build GRCh38
rs770324119 Chr.1: 6085257-6085257 on Build GRCh38
rs200139970 Chr.1: 6085269-6085269 on Build GRCh38
rs554862095 Chr.1: 6085408-6085408 on Build GRCh38
rs888195734 Chr.1: 6085170-6085170 on Build GRCh38
rs1044566026 Chr.1: 6085558-6085558 on Build GRCh38
rs540160792 Chr.1: 6085291-6085291 on Build GRCh38
rs34975389 Chr.1: 6085213-6085213 on Build GRCh38
rs941395226 Chr.1: 6085497-6085497 on Build GRCh38
rs144471831 Chr.1: 6085212-6085212 on Build GRCh38
rs1023624633 Chr.1: 6085423-6085423 on Build GRCh38
rs1049370077 Chr.1: 6085140-6085140 on Build GRCh38
rs932321728 Chr.1: 6085138-6085138 on Build GRCh38
rs7512313 Chr.1: 6085405-6085405 on Build GRCh38
rs200509611 Chr.1: 6085172-6085172 on Build GRCh38
rs751055478 Chr.1: 6085186-6085186 on Build GRCh38
rs1029535778 Chr.1: 6085479-6085479 on Build GRCh38
rs764624561 Chr.1: 6085184-6085184 on Build GRCh38
rs985778605 Chr.1: 6085308-6085308 on Build GRCh38
rs2294936 Chr.1: 6085305-6085305 on Build GRCh38
rs574931437 Chr.1: 6085484-6085484 on Build GRCh38
rs144038305 Chr.1: 6085295-6085295 on Build GRCh38
rs896633827 Chr.1: 6085550-6085550 on Build GRCh38
rs1012400070 Chr.1: 6085554-6085554 on Build GRCh38
rs756736558 Chr.1: 6085187-6085187 on Build GRCh38
rs151230534 Chr.1: 6085329-6085329 on Build GRCh38
rs778781829 Chr.1: 6085290-6085290 on Build GRCh38
rs778849824 Chr.1: 6085217-6085217 on Build GRCh38
rs774858303 Chr.1: 6085268-6085268 on Build GRCh38
rs2294935 Chr.1: 6085319-6085319 on Build GRCh38
rs999850554 Chr.1: 6085406-6085406 on Build GRCh38
rs761354245 Chr.1: 6085232-6085232 on Build GRCh38
rs2294934 Chr.1: 6085347-6085347 on Build GRCh38
rs368379992 Chr.1: 6085228-6085228 on Build GRCh38
rs758130104 Chr.1: 6085269-6085269 on Build GRCh38
rs201718200 Chr.1: 6085260-6085260 on Build GRCh38
rs76508254 Chr.1: 6085317-6085317 on Build GRCh38

View Full Product Details