Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6234792-6235330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICMT
Entrez Gene ID
23463
Gene Name
isoprenylcysteine carboxyl methyltransferase
Gene Aliases
HSTE14, MST098, MSTP098, PCCMT, PCMT, PPMT
UniGene
Hs.515688
Gene Chromosome Location
Chr.1: 6221193-6235984 on Build GRCh38
Transcripts
NM_012405.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4009239 c.245T>C p.L82P Chr.1: 6234925-6234925 on Build GRCh38
3360979 c.206G>A p.R69Q Chr.1: 6234964-6234964 on Build GRCh38
395172 c.200C>T p.A67V Chr.1: 6234970-6234970 on Build GRCh38
4691727 c.243G>A p.T81T Chr.1: 6234927-6234927 on Build GRCh38
6211955 c.284+7A>T p.? Chr.1: 6234879-6234879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375689804 Chr.1: 6234856-6234856 on Build GRCh38
rs190451851 Chr.1: 6235290-6235290 on Build GRCh38
rs989504693 Chr.1: 6234953-6234953 on Build GRCh38
rs1033664787 Chr.1: 6235222-6235222 on Build GRCh38
rs930405676 Chr.1: 6235072-6235072 on Build GRCh38
rs549148083 Chr.1: 6235099-6235099 on Build GRCh38
rs186824400 Chr.1: 6234898-6234898 on Build GRCh38
rs756802838 Chr.1: 6235235-6235235 on Build GRCh38
rs367569358 Chr.1: 6235247-6235247 on Build GRCh38
rs776161046 Chr.1: 6234881-6234881 on Build GRCh38
rs972021929 Chr.1: 6235186-6235187 on Build GRCh38
rs558788817 Chr.1: 6234921-6234921 on Build GRCh38
rs771526458 Chr.1: 6235009-6235009 on Build GRCh38
rs775283376 Chr.1: 6234844-6234853 on Build GRCh38
rs770581951 Chr.1: 6234867-6234867 on Build GRCh38
rs963583797 Chr.1: 6235171-6235171 on Build GRCh38
rs911799928 Chr.1: 6235031-6235031 on Build GRCh38
rs567836909 Chr.1: 6235271-6235271 on Build GRCh38
rs775203958 Chr.1: 6235020-6235020 on Build GRCh38
rs926142882 Chr.1: 6235248-6235248 on Build GRCh38
rs755176396 Chr.1: 6234847-6234847 on Build GRCh38
rs1007389706 Chr.1: 6235128-6235128 on Build GRCh38
rs530411828 Chr.1: 6235034-6235034 on Build GRCh38
rs943231215 Chr.1: 6235037-6235037 on Build GRCh38
rs759146026 Chr.1: 6234895-6234895 on Build GRCh38
rs756124227 Chr.1: 6235071-6235071 on Build GRCh38
rs531252816 Chr.1: 6235226-6235226 on Build GRCh38
rs1025642290 Chr.1: 6234832-6234832 on Build GRCh38
rs934879220 Chr.1: 6235258-6235258 on Build GRCh38
rs549681000 Chr.1: 6235141-6235141 on Build GRCh38
rs113245791 Chr.1: 6234861-6234861 on Build GRCh38
rs901445349 Chr.1: 6235275-6235276 on Build GRCh38
rs980742317 Chr.1: 6234849-6234849 on Build GRCh38
rs1002382242 Chr.1: 6235194-6235194 on Build GRCh38
rs145062634 Chr.1: 6234959-6234959 on Build GRCh38
rs553577614 Chr.1: 6235108-6235108 on Build GRCh38
rs537451128 Chr.1: 6234915-6234915 on Build GRCh38
rs775200107 Chr.1: 6234906-6234906 on Build GRCh38
rs563995964 Chr.1: 6235223-6235223 on Build GRCh38
rs776047014 Chr.1: 6235011-6235011 on Build GRCh38
rs1053360399 Chr.1: 6235266-6235266 on Build GRCh38
rs747543998 Chr.1: 6234995-6234995 on Build GRCh38
rs778229072 Chr.1: 6234993-6234993 on Build GRCh38
rs748514612 Chr.1: 6234981-6234981 on Build GRCh38
rs752598768 Chr.1: 6235040-6235040 on Build GRCh38
rs751218475 Chr.1: 6235006-6235007 on Build GRCh38
rs766467773 Chr.1: 6234955-6234955 on Build GRCh38
rs769391242 Chr.1: 6235018-6235018 on Build GRCh38
rs753911724 Chr.1: 6234957-6234957 on Build GRCh38
rs757126497 Chr.1: 6234923-6234923 on Build GRCh38
rs777109510 Chr.1: 6235083-6235083 on Build GRCh38
rs1056872779 Chr.1: 6234872-6234872 on Build GRCh38
rs762795960 Chr.1: 6234912-6234912 on Build GRCh38
rs987443473 Chr.1: 6234984-6234984 on Build GRCh38
rs773837622 Chr.1: 6234858-6234858 on Build GRCh38
rs541662841 Chr.1: 6235090-6235090 on Build GRCh38
rs954149093 Chr.1: 6235084-6235084 on Build GRCh38
rs145910484 Chr.1: 6235111-6235111 on Build GRCh38
rs758843002 Chr.1: 6234988-6234988 on Build GRCh38
rs959716544 Chr.1: 6235204-6235204 on Build GRCh38
rs898811739 Chr.1: 6235052-6235052 on Build GRCh38
rs972104197 Chr.1: 6234821-6234821 on Build GRCh38
rs779169900 Chr.1: 6234974-6234974 on Build GRCh38
rs554429238 Chr.1: 6234827-6234827 on Build GRCh38
rs1047924279 Chr.1: 6235079-6235079 on Build GRCh38
rs906802118 Chr.1: 6235157-6235157 on Build GRCh38
rs778695223 Chr.1: 6235236-6235236 on Build GRCh38
rs763657504 Chr.1: 6234997-6235000 on Build GRCh38
rs762676875 Chr.1: 6234988-6234989 on Build GRCh38
rs745362126 Chr.1: 6235017-6235017 on Build GRCh38
rs117867161 Chr.1: 6235155-6235155 on Build GRCh38
rs564838441 Chr.1: 6235254-6235254 on Build GRCh38
rs35298962 Chr.1: 6235206-6235206 on Build GRCh38
rs945195580 Chr.1: 6234964-6234964 on Build GRCh38
rs749484160 Chr.1: 6234837-6234837 on Build GRCh38
rs753395251 Chr.1: 6235229-6235229 on Build GRCh38
rs1004787710 Chr.1: 6235139-6235139 on Build GRCh38
rs901186515 Chr.1: 6235063-6235063 on Build GRCh38
rs779173069 Chr.1: 6234852-6234852 on Build GRCh38
rs370554485 Chr.1: 6234823-6234823 on Build GRCh38

View Full Product Details