Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49869018-49869524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAIM2
Entrez Gene ID
23017
Gene Name
Fas apoptotic inhibitory molecule 2
Gene Aliases
LFG, LFG2, NGP35, NMP35, TMBIM2
UniGene
Hs.567424
Gene Chromosome Location
Chr.12: 49866896-49904275 on Build GRCh38
Transcripts
NM_012306.3 XM_005268730.3 XM_017019040.1

SNP Detail

SNP ID Chromosome Location
rs913319283 Chr.12: 49869246-49869246 on Build GRCh38
rs1025823946 Chr.12: 49869167-49869167 on Build GRCh38
rs1038010179 Chr.12: 49869128-49869128 on Build GRCh38
rs979775424 Chr.12: 49869204-49869204 on Build GRCh38
rs1005606942 Chr.12: 49869458-49869458 on Build GRCh38
rs960478353 Chr.12: 49869177-49869177 on Build GRCh38
rs186867185 Chr.12: 49869487-49869487 on Build GRCh38
rs554002763 Chr.12: 49869081-49869081 on Build GRCh38
rs889466363 Chr.12: 49869275-49869276 on Build GRCh38
rs544071225 Chr.12: 49869184-49869184 on Build GRCh38
rs1047947622 Chr.12: 49869439-49869439 on Build GRCh38
rs1051253897 Chr.12: 49869352-49869352 on Build GRCh38
rs1025678779 Chr.12: 49869059-49869059 on Build GRCh38
rs560447248 Chr.12: 49869457-49869457 on Build GRCh38
rs1039030599 Chr.12: 49869362-49869362 on Build GRCh38
rs911408551 Chr.12: 49869353-49869353 on Build GRCh38
rs923682420 Chr.12: 49869242-49869242 on Build GRCh38
rs925936227 Chr.12: 49869209-49869209 on Build GRCh38
rs142694534 Chr.12: 49869259-49869259 on Build GRCh38
rs181163662 Chr.12: 49869178-49869178 on Build GRCh38
rs1041300189 Chr.12: 49869290-49869290 on Build GRCh38
rs969247423 Chr.12: 49869186-49869186 on Build GRCh38
rs942944141 Chr.12: 49869358-49869358 on Build GRCh38
rs533846057 Chr.12: 49869043-49869043 on Build GRCh38
rs1007175347 Chr.12: 49869116-49869116 on Build GRCh38
rs796483052 Chr.12: 49869230-49869230 on Build GRCh38
rs575856668 Chr.12: 49869192-49869192 on Build GRCh38
rs573293676 Chr.12: 49869139-49869139 on Build GRCh38
rs952839607 Chr.12: 49869089-49869089 on Build GRCh38
rs890021318 Chr.12: 49869105-49869105 on Build GRCh38
rs35329567 Chr.12: 49869179-49869179 on Build GRCh38
rs994230575 Chr.12: 49869389-49869389 on Build GRCh38
rs7132908 Chr.12: 49869365-49869365 on Build GRCh38
rs75194253 Chr.12: 49869210-49869210 on Build GRCh38
rs780567477 Chr.12: 49869478-49869478 on Build GRCh38
rs76488009 Chr.12: 49869438-49869438 on Build GRCh38
rs1023400235 Chr.12: 49869480-49869480 on Build GRCh38
rs1053435396 Chr.12: 49869263-49869263 on Build GRCh38
rs568502300 Chr.12: 49869501-49869501 on Build GRCh38
rs756760675 Chr.12: 49869144-49869144 on Build GRCh38
rs942920630 Chr.12: 49869284-49869284 on Build GRCh38
rs561333964 Chr.12: 49869234-49869234 on Build GRCh38
rs191649983 Chr.12: 49869490-49869490 on Build GRCh38
rs907955571 Chr.12: 49869445-49869445 on Build GRCh38
rs984069314 Chr.12: 49869098-49869098 on Build GRCh38
rs993893408 Chr.12: 49869152-49869152 on Build GRCh38
rs1034343707 Chr.12: 49869146-49869146 on Build GRCh38
rs1034289977 Chr.12: 49869469-49869469 on Build GRCh38
rs896226651 Chr.12: 49869446-49869446 on Build GRCh38

View Full Product Details