Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51187815-51188310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POU6F1
Entrez Gene ID
5463
Gene Name
POU class 6 homeobox 1
Gene Aliases
BRN5, MPOU, TCFB1
UniGene
Hs.555886
Gene Chromosome Location
Chr.12: 51186936-51218134 on Build GRCh38
Transcripts
NM_002702.3 NR_026893.1 XM_017019507.1 XM_017019508.1 XM_017019509.1 XM_017019510.1 XM_017019511.1 XM_017019512.1 XM_017019513.1 XM_017019514.1 XM_017019515.1 XM_017019516.1 XM_017019517.1 XM_017019518.1 XM_017019519.1 XM_017019520.1 XM_017019521.1 XM_017019522.1 XM_017019523.1 XM_017019524.1 XM_017019525.1 XM_017019526.1 XM_017019527.1 XM_017019528.1 XM_017019529.1 XM_017019530.1

SNP Detail

SNP ID Chromosome Location
rs913208116 Chr.12: 51187981-51187981 on Build GRCh38
rs571233922 Chr.12: 51187931-51187931 on Build GRCh38
rs185869567 Chr.12: 51188208-51188208 on Build GRCh38
rs1053639633 Chr.12: 51188171-51188171 on Build GRCh38
rs904614782 Chr.12: 51187964-51187964 on Build GRCh38
rs1015016498 Chr.12: 51187848-51187848 on Build GRCh38
rs138938883 Chr.12: 51188217-51188217 on Build GRCh38
rs548962841 Chr.12: 51188086-51188086 on Build GRCh38
rs1053141561 Chr.12: 51188002-51188002 on Build GRCh38
rs948831061 Chr.12: 51188139-51188139 on Build GRCh38
rs550016955 Chr.12: 51188024-51188024 on Build GRCh38
rs962511596 Chr.12: 51188060-51188060 on Build GRCh38
rs980277843 Chr.12: 51188147-51188147 on Build GRCh38
rs963103150 Chr.12: 51187876-51187876 on Build GRCh38
rs1023422507 Chr.12: 51187908-51187908 on Build GRCh38
rs745742134 Chr.12: 51188221-51188221 on Build GRCh38
rs980269378 Chr.12: 51187923-51187923 on Build GRCh38
rs933990352 Chr.12: 51188185-51188185 on Build GRCh38
rs76213841 Chr.12: 51187930-51187930 on Build GRCh38
rs970180023 Chr.12: 51187915-51187915 on Build GRCh38
rs926001500 Chr.12: 51188148-51188148 on Build GRCh38
rs775854501 Chr.12: 51187958-51187958 on Build GRCh38
rs762613972 Chr.12: 51187875-51187875 on Build GRCh38
rs972918996 Chr.12: 51188072-51188072 on Build GRCh38
rs10876151 Chr.12: 51187957-51187957 on Build GRCh38
rs200488247 Chr.12: 51188117-51188118 on Build GRCh38
rs1006688128 Chr.12: 51188052-51188052 on Build GRCh38
rs570024767 Chr.12: 51188044-51188044 on Build GRCh38
rs367854102 Chr.12: 51187957-51187958 on Build GRCh38
rs1029575849 Chr.12: 51188030-51188030 on Build GRCh38
rs143971014 Chr.12: 51188075-51188075 on Build GRCh38
rs150676262 Chr.12: 51188194-51188194 on Build GRCh38
rs993092128 Chr.12: 51188081-51188081 on Build GRCh38
rs936079277 Chr.12: 51188170-51188170 on Build GRCh38
rs917414441 Chr.12: 51187934-51187934 on Build GRCh38
rs891812301 Chr.12: 51188023-51188023 on Build GRCh38
rs917507668 Chr.12: 51188117-51188117 on Build GRCh38
rs551218905 Chr.12: 51187895-51187895 on Build GRCh38
rs1035848631 Chr.12: 51188220-51188220 on Build GRCh38
rs994546439 Chr.12: 51188135-51188135 on Build GRCh38
rs35515176 Chr.12: 51188107-51188107 on Build GRCh38
rs1023766939 Chr.12: 51188142-51188142 on Build GRCh38
rs866120034 Chr.12: 51187994-51187994 on Build GRCh38
rs193099734 Chr.12: 51188180-51188180 on Build GRCh38
rs1036303949 Chr.12: 51188091-51188091 on Build GRCh38
rs960686937 Chr.12: 51188079-51188079 on Build GRCh38
rs1043529886 Chr.12: 51188022-51188022 on Build GRCh38
rs1000574020 Chr.12: 51187987-51187987 on Build GRCh38
rs772886922 Chr.12: 51187873-51187873 on Build GRCh38
rs929557141 Chr.12: 51187838-51187838 on Build GRCh38
rs528165153 Chr.12: 51187893-51187893 on Build GRCh38
rs111665877 Chr.12: 51188265-51188265 on Build GRCh38
rs754373382 Chr.12: 51188239-51188239 on Build GRCh38
rs1016611599 Chr.12: 51188053-51188053 on Build GRCh38

View Full Product Details