Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52493274-52493783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT6A
Entrez Gene ID
3853
Gene Name
keratin 6A
Gene Aliases
CK-6C, CK-6E, CK6A, CK6C, CK6D, K6A, K6C, K6D, KRT6C, KRT6D, PC3
UniGene
Hs.700779
Gene Chromosome Location
Chr.12: 52487174-52493397 on Build GRCh38
Transcripts
NM_005554.3

SNP Detail

SNP ID Chromosome Location
rs907841362 Chr.12: 52493684-52493684 on Build GRCh38
rs905652025 Chr.12: 52493497-52493497 on Build GRCh38
rs956142380 Chr.12: 52493484-52493484 on Build GRCh38
rs899301249 Chr.12: 52493439-52493439 on Build GRCh38
rs905840775 Chr.12: 52493680-52493680 on Build GRCh38
rs17104569 Chr.12: 52493679-52493679 on Build GRCh38
rs964434678 Chr.12: 52493583-52493583 on Build GRCh38
rs775749128 Chr.12: 52493572-52493572 on Build GRCh38
rs954626520 Chr.12: 52493538-52493538 on Build GRCh38
rs974052586 Chr.12: 52493710-52493710 on Build GRCh38
rs948220568 Chr.12: 52493745-52493745 on Build GRCh38
rs1010543827 Chr.12: 52493562-52493562 on Build GRCh38
rs973499296 Chr.12: 52493599-52493599 on Build GRCh38
rs528286530 Chr.12: 52493300-52493300 on Build GRCh38
rs920880137 Chr.12: 52493729-52493729 on Build GRCh38
rs550274063 Chr.12: 52493417-52493417 on Build GRCh38
rs891845878 Chr.12: 52493704-52493704 on Build GRCh38
rs774471929 Chr.12: 52493730-52493730 on Build GRCh38
rs891472622 Chr.12: 52493553-52493553 on Build GRCh38
rs1047885404 Chr.12: 52493661-52493661 on Build GRCh38
rs1002534928 Chr.12: 52493503-52493503 on Build GRCh38
rs773539879 Chr.12: 52493480-52493480 on Build GRCh38
rs1010317481 Chr.12: 52493728-52493728 on Build GRCh38
rs1014515863 Chr.12: 52493390-52493390 on Build GRCh38
rs908125641 Chr.12: 52493341-52493341 on Build GRCh38
rs866329578 Chr.12: 52493647-52493647 on Build GRCh38
rs1054876439 Chr.12: 52493690-52493690 on Build GRCh38
rs768877500 Chr.12: 52493693-52493693 on Build GRCh38
rs987383675 Chr.12: 52493566-52493566 on Build GRCh38
rs917460184 Chr.12: 52493663-52493663 on Build GRCh38
rs117946106 Chr.12: 52493406-52493406 on Build GRCh38
rs1000673667 Chr.12: 52493479-52493479 on Build GRCh38
rs567229558 Chr.12: 52493511-52493511 on Build GRCh38
rs535818052 Chr.12: 52493631-52493631 on Build GRCh38
rs954826787 Chr.12: 52493670-52493670 on Build GRCh38
rs777096774 Chr.12: 52493366-52493366 on Build GRCh38
rs761992918 Chr.12: 52493402-52493402 on Build GRCh38
rs769986114 Chr.12: 52493532-52493532 on Build GRCh38
rs551513949 Chr.12: 52493324-52493324 on Build GRCh38
rs1020737965 Chr.12: 52493753-52493753 on Build GRCh38
rs556013488 Chr.12: 52493664-52493664 on Build GRCh38
rs557851513 Chr.12: 52493708-52493708 on Build GRCh38
rs777708175 Chr.12: 52493427-52493427 on Build GRCh38
rs145819581 Chr.12: 52493689-52493689 on Build GRCh38
rs571319747 Chr.12: 52493329-52493329 on Build GRCh38

View Full Product Details