Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53253132-53253642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD5
Entrez Gene ID
84975
Gene Name
major facilitator superfamily domain containing 5
Gene Aliases
hsMOT2
UniGene
Hs.654660
Gene Chromosome Location
Chr.12: 53251251-53254406 on Build GRCh38
Transcripts
NM_001170790.1 NM_032889.4 XM_005269197.1 XM_005269198.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
292045 c.426C>T p.A142A Chr.12: 53253261-53253261 on Build GRCh38
6351887 c.754T>C p.L252L Chr.12: 53253589-53253589 on Build GRCh38
5361217 c.915delG p.P307fs*2 Chr.12: 53253429-53253429 on Build GRCh38
3871933 c.766G>A p.E256K Chr.12: 53253280-53253280 on Build GRCh38
7097827 c.651C>T p.L217L Chr.12: 53253165-53253165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755165824 Chr.12: 53253304-53253304 on Build GRCh38
rs200458134 Chr.12: 53253198-53253198 on Build GRCh38
rs200829817 Chr.12: 53253521-53253521 on Build GRCh38
rs531505170 Chr.12: 53253441-53253441 on Build GRCh38
rs767229928 Chr.12: 53253490-53253490 on Build GRCh38
rs768719528 Chr.12: 53253530-53253530 on Build GRCh38
rs770083749 Chr.12: 53253562-53253562 on Build GRCh38
rs373745086 Chr.12: 53253370-53253370 on Build GRCh38
rs149660360 Chr.12: 53253569-53253569 on Build GRCh38
rs751573232 Chr.12: 53253575-53253575 on Build GRCh38
rs762649843 Chr.12: 53253310-53253310 on Build GRCh38
rs750845106 Chr.12: 53253269-53253269 on Build GRCh38
rs761089465 Chr.12: 53253401-53253401 on Build GRCh38
rs781530496 Chr.12: 53253522-53253522 on Build GRCh38
rs761590101 Chr.12: 53253189-53253189 on Build GRCh38
rs759026171 Chr.12: 53253537-53253537 on Build GRCh38
rs772983521 Chr.12: 53253556-53253556 on Build GRCh38
rs367908569 Chr.12: 53253444-53253444 on Build GRCh38
rs150771447 Chr.12: 53253330-53253330 on Build GRCh38
rs766004805 Chr.12: 53253611-53253622 on Build GRCh38
rs528600211 Chr.12: 53253400-53253400 on Build GRCh38
rs770687144 Chr.12: 53253382-53253382 on Build GRCh38
rs772943235 Chr.12: 53253297-53253297 on Build GRCh38
rs772320341 Chr.12: 53253398-53253404 on Build GRCh38
rs267603536 Chr.12: 53253507-53253507 on Build GRCh38
rs772311798 Chr.12: 53253294-53253294 on Build GRCh38
rs747138972 Chr.12: 53253415-53253415 on Build GRCh38
rs377408812 Chr.12: 53253229-53253229 on Build GRCh38
rs761996974 Chr.12: 53253577-53253577 on Build GRCh38
rs766251464 Chr.12: 53253226-53253226 on Build GRCh38
rs776495680 Chr.12: 53253383-53253383 on Build GRCh38
rs748301666 Chr.12: 53253390-53253391 on Build GRCh38
rs748477955 Chr.12: 53253499-53253499 on Build GRCh38
rs762604625 Chr.12: 53253398-53253398 on Build GRCh38
rs765727048 Chr.12: 53253500-53253500 on Build GRCh38
rs778394694 Chr.12: 53253578-53253578 on Build GRCh38
rs778897799 Chr.12: 53253362-53253362 on Build GRCh38
rs781639190 Chr.12: 53253259-53253259 on Build GRCh38
rs759879003 Chr.12: 53253557-53253557 on Build GRCh38
rs765782010 Chr.12: 53253404-53253404 on Build GRCh38
rs10876422 Chr.12: 53253589-53253589 on Build GRCh38
rs763810518 Chr.12: 53253399-53253399 on Build GRCh38
rs530817927 Chr.12: 53253307-53253307 on Build GRCh38
rs748805187 Chr.12: 53253448-53253448 on Build GRCh38
rs577395856 Chr.12: 53253502-53253502 on Build GRCh38
rs768341612 Chr.12: 53253462-53253462 on Build GRCh38
rs773926474 Chr.12: 53253474-53253474 on Build GRCh38
rs368608562 Chr.12: 53253604-53253604 on Build GRCh38
rs780605054 Chr.12: 53253339-53253339 on Build GRCh38
rs201292974 Chr.12: 53253314-53253314 on Build GRCh38
rs750717532 Chr.12: 53253588-53253588 on Build GRCh38
rs759011096 Chr.12: 53253236-53253236 on Build GRCh38
rs370069764 Chr.12: 53253353-53253353 on Build GRCh38
rs745702668 Chr.12: 53253440-53253440 on Build GRCh38
rs775094756 Chr.12: 53253395-53253395 on Build GRCh38
rs767632709 Chr.12: 53253580-53253580 on Build GRCh38
rs757501586 Chr.12: 53253423-53253423 on Build GRCh38
rs563549259 Chr.12: 53253280-53253280 on Build GRCh38
rs764983507 Chr.12: 53253556-53253566 on Build GRCh38
rs199567120 Chr.12: 53253493-53253493 on Build GRCh38
rs199705102 Chr.12: 53253518-53253518 on Build GRCh38
rs764679069 Chr.12: 53253570-53253570 on Build GRCh38
rs770424708 Chr.12: 53253153-53253153 on Build GRCh38
rs752264052 Chr.12: 53253309-53253309 on Build GRCh38
rs756042872 Chr.12: 53253524-53253524 on Build GRCh38
rs181381216 Chr.12: 53253215-53253215 on Build GRCh38
rs780128042 Chr.12: 53253526-53253526 on Build GRCh38
rs748311439 Chr.12: 53253293-53253293 on Build GRCh38
rs749347175 Chr.12: 53253164-53253164 on Build GRCh38
rs750133740 Chr.12: 53253555-53253556 on Build GRCh38
rs961913269 Chr.12: 53253504-53253504 on Build GRCh38
rs771592541 Chr.12: 53253555-53253555 on Build GRCh38
rs778246487 Chr.12: 53253548-53253548 on Build GRCh38
rs753718042 Chr.12: 53253597-53253597 on Build GRCh38
rs754675488 Chr.12: 53253385-53253386 on Build GRCh38
rs145868928 Chr.12: 53253318-53253318 on Build GRCh38
rs754358858 Chr.12: 53253351-53253351 on Build GRCh38
rs773004522 Chr.12: 53253203-53253203 on Build GRCh38
rs758140779 Chr.12: 53253255-53253255 on Build GRCh38
rs749181599 Chr.12: 53253529-53253529 on Build GRCh38
rs777955343 Chr.12: 53253410-53253410 on Build GRCh38
rs763857729 Chr.12: 53253311-53253311 on Build GRCh38
rs775983106 Chr.12: 53253399-53253400 on Build GRCh38
rs752458422 Chr.12: 53253246-53253246 on Build GRCh38
rs36054910 Chr.12: 53253567-53253567 on Build GRCh38
rs988592678 Chr.12: 53253221-53253221 on Build GRCh38
rs754235430 Chr.12: 53253402-53253402 on Build GRCh38
rs752741759 Chr.12: 53253406-53253406 on Build GRCh38
rs781104091 Chr.12: 53253388-53253389 on Build GRCh38
rs551001881 Chr.12: 53253523-53253523 on Build GRCh38
rs775596714 Chr.12: 53253160-53253160 on Build GRCh38
rs768683940 Chr.12: 53253285-53253285 on Build GRCh38
rs762507257 Chr.12: 53253569-53253570 on Build GRCh38
rs761351551 Chr.12: 53253553-53253554 on Build GRCh38
rs768898335 Chr.12: 53253166-53253166 on Build GRCh38
rs938781390 Chr.12: 53253579-53253579 on Build GRCh38
rs769796737 Chr.12: 53253393-53253393 on Build GRCh38
rs746812903 Chr.12: 53253380-53253380 on Build GRCh38
rs564304409 Chr.12: 53253375-53253375 on Build GRCh38
rs1019320608 Chr.12: 53253496-53253496 on Build GRCh38
rs139258866 Chr.12: 53253409-53253409 on Build GRCh38
rs755391705 Chr.12: 53253403-53253403 on Build GRCh38
rs774476002 Chr.12: 53253183-53253183 on Build GRCh38
rs201678997 Chr.12: 53253517-53253517 on Build GRCh38
rs370578711 Chr.12: 53253489-53253489 on Build GRCh38
rs778009674 Chr.12: 53253376-53253376 on Build GRCh38
rs145958473 Chr.12: 53253494-53253494 on Build GRCh38
rs779929287 Chr.12: 53253594-53253594 on Build GRCh38
rs756592209 Chr.12: 53253279-53253279 on Build GRCh38
rs779092319 Chr.12: 53253292-53253292 on Build GRCh38
rs769975222 Chr.12: 53253162-53253162 on Build GRCh38
rs911171862 Chr.12: 53253328-53253328 on Build GRCh38
rs776501438 Chr.12: 53253305-53253305 on Build GRCh38
rs760514479 Chr.12: 53253303-53253303 on Build GRCh38
rs780805200 Chr.12: 53253438-53253438 on Build GRCh38
rs148269175 Chr.12: 53253241-53253241 on Build GRCh38
rs760092178 Chr.12: 53253398-53253402 on Build GRCh38
rs376991225 Chr.12: 53253283-53253283 on Build GRCh38
rs938645252 Chr.12: 53253208-53253208 on Build GRCh38
rs747742369 Chr.12: 53253553-53253553 on Build GRCh38
rs1037687482 Chr.12: 53253337-53253337 on Build GRCh38
rs929351919 Chr.12: 53253381-53253381 on Build GRCh38

View Full Product Details