Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54644513-54645023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCD
Entrez Gene ID
117159
Gene Name
dermcidin
Gene Aliases
AIDD, DCD-1, DSEP, HCAP, PIF
UniGene
Hs.350570
Gene Chromosome Location
Chr.12: 54644591-54648493 on Build GRCh38
Transcripts
NM_001300854.1 NM_053283.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
70403 c.319G>C p.D107H Chr.12: 54644727-54644727 on Build GRCh38
6311004 c.298C>A p.H100N Chr.12: 54644748-54644748 on Build GRCh38
7159804 c.304G>A p.V102I Chr.12: 54644742-54644742 on Build GRCh38
3462886 c.328C>T p.L110L Chr.12: 54644718-54644718 on Build GRCh38
3462887 c.315C>T p.V105V Chr.12: 54644731-54644731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745977741 Chr.12: 54644710-54644710 on Build GRCh38
rs781595221 Chr.12: 54644770-54644770 on Build GRCh38
rs954138007 Chr.12: 54644731-54644731 on Build GRCh38
rs774594952 Chr.12: 54644790-54644790 on Build GRCh38
rs7301225 Chr.12: 54644920-54644920 on Build GRCh38
rs952019773 Chr.12: 54644557-54644557 on Build GRCh38
rs56013373 Chr.12: 54644598-54644598 on Build GRCh38
rs962375303 Chr.12: 54644811-54644811 on Build GRCh38
rs775320876 Chr.12: 54644819-54644819 on Build GRCh38
rs569588793 Chr.12: 54644990-54644990 on Build GRCh38
rs771879520 Chr.12: 54644793-54644793 on Build GRCh38
rs373661792 Chr.12: 54644743-54644743 on Build GRCh38
rs759094075 Chr.12: 54644996-54644996 on Build GRCh38
rs771365688 Chr.12: 54644690-54644690 on Build GRCh38
rs767916837 Chr.12: 54644882-54644882 on Build GRCh38
rs1050553362 Chr.12: 54644979-54644979 on Build GRCh38
rs932226321 Chr.12: 54644964-54644964 on Build GRCh38
rs781696713 Chr.12: 54644693-54644693 on Build GRCh38
rs1029124458 Chr.12: 54644538-54644538 on Build GRCh38
rs369155507 Chr.12: 54644663-54644663 on Build GRCh38
rs760602440 Chr.12: 54644800-54644800 on Build GRCh38
rs11338265 Chr.12: 54644647-54644647 on Build GRCh38
rs749521523 Chr.12: 54644715-54644715 on Build GRCh38
rs369454217 Chr.12: 54644675-54644675 on Build GRCh38
rs868112354 Chr.12: 54644733-54644733 on Build GRCh38
rs766246933 Chr.12: 54644836-54644836 on Build GRCh38
rs943743153 Chr.12: 54644925-54644925 on Build GRCh38
rs961957735 Chr.12: 54644586-54644586 on Build GRCh38
rs1056689784 Chr.12: 54644878-54644878 on Build GRCh38
rs1015355356 Chr.12: 54644584-54644584 on Build GRCh38
rs775554082 Chr.12: 54644780-54644780 on Build GRCh38
rs775916647 Chr.12: 54644711-54644711 on Build GRCh38
rs769025831 Chr.12: 54644787-54644787 on Build GRCh38
rs760381297 Chr.12: 54644842-54644843 on Build GRCh38
rs776140135 Chr.12: 54644841-54644841 on Build GRCh38
rs377645047 Chr.12: 54644666-54644666 on Build GRCh38
rs756706265 Chr.12: 54644897-54644897 on Build GRCh38
rs73112792 Chr.12: 54644842-54644842 on Build GRCh38
rs757617852 Chr.12: 54644871-54644871 on Build GRCh38
rs914288292 Chr.12: 54644651-54644651 on Build GRCh38
rs958571226 Chr.12: 54644616-54644616 on Build GRCh38
rs758820272 Chr.12: 54644679-54644679 on Build GRCh38
rs780661574 Chr.12: 54644899-54644899 on Build GRCh38
rs938275576 Chr.12: 54644550-54644550 on Build GRCh38
rs77605968 Chr.12: 54644742-54644742 on Build GRCh38
rs923484452 Chr.12: 54644945-54644945 on Build GRCh38
rs1006729670 Chr.12: 54644807-54644807 on Build GRCh38
rs764835374 Chr.12: 54644843-54644843 on Build GRCh38
rs1040643364 Chr.12: 54644930-54644930 on Build GRCh38
rs762354994 Chr.12: 54644844-54644849 on Build GRCh38
rs773083042 Chr.12: 54644797-54644797 on Build GRCh38
rs757608223 Chr.12: 54644763-54644763 on Build GRCh38
rs1042472807 Chr.12: 54644622-54644622 on Build GRCh38
rs139716642 Chr.12: 54644752-54644752 on Build GRCh38
rs991242280 Chr.12: 54644883-54644883 on Build GRCh38
rs1014911613 Chr.12: 54644810-54644810 on Build GRCh38
rs778115220 Chr.12: 54644681-54644681 on Build GRCh38
rs765861179 Chr.12: 54644750-54644750 on Build GRCh38
rs1006190159 Chr.12: 54644574-54644574 on Build GRCh38
rs746757901 Chr.12: 54644944-54644944 on Build GRCh38
rs547857899 Chr.12: 54644901-54644901 on Build GRCh38
rs1002789379 Chr.12: 54644818-54644818 on Build GRCh38
rs560030993 Chr.12: 54644734-54644734 on Build GRCh38
rs755608119 Chr.12: 54644671-54644671 on Build GRCh38
rs770885111 Chr.12: 54644947-54644947 on Build GRCh38
rs150921024 Chr.12: 54644918-54644918 on Build GRCh38
rs73310828 Chr.12: 54644792-54644792 on Build GRCh38
rs199825782 Chr.12: 54644785-54644785 on Build GRCh38
rs938273257 Chr.12: 54644876-54644876 on Build GRCh38
rs796644696 Chr.12: 54644623-54644623 on Build GRCh38
rs532438783 Chr.12: 54644896-54644896 on Build GRCh38
rs879117946 Chr.12: 54644625-54644626 on Build GRCh38
rs765479391 Chr.12: 54644976-54644976 on Build GRCh38
rs539008225 Chr.12: 54644923-54644923 on Build GRCh38
rs774931561 Chr.12: 54644625-54644627 on Build GRCh38
rs749986897 Chr.12: 54644626-54644627 on Build GRCh38
rs567531486 Chr.12: 54644941-54644941 on Build GRCh38
rs532310980 Chr.12: 54644741-54644741 on Build GRCh38
rs780289515 Chr.12: 54644778-54644778 on Build GRCh38
rs200535792 Chr.12: 54644761-54644761 on Build GRCh38
rs756325842 Chr.12: 54644776-54644776 on Build GRCh38
rs928451295 Chr.12: 54644612-54644612 on Build GRCh38
rs761970067 Chr.12: 54644975-54644976 on Build GRCh38
rs143447971 Chr.12: 54644665-54644665 on Build GRCh38
rs758805368 Chr.12: 54644849-54644850 on Build GRCh38
rs746369300 Chr.12: 54644772-54644772 on Build GRCh38
rs199541255 Chr.12: 54644751-54644751 on Build GRCh38
rs984964542 Chr.12: 54644865-54644865 on Build GRCh38
rs905489147 Chr.12: 54644539-54644539 on Build GRCh38
rs536287837 Chr.12: 54644843-54644844 on Build GRCh38
rs914425854 Chr.12: 54644898-54644898 on Build GRCh38
rs752369587 Chr.12: 54644869-54644869 on Build GRCh38
rs1049591734 Chr.12: 54644937-54644937 on Build GRCh38
rs747340822 Chr.12: 54644688-54644688 on Build GRCh38

View Full Product Details