Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54857978-54858491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUCL1
Entrez Gene ID
118430
Gene Name
mucin like 1
Gene Aliases
SBEM
UniGene
Hs.348419
Gene Chromosome Location
Chr.12: 54854515-54858393 on Build GRCh38
Transcripts
NM_058173.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3462894 c.235T>G p.W79G Chr.12: 54858204-54858204 on Build GRCh38
6718185 c.247C>T p.L83F Chr.12: 54858216-54858216 on Build GRCh38
3462897 c.272G>A p.*91* Chr.12: 54858241-54858241 on Build GRCh38
5545347 c.224-1G>A p.? Chr.12: 54858192-54858192 on Build GRCh38
3462895 c.237G>A p.W79* Chr.12: 54858206-54858206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756454064 Chr.12: 54858159-54858159 on Build GRCh38
rs573642425 Chr.12: 54858406-54858406 on Build GRCh38
rs759372635 Chr.12: 54858272-54858272 on Build GRCh38
rs774459388 Chr.12: 54858276-54858278 on Build GRCh38
rs539263727 Chr.12: 54858220-54858220 on Build GRCh38
rs566791248 Chr.12: 54858223-54858223 on Build GRCh38
rs893434688 Chr.12: 54858051-54858051 on Build GRCh38
rs1020325031 Chr.12: 54858103-54858103 on Build GRCh38
rs1001916534 Chr.12: 54858039-54858039 on Build GRCh38
rs1050064910 Chr.12: 54858062-54858062 on Build GRCh38
rs767941898 Chr.12: 54858151-54858151 on Build GRCh38
rs964154725 Chr.12: 54858409-54858409 on Build GRCh38
rs375464814 Chr.12: 54858126-54858126 on Build GRCh38
rs755513344 Chr.12: 54858237-54858237 on Build GRCh38
rs138235110 Chr.12: 54858221-54858221 on Build GRCh38
rs753659452 Chr.12: 54858172-54858172 on Build GRCh38
rs903434467 Chr.12: 54858376-54858376 on Build GRCh38
rs553671545 Chr.12: 54858373-54858373 on Build GRCh38
rs772183799 Chr.12: 54858257-54858257 on Build GRCh38
rs776504221 Chr.12: 54858212-54858212 on Build GRCh38
rs897433655 Chr.12: 54858328-54858328 on Build GRCh38
rs187155476 Chr.12: 54858416-54858416 on Build GRCh38
rs1008399605 Chr.12: 54858388-54858388 on Build GRCh38
rs771221611 Chr.12: 54858269-54858269 on Build GRCh38
rs370851754 Chr.12: 54858175-54858175 on Build GRCh38
rs996910373 Chr.12: 54858411-54858411 on Build GRCh38
rs747239549 Chr.12: 54858268-54858268 on Build GRCh38
rs908501331 Chr.12: 54858425-54858425 on Build GRCh38
rs913186667 Chr.12: 54858185-54858185 on Build GRCh38
rs766274450 Chr.12: 54858206-54858206 on Build GRCh38
rs780407333 Chr.12: 54858228-54858228 on Build GRCh38
rs1008807306 Chr.12: 54858085-54858085 on Build GRCh38
rs769091768 Chr.12: 54858164-54858164 on Build GRCh38
rs1011852074 Chr.12: 54858058-54858058 on Build GRCh38
rs200845354 Chr.12: 54858270-54858270 on Build GRCh38
rs867390271 Chr.12: 54858238-54858238 on Build GRCh38
rs767045892 Chr.12: 54858065-54858065 on Build GRCh38
rs978681607 Chr.12: 54858424-54858424 on Build GRCh38
rs763660147 Chr.12: 54858283-54858283 on Build GRCh38
rs756832073 Chr.12: 54858226-54858226 on Build GRCh38
rs753053136 Chr.12: 54858177-54858177 on Build GRCh38
rs750973715 Chr.12: 54858287-54858287 on Build GRCh38
rs983185590 Chr.12: 54858420-54858420 on Build GRCh38
rs373133585 Chr.12: 54858150-54858150 on Build GRCh38
rs1042821081 Chr.12: 54858030-54858030 on Build GRCh38
rs945902239 Chr.12: 54858012-54858012 on Build GRCh38
rs113433295 Chr.12: 54858227-54858227 on Build GRCh38
rs375718668 Chr.12: 54858214-54858214 on Build GRCh38
rs765136019 Chr.12: 54858273-54858273 on Build GRCh38
rs1042008202 Chr.12: 54858367-54858367 on Build GRCh38
rs773373990 Chr.12: 54858261-54858261 on Build GRCh38
rs752201508 Chr.12: 54858149-54858149 on Build GRCh38
rs555435001 Chr.12: 54858395-54858395 on Build GRCh38
rs1034889509 Chr.12: 54858392-54858392 on Build GRCh38
rs76751755 Chr.12: 54858419-54858419 on Build GRCh38
rs202045449 Chr.12: 54858173-54858173 on Build GRCh38
rs1008439958 Chr.12: 54858096-54858096 on Build GRCh38
rs200001020 Chr.12: 54858176-54858176 on Build GRCh38
rs538619675 Chr.12: 54858282-54858282 on Build GRCh38
rs927284106 Chr.12: 54858176-54858177 on Build GRCh38
rs375293752 Chr.12: 54858152-54858152 on Build GRCh38
rs760605577 Chr.12: 54858190-54858190 on Build GRCh38
rs772226522 Chr.12: 54858178-54858178 on Build GRCh38
rs750382208 Chr.12: 54858094-54858094 on Build GRCh38
rs1020938601 Chr.12: 54858067-54858067 on Build GRCh38
rs60749539 Chr.12: 54858428-54858428 on Build GRCh38
rs779446904 Chr.12: 54858253-54858253 on Build GRCh38
rs1057423114 Chr.12: 54857999-54857999 on Build GRCh38
rs555972184 Chr.12: 54858435-54858435 on Build GRCh38
rs2168139 Chr.12: 54858000-54858000 on Build GRCh38
rs866236928 Chr.12: 54858297-54858297 on Build GRCh38
rs1948139 Chr.12: 54858418-54858418 on Build GRCh38
rs112968647 Chr.12: 54858083-54858083 on Build GRCh38
rs539463530 Chr.12: 54858429-54858429 on Build GRCh38
rs79678008 Chr.12: 54858461-54858461 on Build GRCh38
rs1041266979 Chr.12: 54858325-54858325 on Build GRCh38
rs2306849 Chr.12: 54858050-54858050 on Build GRCh38
rs774320240 Chr.12: 54858174-54858174 on Build GRCh38
rs985891918 Chr.12: 54858454-54858454 on Build GRCh38
rs749603693 Chr.12: 54858236-54858236 on Build GRCh38
rs1046267443 Chr.12: 54858251-54858251 on Build GRCh38
rs150470196 Chr.12: 54858153-54858153 on Build GRCh38
rs773466348 Chr.12: 54858182-54858182 on Build GRCh38
rs960317765 Chr.12: 54858177-54858177 on Build GRCh38
rs778572567 Chr.12: 54858143-54858143 on Build GRCh38
rs749797825 Chr.12: 54858160-54858160 on Build GRCh38

View Full Product Details