Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55681852-55682349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL7B
Entrez Gene ID
196410
Gene Name
methyltransferase like 7B
Gene Aliases
ALDI
UniGene
Hs.51483
Gene Chromosome Location
Chr.12: 55681546-55684611 on Build GRCh38
Transcripts
NM_152637.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6988078 c.453G>T p.Q151H Chr.12: 55682207-55682207 on Build GRCh38
6052576 c.344G>T p.R115I Chr.12: 55682248-55682248 on Build GRCh38
3463156 c.415G>A p.D139N Chr.12: 55682169-55682169 on Build GRCh38
3463157 c.265G>A p.D89N Chr.12: 55682169-55682169 on Build GRCh38
3463153 c.94G>A p.G32R Chr.12: 55681998-55681998 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768410706 Chr.12: 55682290-55682290 on Build GRCh38
rs774200244 Chr.12: 55682295-55682295 on Build GRCh38
rs574128558 Chr.12: 55682272-55682272 on Build GRCh38
rs75289684 Chr.12: 55682131-55682131 on Build GRCh38
rs542381721 Chr.12: 55682318-55682318 on Build GRCh38
rs111293312 Chr.12: 55682281-55682281 on Build GRCh38
rs1006918177 Chr.12: 55682240-55682240 on Build GRCh38
rs770736277 Chr.12: 55682023-55682023 on Build GRCh38
rs138407179 Chr.12: 55681992-55681992 on Build GRCh38
rs753513397 Chr.12: 55681969-55681969 on Build GRCh38
rs749675020 Chr.12: 55681937-55681937 on Build GRCh38
rs975941682 Chr.12: 55681988-55681988 on Build GRCh38
rs758630679 Chr.12: 55682089-55682089 on Build GRCh38
rs997346591 Chr.12: 55682305-55682305 on Build GRCh38
rs762519717 Chr.12: 55681921-55681921 on Build GRCh38
rs772481213 Chr.12: 55682016-55682016 on Build GRCh38
rs773492285 Chr.12: 55681960-55681960 on Build GRCh38
rs760734677 Chr.12: 55681879-55681879 on Build GRCh38
rs555960398 Chr.12: 55682251-55682251 on Build GRCh38
rs116592765 Chr.12: 55682222-55682222 on Build GRCh38
rs772247957 Chr.12: 55681953-55681953 on Build GRCh38
rs752320926 Chr.12: 55681976-55681976 on Build GRCh38
rs143952871 Chr.12: 55682022-55682022 on Build GRCh38
rs751128037 Chr.12: 55681991-55681991 on Build GRCh38
rs146536505 Chr.12: 55681877-55681877 on Build GRCh38
rs773371475 Chr.12: 55682020-55682020 on Build GRCh38
rs185374665 Chr.12: 55681899-55681899 on Build GRCh38
rs756820350 Chr.12: 55682057-55682057 on Build GRCh38
rs780267588 Chr.12: 55681932-55681932 on Build GRCh38
rs368189344 Chr.12: 55682133-55682133 on Build GRCh38
rs749959695 Chr.12: 55682190-55682190 on Build GRCh38
rs1024688495 Chr.12: 55682068-55682068 on Build GRCh38
rs765848131 Chr.12: 55682207-55682207 on Build GRCh38
rs755456149 Chr.12: 55682200-55682200 on Build GRCh38
rs770402613 Chr.12: 55682116-55682116 on Build GRCh38
rs767051732 Chr.12: 55682154-55682154 on Build GRCh38
rs139449707 Chr.12: 55682127-55682127 on Build GRCh38
rs763695166 Chr.12: 55682135-55682135 on Build GRCh38
rs1026568401 Chr.12: 55682306-55682306 on Build GRCh38
rs747113874 Chr.12: 55681960-55681960 on Build GRCh38
rs762176191 Chr.12: 55681951-55681951 on Build GRCh38
rs546990666 Chr.12: 55682241-55682241 on Build GRCh38
rs781314085 Chr.12: 55682258-55682258 on Build GRCh38
rs201358277 Chr.12: 55682223-55682223 on Build GRCh38
rs766987012 Chr.12: 55682059-55682059 on Build GRCh38
rs904701086 Chr.12: 55682051-55682051 on Build GRCh38
rs144071589 Chr.12: 55682252-55682252 on Build GRCh38
rs777309449 Chr.12: 55681908-55681908 on Build GRCh38
rs1044107498 Chr.12: 55682123-55682123 on Build GRCh38
rs147300461 Chr.12: 55682042-55682042 on Build GRCh38
rs150913422 Chr.12: 55682085-55682085 on Build GRCh38
rs956469846 Chr.12: 55682061-55682062 on Build GRCh38
rs775347836 Chr.12: 55681883-55681883 on Build GRCh38
rs763551912 Chr.12: 55681927-55681927 on Build GRCh38
rs141569755 Chr.12: 55682064-55682064 on Build GRCh38
rs375664659 Chr.12: 55682063-55682063 on Build GRCh38
rs139209159 Chr.12: 55682235-55682235 on Build GRCh38
rs950635144 Chr.12: 55682319-55682319 on Build GRCh38
rs1016422404 Chr.12: 55682094-55682106 on Build GRCh38
rs544762153 Chr.12: 55682282-55682282 on Build GRCh38
rs779323040 Chr.12: 55682083-55682083 on Build GRCh38
rs1000839579 Chr.12: 55682317-55682317 on Build GRCh38
rs758082916 Chr.12: 55681978-55681978 on Build GRCh38
rs761414940 Chr.12: 55682147-55682147 on Build GRCh38
rs778175797 Chr.12: 55682097-55682097 on Build GRCh38
rs1053738126 Chr.12: 55682136-55682136 on Build GRCh38
rs762580730 Chr.12: 55682047-55682047 on Build GRCh38
rs980679753 Chr.12: 55681900-55681900 on Build GRCh38
rs201074903 Chr.12: 55682050-55682050 on Build GRCh38
rs759180049 Chr.12: 55682173-55682173 on Build GRCh38
rs377765746 Chr.12: 55682266-55682266 on Build GRCh38
rs779187102 Chr.12: 55682002-55682002 on Build GRCh38
rs758859367 Chr.12: 55682229-55682229 on Build GRCh38
rs544205263 Chr.12: 55682115-55682115 on Build GRCh38
rs760681089 Chr.12: 55681961-55681961 on Build GRCh38
rs754532522 Chr.12: 55681892-55681892 on Build GRCh38
rs147055776 Chr.12: 55681981-55681981 on Build GRCh38
rs140926536 Chr.12: 55681972-55681972 on Build GRCh38
rs769045151 Chr.12: 55681999-55681999 on Build GRCh38
rs889807347 Chr.12: 55682238-55682238 on Build GRCh38
rs768692231 Chr.12: 55681940-55681940 on Build GRCh38
rs746339120 Chr.12: 55681920-55681920 on Build GRCh38
rs768988830 Chr.12: 55682172-55682174 on Build GRCh38
rs776649757 Chr.12: 55682025-55682025 on Build GRCh38
rs766276829 Chr.12: 55681881-55681881 on Build GRCh38
rs757923363 Chr.12: 55681904-55681904 on Build GRCh38
rs74421938 Chr.12: 55682056-55682056 on Build GRCh38
rs748377831 Chr.12: 55682012-55682012 on Build GRCh38
rs931158232 Chr.12: 55682117-55682117 on Build GRCh38
rs537623851 Chr.12: 55682280-55682280 on Build GRCh38
rs752105398 Chr.12: 55681901-55681901 on Build GRCh38
rs575068928 Chr.12: 55682149-55682149 on Build GRCh38
rs749574114 Chr.12: 55681998-55681998 on Build GRCh38
rs981105890 Chr.12: 55682109-55682109 on Build GRCh38
rs367964396 Chr.12: 55682036-55682036 on Build GRCh38
rs149358153 Chr.12: 55681964-55681964 on Build GRCh38
rs375169143 Chr.12: 55682043-55682043 on Build GRCh38
rs774658111 Chr.12: 55681943-55681943 on Build GRCh38
rs965848463 Chr.12: 55681980-55681980 on Build GRCh38
rs747039214 Chr.12: 55682098-55682098 on Build GRCh38
rs181513519 Chr.12: 55682312-55682312 on Build GRCh38
rs755504620 Chr.12: 55682082-55682082 on Build GRCh38
rs892009546 Chr.12: 55682216-55682216 on Build GRCh38
rs748541304 Chr.12: 55682088-55682088 on Build GRCh38
rs774012372 Chr.12: 55682139-55682139 on Build GRCh38

View Full Product Details