Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56712647-56713185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NACA
Entrez Gene ID
4666
Gene Name
nascent polypeptide-associated complex alpha subunit
Gene Aliases
HSD48, NAC-alpha, NACA1, skNAC
UniGene
Hs.505735
Gene Chromosome Location
Chr.12: 56712427-56726275 on Build GRCh38
Transcripts
NM_001113201.2 NM_001113202.1 NM_001113203.2 NM_001320193.1 NM_001320194.1 NM_005594.5 XM_006719412.1 XM_006719413.2 XM_006719414.1 XM_006719418.1 XM_006719420.3 XM_011538389.1 XM_017019337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6336547 c.2526C>T p.S842S Chr.12: 56713176-56713176 on Build GRCh38
5739691 c.582G>T p.K194N Chr.12: 56712837-56712837 on Build GRCh38
6336544 c.2527C>G p.Q843E Chr.12: 56713175-56713175 on Build GRCh38
1180644 c.2674G>C p.E892Q Chr.12: 56712875-56712875 on Build GRCh38
3417013 c.6179G>A p.R2060Q Chr.12: 56712829-56712829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773075256 Chr.12: 56712783-56712783 on Build GRCh38
rs372164873 Chr.12: 56713043-56713043 on Build GRCh38
rs186711868 Chr.12: 56712914-56712914 on Build GRCh38
rs1034010346 Chr.12: 56712990-56712990 on Build GRCh38
rs759903795 Chr.12: 56712946-56712948 on Build GRCh38
rs1033168651 Chr.12: 56712714-56712714 on Build GRCh38
rs772283432 Chr.12: 56712736-56712736 on Build GRCh38
rs140448348 Chr.12: 56713086-56713086 on Build GRCh38
rs898609 Chr.12: 56713026-56713026 on Build GRCh38
rs765004308 Chr.12: 56712872-56712872 on Build GRCh38
rs61741738 Chr.12: 56713122-56713122 on Build GRCh38
rs779462506 Chr.12: 56712723-56712723 on Build GRCh38
rs759347986 Chr.12: 56712846-56712846 on Build GRCh38
rs772163703 Chr.12: 56712774-56712774 on Build GRCh38
rs757894002 Chr.12: 56712879-56712879 on Build GRCh38
rs774751792 Chr.12: 56712918-56712919 on Build GRCh38
rs754316696 Chr.12: 56713081-56713081 on Build GRCh38
rs556881553 Chr.12: 56713020-56713020 on Build GRCh38
rs879091640 Chr.12: 56713059-56713059 on Build GRCh38
rs753691485 Chr.12: 56712827-56712827 on Build GRCh38
rs777322987 Chr.12: 56712893-56712893 on Build GRCh38
rs760650214 Chr.12: 56712792-56712792 on Build GRCh38
rs779325302 Chr.12: 56713096-56713096 on Build GRCh38
rs766346119 Chr.12: 56712804-56712804 on Build GRCh38
rs149732816 Chr.12: 56712905-56712905 on Build GRCh38
rs780390926 Chr.12: 56712735-56712735 on Build GRCh38
rs4788 Chr.12: 56712876-56712876 on Build GRCh38
rs955037728 Chr.12: 56713056-56713056 on Build GRCh38
rs556518624 Chr.12: 56712961-56712961 on Build GRCh38
rs936969767 Chr.12: 56713097-56713097 on Build GRCh38
rs200124165 Chr.12: 56713029-56713029 on Build GRCh38
rs986355282 Chr.12: 56713057-56713057 on Build GRCh38
rs761834240 Chr.12: 56712756-56712756 on Build GRCh38
rs749671400 Chr.12: 56712915-56712915 on Build GRCh38
rs527361962 Chr.12: 56713049-56713049 on Build GRCh38
rs146755279 Chr.12: 56713079-56713079 on Build GRCh38
rs1002075686 Chr.12: 56712959-56712959 on Build GRCh38
rs534417377 Chr.12: 56712906-56712906 on Build GRCh38
rs779150027 Chr.12: 56712948-56712948 on Build GRCh38
rs375914904 Chr.12: 56713028-56713028 on Build GRCh38
rs201182580 Chr.12: 56712957-56712957 on Build GRCh38
rs368635646 Chr.12: 56712951-56712951 on Build GRCh38
rs748502679 Chr.12: 56712727-56712727 on Build GRCh38
rs768875387 Chr.12: 56712747-56712747 on Build GRCh38
rs936333037 Chr.12: 56712743-56712743 on Build GRCh38
rs933549764 Chr.12: 56712710-56712710 on Build GRCh38
rs748884842 Chr.12: 56712750-56712750 on Build GRCh38
rs771007076 Chr.12: 56713025-56713025 on Build GRCh38
rs923451652 Chr.12: 56712697-56712697 on Build GRCh38
rs544951704 Chr.12: 56712967-56712967 on Build GRCh38
rs1009287289 Chr.12: 56712917-56712917 on Build GRCh38
rs542829900 Chr.12: 56713071-56713071 on Build GRCh38
rs763666769 Chr.12: 56712957-56712959 on Build GRCh38
rs753523188 Chr.12: 56713025-56713025 on Build GRCh38
rs894486492 Chr.12: 56713082-56713082 on Build GRCh38
rs920956901 Chr.12: 56712807-56712807 on Build GRCh38
rs200078093 Chr.12: 56712741-56712741 on Build GRCh38
rs748387766 Chr.12: 56713146-56713146 on Build GRCh38
rs975643699 Chr.12: 56712751-56712751 on Build GRCh38
rs939766615 Chr.12: 56712996-56712996 on Build GRCh38
rs202147902 Chr.12: 56713010-56713010 on Build GRCh38
rs756703844 Chr.12: 56713068-56713068 on Build GRCh38
rs972197813 Chr.12: 56712692-56712692 on Build GRCh38
rs755291310 Chr.12: 56712933-56712933 on Build GRCh38

View Full Product Details