Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 62603335-62603835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01465
Entrez Gene ID
283416
Gene Name
long intergenic non-protein coding RNA 1465
Gene Aliases
C12orf61
UniGene
Hs.375881
Gene Chromosome Location
Chr.12: 62601751-62603434 on Build GRCh38
Transcripts
NR_121682.1 NR_029661.1

Gene Symbol
MIRLET7I
Entrez Gene ID
406891
Gene Name
microRNA let-7i
Gene Aliases
LET7I, MIRNLET7I, hsa-let-7i, let-7i
UniGene
Gene Chromosome Location
Chr.12: 62603686-62603769 on Build GRCh38
Transcripts
NR_121682.1 NR_029661.1

SNP Detail

SNP ID Chromosome Location
rs758294531 Chr.12: 62603768-62603768 on Build GRCh38
rs1037277511 Chr.12: 62603659-62603659 on Build GRCh38
rs1033929277 Chr.12: 62603374-62603374 on Build GRCh38
rs532384943 Chr.12: 62603604-62603604 on Build GRCh38
rs547302253 Chr.12: 62603397-62603397 on Build GRCh38
rs374744792 Chr.12: 62603755-62603755 on Build GRCh38
rs370685868 Chr.12: 62603417-62603417 on Build GRCh38
rs944627822 Chr.12: 62603426-62603426 on Build GRCh38
rs953908445 Chr.12: 62603479-62603479 on Build GRCh38
rs1056650351 Chr.12: 62603415-62603415 on Build GRCh38
rs961327234 Chr.12: 62603471-62603471 on Build GRCh38
rs536001911 Chr.12: 62603402-62603402 on Build GRCh38
rs749562233 Chr.12: 62603411-62603411 on Build GRCh38
rs373300999 Chr.12: 62603730-62603730 on Build GRCh38
rs750854486 Chr.12: 62603713-62603713 on Build GRCh38
rs748411870 Chr.12: 62603815-62603815 on Build GRCh38
rs1041432016 Chr.12: 62603567-62603594 on Build GRCh38
rs572685822 Chr.12: 62603525-62603525 on Build GRCh38
rs567356055 Chr.12: 62603782-62603782 on Build GRCh38
rs779963385 Chr.12: 62603635-62603635 on Build GRCh38
rs559310321 Chr.12: 62603497-62603497 on Build GRCh38
rs887756665 Chr.12: 62603444-62603444 on Build GRCh38
rs924147793 Chr.12: 62603564-62603564 on Build GRCh38
rs1056618686 Chr.12: 62603584-62603584 on Build GRCh38
rs1040155553 Chr.12: 62603432-62603432 on Build GRCh38
rs796184182 Chr.12: 62603400-62603401 on Build GRCh38
rs1005260002 Chr.12: 62603634-62603634 on Build GRCh38
rs769835716 Chr.12: 62603668-62603668 on Build GRCh38
rs897033216 Chr.12: 62603655-62603655 on Build GRCh38
rs369078761 Chr.12: 62603801-62603801 on Build GRCh38
rs887904946 Chr.12: 62603633-62603633 on Build GRCh38
rs186694256 Chr.12: 62603566-62603566 on Build GRCh38
rs547658149 Chr.12: 62603690-62603690 on Build GRCh38
rs118126121 Chr.12: 62603804-62603804 on Build GRCh38
rs552186135 Chr.12: 62603631-62603631 on Build GRCh38
rs370128294 Chr.12: 62603674-62603674 on Build GRCh38
rs543513304 Chr.12: 62603575-62603575 on Build GRCh38
rs1022429218 Chr.12: 62603750-62603750 on Build GRCh38
rs985157525 Chr.12: 62603483-62603483 on Build GRCh38
rs1049612014 Chr.12: 62603629-62603629 on Build GRCh38
rs751365433 Chr.12: 62603800-62603800 on Build GRCh38
rs895210928 Chr.12: 62603422-62603422 on Build GRCh38
rs939464186 Chr.12: 62603401-62603401 on Build GRCh38
rs766127565 Chr.12: 62603789-62603789 on Build GRCh38
rs747189338 Chr.12: 62603656-62603656 on Build GRCh38
rs937768467 Chr.12: 62603602-62603602 on Build GRCh38
rs748215585 Chr.12: 62603665-62603665 on Build GRCh38
rs776491984 Chr.12: 62603734-62603734 on Build GRCh38
rs763423413 Chr.12: 62603683-62603683 on Build GRCh38
rs985903171 Chr.12: 62603414-62603414 on Build GRCh38
rs773742345 Chr.12: 62603673-62603673 on Build GRCh38
rs897450253 Chr.12: 62603661-62603661 on Build GRCh38
rs766726101 Chr.12: 62603380-62603380 on Build GRCh38
rs760338122 Chr.12: 62603727-62603727 on Build GRCh38
rs565248118 Chr.12: 62603600-62603600 on Build GRCh38
rs10877887 Chr.12: 62603400-62603400 on Build GRCh38
rs556257065 Chr.12: 62603442-62603442 on Build GRCh38
rs914877477 Chr.12: 62603384-62603384 on Build GRCh38
rs946426558 Chr.12: 62603393-62603393 on Build GRCh38
rs954271945 Chr.12: 62603532-62603532 on Build GRCh38
rs945236906 Chr.12: 62603437-62603437 on Build GRCh38
rs1029462987 Chr.12: 62603477-62603477 on Build GRCh38
rs538612367 Chr.12: 62603474-62603474 on Build GRCh38
rs897313369 Chr.12: 62603495-62603495 on Build GRCh38
rs757204594 Chr.12: 62603366-62603366 on Build GRCh38
rs559675799 Chr.12: 62603649-62603649 on Build GRCh38
rs113188524 Chr.12: 62603373-62603373 on Build GRCh38
rs373730636 Chr.12: 62603663-62603663 on Build GRCh38
rs750365860 Chr.12: 62603645-62603645 on Build GRCh38
rs1012353015 Chr.12: 62603441-62603441 on Build GRCh38
rs781390203 Chr.12: 62603809-62603809 on Build GRCh38
rs576113439 Chr.12: 62603450-62603450 on Build GRCh38
rs1027067357 Chr.12: 62603518-62603518 on Build GRCh38
rs774888189 Chr.12: 62603721-62603721 on Build GRCh38
rs1015918983 Chr.12: 62603457-62603457 on Build GRCh38
rs1004925218 Chr.12: 62603452-62603452 on Build GRCh38
rs961290113 Chr.12: 62603376-62603376 on Build GRCh38
rs867804200 Chr.12: 62603511-62603511 on Build GRCh38
rs761374780 Chr.12: 62603737-62603737 on Build GRCh38
rs113451066 Chr.12: 62603418-62603418 on Build GRCh38
rs182504454 Chr.12: 62603557-62603557 on Build GRCh38
rs954063919 Chr.12: 62603405-62603405 on Build GRCh38
rs534077094 Chr.12: 62603385-62603385 on Build GRCh38
rs1055204401 Chr.12: 62603454-62603454 on Build GRCh38
rs752740994 Chr.12: 62603424-62603424 on Build GRCh38
rs965337124 Chr.12: 62603745-62603745 on Build GRCh38
rs948225366 Chr.12: 62603624-62603624 on Build GRCh38
rs968108123 Chr.12: 62603544-62603544 on Build GRCh38
rs913755984 Chr.12: 62603435-62603435 on Build GRCh38
rs541690178 Chr.12: 62603541-62603541 on Build GRCh38
rs903636850 Chr.12: 62603446-62603446 on Build GRCh38
rs1054773633 Chr.12: 62603609-62603609 on Build GRCh38
rs750326290 Chr.12: 62603766-62603766 on Build GRCh38
rs79508661 Chr.12: 62603652-62603652 on Build GRCh38

View Full Product Details