Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64697271-64697780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASSF3
Entrez Gene ID
283349
Gene Name
Ras association domain family member 3
Gene Aliases
RASSF5
UniGene
Hs.643605
Gene Chromosome Location
Chr.12: 64610513-64697567 on Build GRCh38
Transcripts
NM_178169.3 NR_040718.1 XM_006719345.3 XM_011538195.2 XM_017019182.1

SNP Detail

SNP ID Chromosome Location
rs905675441 Chr.12: 64697624-64697624 on Build GRCh38
rs781539996 Chr.12: 64697327-64697327 on Build GRCh38
rs141464309 Chr.12: 64697688-64697688 on Build GRCh38
rs15958 Chr.12: 64697534-64697534 on Build GRCh38
rs184157681 Chr.12: 64697623-64697623 on Build GRCh38
rs1017033069 Chr.12: 64697309-64697309 on Build GRCh38
rs1038113079 Chr.12: 64697640-64697640 on Build GRCh38
rs538906029 Chr.12: 64697386-64697386 on Build GRCh38
rs574626106 Chr.12: 64697586-64697591 on Build GRCh38
rs1048466392 Chr.12: 64697391-64697391 on Build GRCh38
rs964900120 Chr.12: 64697712-64697712 on Build GRCh38
rs751452424 Chr.12: 64697321-64697321 on Build GRCh38
rs901314666 Chr.12: 64697547-64697547 on Build GRCh38
rs993794654 Chr.12: 64697651-64697651 on Build GRCh38
rs913129071 Chr.12: 64697573-64697573 on Build GRCh38
rs996675687 Chr.12: 64697721-64697721 on Build GRCh38
rs988020556 Chr.12: 64697392-64697392 on Build GRCh38
rs974852773 Chr.12: 64697502-64697502 on Build GRCh38
rs1019099778 Chr.12: 64697446-64697446 on Build GRCh38
rs898158051 Chr.12: 64697316-64697316 on Build GRCh38
rs767378379 Chr.12: 64697591-64697591 on Build GRCh38
rs773311393 Chr.12: 64697472-64697477 on Build GRCh38
rs898171139 Chr.12: 64697642-64697642 on Build GRCh38
rs1048026108 Chr.12: 64697671-64697671 on Build GRCh38
rs1012379556 Chr.12: 64697643-64697643 on Build GRCh38
rs1019571536 Chr.12: 64697708-64697708 on Build GRCh38
rs929955731 Chr.12: 64697380-64697380 on Build GRCh38
rs112672533 Chr.12: 64697334-64697334 on Build GRCh38
rs56953843 Chr.12: 64697567-64697567 on Build GRCh38
rs965181326 Chr.12: 64697467-64697467 on Build GRCh38
rs942809054 Chr.12: 64697528-64697528 on Build GRCh38
rs61932663 Chr.12: 64697625-64697625 on Build GRCh38
rs925981306 Chr.12: 64697509-64697509 on Build GRCh38
rs886660708 Chr.12: 64697674-64697674 on Build GRCh38
rs1023388000 Chr.12: 64697665-64697665 on Build GRCh38
rs551984441 Chr.12: 64697614-64697614 on Build GRCh38
rs1041917678 Chr.12: 64697574-64697574 on Build GRCh38
rs367730972 Chr.12: 64697319-64697319 on Build GRCh38
rs547838753 Chr.12: 64697716-64697716 on Build GRCh38

View Full Product Details