Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 96282419-96282925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK17
Entrez Gene ID
5128
Gene Name
cyclin dependent kinase 17
Gene Aliases
PCTAIRE2, PCTK2
UniGene
Hs.506415
Gene Chromosome Location
Chr.12: 96278261-96400588 on Build GRCh38
Transcripts
NM_001170464.2 NM_002595.4 XM_017019405.1 XM_017019406.1 XM_017019407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4980746 c.1384G>A p.V462I Chr.12: 96282581-96282581 on Build GRCh38
5376242 c.1393G>A p.E465K Chr.12: 96282572-96282572 on Build GRCh38
4980747 c.1384G>A p.V462I Chr.12: 96282581-96282581 on Build GRCh38
3738608 c.1370A>G p.E457G Chr.12: 96282595-96282595 on Build GRCh38
4671116 c.1426C>T p.L476L Chr.12: 96282539-96282539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs60309851 Chr.12: 96282583-96282583 on Build GRCh38
rs563508256 Chr.12: 96282494-96282494 on Build GRCh38
rs973508201 Chr.12: 96282815-96282815 on Build GRCh38
rs755355257 Chr.12: 96282459-96282459 on Build GRCh38
rs1010781980 Chr.12: 96282598-96282598 on Build GRCh38
rs774161512 Chr.12: 96282825-96282825 on Build GRCh38
rs1047747301 Chr.12: 96282795-96282795 on Build GRCh38
rs930264311 Chr.12: 96282793-96282793 on Build GRCh38
rs921646763 Chr.12: 96282490-96282490 on Build GRCh38
rs753694636 Chr.12: 96282630-96282631 on Build GRCh38
rs923153218 Chr.12: 96282688-96282689 on Build GRCh38
rs780279482 Chr.12: 96282701-96282701 on Build GRCh38
rs954689472 Chr.12: 96282703-96282703 on Build GRCh38
rs963512267 Chr.12: 96282813-96282813 on Build GRCh38
rs370751661 Chr.12: 96282615-96282615 on Build GRCh38
rs910249519 Chr.12: 96282448-96282448 on Build GRCh38
rs566595512 Chr.12: 96282564-96282564 on Build GRCh38
rs967238202 Chr.12: 96282674-96282674 on Build GRCh38
rs1050581926 Chr.12: 96282445-96282445 on Build GRCh38
rs752865496 Chr.12: 96282480-96282480 on Build GRCh38
rs34916547 Chr.12: 96282749-96282750 on Build GRCh38
rs560841111 Chr.12: 96282588-96282588 on Build GRCh38
rs139401006 Chr.12: 96282544-96282544 on Build GRCh38
rs757377695 Chr.12: 96282521-96282521 on Build GRCh38
rs747780273 Chr.12: 96282555-96282555 on Build GRCh38
rs757558854 Chr.12: 96282748-96282749 on Build GRCh38
rs1037338326 Chr.12: 96282776-96282776 on Build GRCh38
rs746889872 Chr.12: 96282547-96282547 on Build GRCh38
rs571130238 Chr.12: 96282887-96282887 on Build GRCh38
rs922116140 Chr.12: 96282836-96282836 on Build GRCh38
rs1011491818 Chr.12: 96282635-96282635 on Build GRCh38
rs760020649 Chr.12: 96282589-96282589 on Build GRCh38
rs777668645 Chr.12: 96282555-96282556 on Build GRCh38
rs553714097 Chr.12: 96282869-96282869 on Build GRCh38
rs546557196 Chr.12: 96282781-96282781 on Build GRCh38
rs186159707 Chr.12: 96282737-96282737 on Build GRCh38
rs942241599 Chr.12: 96282475-96282475 on Build GRCh38
rs373935134 Chr.12: 96282647-96282647 on Build GRCh38
rs35751611 Chr.12: 96282552-96282552 on Build GRCh38
rs200728242 Chr.12: 96282460-96282460 on Build GRCh38
rs563703693 Chr.12: 96282807-96282807 on Build GRCh38
rs137990615 Chr.12: 96282573-96282573 on Build GRCh38
rs764448164 Chr.12: 96282616-96282616 on Build GRCh38
rs757430629 Chr.12: 96282651-96282651 on Build GRCh38
rs1036078580 Chr.12: 96282670-96282670 on Build GRCh38
rs374941693 Chr.12: 96282497-96282497 on Build GRCh38
rs753054186 Chr.12: 96282609-96282609 on Build GRCh38
rs373534430 Chr.12: 96282582-96282582 on Build GRCh38
rs574162082 Chr.12: 96282735-96282735 on Build GRCh38
rs1015212297 Chr.12: 96282788-96282788 on Build GRCh38
rs1037406814 Chr.12: 96282484-96282484 on Build GRCh38
rs377274185 Chr.12: 96282605-96282605 on Build GRCh38
rs920190712 Chr.12: 96282782-96282782 on Build GRCh38
rs200312658 Chr.12: 96282515-96282515 on Build GRCh38
rs771091546 Chr.12: 96282721-96282721 on Build GRCh38
rs977676842 Chr.12: 96282683-96282683 on Build GRCh38
rs984095926 Chr.12: 96282770-96282770 on Build GRCh38
rs781003801 Chr.12: 96282554-96282554 on Build GRCh38
rs749525931 Chr.12: 96282664-96282664 on Build GRCh38
rs931709031 Chr.12: 96282857-96282857 on Build GRCh38
rs769755890 Chr.12: 96282576-96282576 on Build GRCh38

View Full Product Details