Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 102843555-102844046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAH
Entrez Gene ID
5053
Gene Name
phenylalanine hydroxylase
Gene Aliases
PH, PKU, PKU1
UniGene
Hs.603740
Gene Chromosome Location
Chr.12: 102838326-102917603 on Build GRCh38
Transcripts
NM_000277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
691583 c.1183G>T p.A395S Chr.12: 102843662-102843662 on Build GRCh38
6462213 c.1078G>A p.E360K Chr.12: 102843767-102843767 on Build GRCh38
4522915 c.1188G>A p.K396K Chr.12: 102843657-102843657 on Build GRCh38
5909278 c.1189G>A p.E397K Chr.12: 102843656-102843656 on Build GRCh38
1706139 c.1118C>T p.A373V Chr.12: 102843727-102843727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199475593 Chr.12: 102843647-102843647 on Build GRCh38
rs62506951 Chr.12: 102843746-102843747 on Build GRCh38
rs5030654 Chr.12: 102843756-102843756 on Build GRCh38
rs62516102 Chr.12: 102843664-102843664 on Build GRCh38
rs754762316 Chr.12: 102844002-102844002 on Build GRCh38
rs62507334 Chr.12: 102843793-102843793 on Build GRCh38
rs751688151 Chr.12: 102843881-102843881 on Build GRCh38
rs1054069544 Chr.12: 102843907-102843907 on Build GRCh38
rs780691974 Chr.12: 102843712-102843712 on Build GRCh38
rs281865458 Chr.12: 102843693-102843693 on Build GRCh38
rs62508595 Chr.12: 102843776-102843776 on Build GRCh38
rs765124844 Chr.12: 102843998-102843998 on Build GRCh38
rs750730715 Chr.12: 102843788-102843788 on Build GRCh38
rs535141809 Chr.12: 102843857-102843857 on Build GRCh38
rs779053558 Chr.12: 102843708-102843708 on Build GRCh38
rs766107583 Chr.12: 102843774-102843774 on Build GRCh38
rs62508574 Chr.12: 102843745-102843745 on Build GRCh38
rs62517163 Chr.12: 102843731-102843731 on Build GRCh38
rs869312997 Chr.12: 102843673-102843673 on Build GRCh38
rs62507320 Chr.12: 102843779-102843779 on Build GRCh38
rs542946164 Chr.12: 102843964-102843964 on Build GRCh38
rs916247986 Chr.12: 102843820-102843820 on Build GRCh38
rs369574052 Chr.12: 102843596-102843596 on Build GRCh38
rs786200861 Chr.12: 102843751-102843753 on Build GRCh38
rs199475685 Chr.12: 102843810-102843810 on Build GRCh38
rs184148104 Chr.12: 102843722-102843722 on Build GRCh38
rs62508674 Chr.12: 102843641-102843641 on Build GRCh38
rs752746091 Chr.12: 102843611-102843611 on Build GRCh38
rs62508684 Chr.12: 102843780-102843780 on Build GRCh38
rs772630527 Chr.12: 102843710-102843710 on Build GRCh38
rs786200862 Chr.12: 102843739-102843753 on Build GRCh38
rs62517194 Chr.12: 102843686-102843686 on Build GRCh38
rs62508737 Chr.12: 102843644-102843644 on Build GRCh38
rs281865435 Chr.12: 102843682-102843682 on Build GRCh38
rs62642937 Chr.12: 102843706-102843706 on Build GRCh38
rs573675426 Chr.12: 102843844-102843844 on Build GRCh38
rs199475695 Chr.12: 102843670-102843670 on Build GRCh38
rs776159017 Chr.12: 102843747-102843747 on Build GRCh38
rs769076484 Chr.12: 102843725-102843725 on Build GRCh38
rs761487922 Chr.12: 102843668-102843668 on Build GRCh38
rs775701242 Chr.12: 102843628-102843628 on Build GRCh38
rs199475629 Chr.12: 102843681-102843682 on Build GRCh38
rs62642911 Chr.12: 102843713-102843713 on Build GRCh38
rs62516101 Chr.12: 102843683-102843683 on Build GRCh38
rs772683682 Chr.12: 102843657-102843657 on Build GRCh38
rs62516096 Chr.12: 102843753-102843755 on Build GRCh38
rs951540129 Chr.12: 102843752-102843752 on Build GRCh38
rs894886853 Chr.12: 102843575-102843575 on Build GRCh38
rs62508736 Chr.12: 102843661-102843661 on Build GRCh38
rs62507329 Chr.12: 102843761-102843761 on Build GRCh38
rs781593136 Chr.12: 102843831-102843831 on Build GRCh38
rs976776708 Chr.12: 102843617-102843617 on Build GRCh38
rs374160054 Chr.12: 102844018-102844018 on Build GRCh38
rs376480977 Chr.12: 102843771-102843771 on Build GRCh38
rs757320744 Chr.12: 102843783-102843783 on Build GRCh38
rs998242445 Chr.12: 102843989-102843989 on Build GRCh38
rs199475584 Chr.12: 102843648-102843648 on Build GRCh38
rs62516099 Chr.12: 102843727-102843728 on Build GRCh38
rs112537797 Chr.12: 102843615-102843615 on Build GRCh38
rs776178623 Chr.12: 102843658-102843658 on Build GRCh38
rs199475667 Chr.12: 102843757-102843758 on Build GRCh38
rs62642942 Chr.12: 102843715-102843715 on Build GRCh38
rs922671240 Chr.12: 102843930-102843930 on Build GRCh38
rs1056671080 Chr.12: 102844022-102844022 on Build GRCh38
rs549833778 Chr.12: 102843801-102843801 on Build GRCh38
rs5030855 Chr.12: 102843790-102843790 on Build GRCh38
rs1054762308 Chr.12: 102843795-102843795 on Build GRCh38
rs1032248318 Chr.12: 102843667-102843667 on Build GRCh38
rs10860928 Chr.12: 102843972-102843972 on Build GRCh38
rs281865453 Chr.12: 102843674-102843674 on Build GRCh38
rs62517203 Chr.12: 102843745-102843746 on Build GRCh38
rs1029016624 Chr.12: 102843950-102843950 on Build GRCh38
rs1037261330 Chr.12: 102843983-102843983 on Build GRCh38
rs62516142 Chr.12: 102843665-102843665 on Build GRCh38
rs918863691 Chr.12: 102843997-102843997 on Build GRCh38
rs780020477 Chr.12: 102843610-102843610 on Build GRCh38
rs199475658 Chr.12: 102843646-102843646 on Build GRCh38
rs755489088 Chr.12: 102843804-102843804 on Build GRCh38
rs62509018 Chr.12: 102843626-102843626 on Build GRCh38
rs372558114 Chr.12: 102843608-102843608 on Build GRCh38
rs910131541 Chr.12: 102843784-102843784 on Build GRCh38
rs977894054 Chr.12: 102843588-102843588 on Build GRCh38
rs281865447 Chr.12: 102843781-102843781 on Build GRCh38
rs772918939 Chr.12: 102843764-102843764 on Build GRCh38
rs62506949 Chr.12: 102843679-102843679 on Build GRCh38
rs62508613 Chr.12: 102843629-102843629 on Build GRCh38
rs281865436 Chr.12: 102843649-102843649 on Build GRCh38
rs772896 Chr.12: 102844015-102844015 on Build GRCh38
rs777846905 Chr.12: 102843620-102843620 on Build GRCh38
rs199475490 Chr.12: 102843778-102843779 on Build GRCh38
rs62516097 Chr.12: 102843741-102843755 on Build GRCh38
rs149595475 Chr.12: 102843684-102843684 on Build GRCh38
rs63329263 Chr.12: 102843756-102843756 on Build GRCh38
rs554043701 Chr.12: 102843946-102843946 on Build GRCh38
rs267603271 Chr.12: 102843704-102843704 on Build GRCh38
rs62516103 Chr.12: 102843662-102843662 on Build GRCh38
rs771420692 Chr.12: 102843616-102843616 on Build GRCh38
rs62516141 Chr.12: 102843688-102843688 on Build GRCh38
rs772897 Chr.12: 102843690-102843690 on Build GRCh38
rs62516095 Chr.12: 102843777-102843777 on Build GRCh38
rs62642941 Chr.12: 102843716-102843716 on Build GRCh38
rs62509015 Chr.12: 102843645-102843645 on Build GRCh38
rs199475593 Chr.12: 102843647-102843647 on Build GRCh38
rs199475603 Chr.12: 102843646-102843649 on Build GRCh38
rs999297649 Chr.12: 102843889-102843889 on Build GRCh38
rs750218310 Chr.12: 102843604-102843604 on Build GRCh38
rs5030856 Chr.12: 102843676-102843676 on Build GRCh38
rs62642921 Chr.12: 102843718-102843718 on Build GRCh38
rs779292646 Chr.12: 102843636-102843636 on Build GRCh38
rs62507268 Chr.12: 102843733-102843733 on Build GRCh38
rs199475590 Chr.12: 102843647-102843647 on Build GRCh38
rs747661840 Chr.12: 102843750-102843750 on Build GRCh38
rs749467239 Chr.12: 102843624-102843624 on Build GRCh38
rs199475691 Chr.12: 102843689-102843689 on Build GRCh38
rs62508620 Chr.12: 102843746-102843746 on Build GRCh38
rs281865457 Chr.12: 102843576-102843576 on Build GRCh38
rs62508717 Chr.12: 102843728-102843728 on Build GRCh38
rs1037293795 Chr.12: 102843698-102843698 on Build GRCh38
rs62507344 Chr.12: 102843782-102843782 on Build GRCh38
rs373763334 Chr.12: 102843705-102843705 on Build GRCh38
rs199475638 Chr.12: 102843651-102843651 on Build GRCh38
rs765490451 Chr.12: 102843787-102843787 on Build GRCh38
rs199475638 Chr.12: 102843651-102843651 on Build GRCh38
rs965828144 Chr.12: 102843880-102843880 on Build GRCh38
rs746203167 Chr.12: 102843709-102843709 on Build GRCh38
rs62508648 Chr.12: 102843744-102843744 on Build GRCh38
rs954194113 Chr.12: 102843941-102843941 on Build GRCh38
rs760256025 Chr.12: 102843660-102843660 on Build GRCh38
rs771059413 Chr.12: 102843634-102843634 on Build GRCh38
rs180819807 Chr.12: 102843671-102843671 on Build GRCh38
rs62516098 Chr.12: 102843748-102843748 on Build GRCh38
rs5030854 Chr.12: 102843769-102843769 on Build GRCh38
rs1057516699 Chr.12: 102843698-102843698 on Build GRCh38

View Full Product Details