Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 106366316-106366824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR3B
Entrez Gene ID
55703
Gene Name
RNA polymerase III subunit B
Gene Aliases
C128, HLD8, INMAP, RPC2
UniGene
Hs.610795
Gene Chromosome Location
Chr.12: 106357658-106510198 on Build GRCh38
Transcripts
NM_001160708.1 NM_018082.5 XM_017019621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
317890 c.218G>T p.W73L Chr.12: 106366713-106366713 on Build GRCh38
1492756 c.227+2T>A p.? Chr.12: 106366724-106366724 on Build GRCh38
7304761 c.201T>C p.S67S Chr.12: 106366696-106366696 on Build GRCh38
6071108 c.112G>T p.G38C Chr.12: 106366522-106366522 on Build GRCh38
7313187 c.130A>C p.I44L Chr.12: 106366540-106366540 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs548719279 Chr.12: 106366518-106366518 on Build GRCh38
rs760544455 Chr.12: 106366467-106366467 on Build GRCh38
rs775696277 Chr.12: 106366693-106366693 on Build GRCh38
rs760770690 Chr.12: 106366702-106366702 on Build GRCh38
rs757656808 Chr.12: 106366580-106366580 on Build GRCh38
rs139226209 Chr.12: 106366339-106366339 on Build GRCh38
rs368827027 Chr.12: 106366705-106366705 on Build GRCh38
rs951242615 Chr.12: 106366391-106366391 on Build GRCh38
rs371743971 Chr.12: 106366609-106366609 on Build GRCh38
rs746599503 Chr.12: 106366628-106366628 on Build GRCh38
rs767784504 Chr.12: 106366688-106366688 on Build GRCh38
rs1001286550 Chr.12: 106366586-106366586 on Build GRCh38
rs111316875 Chr.12: 106366642-106366642 on Build GRCh38
rs983878168 Chr.12: 106366664-106366664 on Build GRCh38
rs371239622 Chr.12: 106366477-106366477 on Build GRCh38
rs879055058 Chr.12: 106366439-106366439 on Build GRCh38
rs1025501079 Chr.12: 106366385-106366385 on Build GRCh38
rs995323491 Chr.12: 106366355-106366355 on Build GRCh38
rs745721628 Chr.12: 106366660-106366662 on Build GRCh38
rs12318640 Chr.12: 106366347-106366347 on Build GRCh38
rs898556406 Chr.12: 106366706-106366706 on Build GRCh38
rs548094849 Chr.12: 106366761-106366761 on Build GRCh38
rs374259169 Chr.12: 106366498-106366498 on Build GRCh38
rs778537362 Chr.12: 106366598-106366598 on Build GRCh38
rs774453914 Chr.12: 106366675-106366675 on Build GRCh38
rs568682674 Chr.12: 106366667-106366667 on Build GRCh38
rs990235768 Chr.12: 106366556-106366556 on Build GRCh38
rs527953383 Chr.12: 106366680-106366680 on Build GRCh38
rs761562811 Chr.12: 106366750-106366750 on Build GRCh38
rs966554577 Chr.12: 106366739-106366739 on Build GRCh38
rs190249858 Chr.12: 106366361-106366361 on Build GRCh38
rs764541119 Chr.12: 106366703-106366703 on Build GRCh38
rs768536925 Chr.12: 106366474-106366474 on Build GRCh38
rs1025767651 Chr.12: 106366783-106366783 on Build GRCh38
rs776245705 Chr.12: 106366653-106366653 on Build GRCh38
rs942704269 Chr.12: 106366684-106366684 on Build GRCh38
rs757150494 Chr.12: 106366503-106366506 on Build GRCh38
rs777182958 Chr.12: 106366477-106366477 on Build GRCh38
rs980873946 Chr.12: 106366511-106366511 on Build GRCh38
rs368216350 Chr.12: 106366513-106366513 on Build GRCh38
rs771959481 Chr.12: 106366669-106366672 on Build GRCh38
rs946254326 Chr.12: 106366632-106366632 on Build GRCh38
rs763829196 Chr.12: 106366516-106366516 on Build GRCh38
rs567829877 Chr.12: 106366484-106366484 on Build GRCh38
rs373936359 Chr.12: 106366608-106366608 on Build GRCh38
rs79677422 Chr.12: 106366655-106366655 on Build GRCh38
rs748769049 Chr.12: 106366499-106366502 on Build GRCh38
rs922079676 Chr.12: 106366349-106366349 on Build GRCh38
rs756485164 Chr.12: 106366536-106366536 on Build GRCh38
rs748135287 Chr.12: 106366660-106366660 on Build GRCh38
rs372689685 Chr.12: 106366476-106366476 on Build GRCh38
rs530257365 Chr.12: 106366351-106366351 on Build GRCh38
rs759803178 Chr.12: 106366506-106366506 on Build GRCh38
rs757826726 Chr.12: 106366708-106366708 on Build GRCh38
rs1027995437 Chr.12: 106366593-106366593 on Build GRCh38
rs969862983 Chr.12: 106366566-106366566 on Build GRCh38
rs1040124471 Chr.12: 106366697-106366697 on Build GRCh38
rs754400950 Chr.12: 106366542-106366542 on Build GRCh38
rs1013947151 Chr.12: 106366529-106366529 on Build GRCh38
rs779843065 Chr.12: 106366627-106366627 on Build GRCh38
rs753126070 Chr.12: 106366528-106366528 on Build GRCh38
rs992068401 Chr.12: 106366440-106366440 on Build GRCh38
rs963148997 Chr.12: 106366353-106366353 on Build GRCh38
rs774242810 Chr.12: 106366500-106366500 on Build GRCh38
rs746687747 Chr.12: 106366764-106366764 on Build GRCh38
rs764300320 Chr.12: 106366540-106366540 on Build GRCh38
rs750015005 Chr.12: 106366723-106366723 on Build GRCh38
rs765662843 Chr.12: 106366709-106366709 on Build GRCh38
rs1052757811 Chr.12: 106366496-106366496 on Build GRCh38
rs936434505 Chr.12: 106366541-106366541 on Build GRCh38
rs778977143 Chr.12: 106366600-106366601 on Build GRCh38
rs891396995 Chr.12: 106366647-106366647 on Build GRCh38
rs200767121 Chr.12: 106366479-106366479 on Build GRCh38
rs563037224 Chr.12: 106366432-106366432 on Build GRCh38

View Full Product Details