Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 106681902-106682399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100287944
Entrez Gene ID
100287944
Gene Name
uncharacterized LOC100287944
Gene Aliases
-
UniGene
Hs.742087
Gene Chromosome Location
Chr.12: 106496410-106774831 on Build GRCh38
Transcripts
NR_040246.1 NR_038912.1 NM_001206691.1 NM_213594.2

Gene Symbol
LOC100505978
Entrez Gene ID
100505978
Gene Name
uncharacterized LOC100505978
Gene Aliases
-
UniGene
Hs.662583
Gene Chromosome Location
Chr.12: 106680758-106684700 on Build GRCh38
Transcripts
NR_040246.1 NR_038912.1 NM_001206691.1 NM_213594.2

Gene Symbol
RFX4
Entrez Gene ID
5992
Gene Name
regulatory factor X4
Gene Aliases
NYD-SP10
UniGene
Hs.388827
Gene Chromosome Location
Chr.12: 106582907-106762804 on Build GRCh38
Transcripts
NR_040246.1 NR_038912.1 NM_001206691.1 NM_213594.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1318064 c.385G>A p.G129R Chr.12: 106682035-106682035 on Build GRCh38
314760 c.363C>A p.T121T Chr.12: 106682040-106682040 on Build GRCh38
6071095 c.386G>T p.G129V Chr.12: 106682036-106682036 on Build GRCh38
3670939 c.385G>T p.G129W Chr.12: 106682035-106682035 on Build GRCh38
3416425 c.356A>G p.Q119R Chr.12: 106682006-106682006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748273668 Chr.12: 106682086-106682086 on Build GRCh38
rs774783070 Chr.12: 106682100-106682100 on Build GRCh38
rs768911487 Chr.12: 106681957-106681957 on Build GRCh38
rs775524200 Chr.12: 106682085-106682085 on Build GRCh38
rs931480904 Chr.12: 106682136-106682136 on Build GRCh38
rs1047792967 Chr.12: 106682160-106682160 on Build GRCh38
rs778753683 Chr.12: 106682072-106682072 on Build GRCh38
rs969074837 Chr.12: 106682234-106682234 on Build GRCh38
rs887372555 Chr.12: 106682176-106682176 on Build GRCh38
rs201502358 Chr.12: 106682078-106682078 on Build GRCh38
rs752216540 Chr.12: 106682034-106682034 on Build GRCh38
rs912467372 Chr.12: 106682339-106682339 on Build GRCh38
rs115876619 Chr.12: 106682134-106682134 on Build GRCh38
rs138508135 Chr.12: 106682372-106682372 on Build GRCh38
rs905168533 Chr.12: 106682236-106682236 on Build GRCh38
rs530712727 Chr.12: 106682199-106682199 on Build GRCh38
rs759491867 Chr.12: 106681953-106681953 on Build GRCh38
rs910182481 Chr.12: 106682089-106682089 on Build GRCh38
rs759836303 Chr.12: 106682104-106682104 on Build GRCh38
rs919851971 Chr.12: 106681933-106681933 on Build GRCh38
rs181400590 Chr.12: 106681994-106681994 on Build GRCh38
rs371836622 Chr.12: 106681966-106681966 on Build GRCh38
rs867953455 Chr.12: 106682011-106682011 on Build GRCh38
rs1032596098 Chr.12: 106682004-106682004 on Build GRCh38
rs539732370 Chr.12: 106682135-106682135 on Build GRCh38
rs949614857 Chr.12: 106682198-106682198 on Build GRCh38
rs773446302 Chr.12: 106681979-106681979 on Build GRCh38
rs893652448 Chr.12: 106682327-106682327 on Build GRCh38
rs374526018 Chr.12: 106682064-106682064 on Build GRCh38
rs368598293 Chr.12: 106682068-106682068 on Build GRCh38
rs776842196 Chr.12: 106681958-106681958 on Build GRCh38
rs759865442 Chr.12: 106682254-106682254 on Build GRCh38
rs749058234 Chr.12: 106681944-106681944 on Build GRCh38
rs763269186 Chr.12: 106681985-106681985 on Build GRCh38
rs763225606 Chr.12: 106682091-106682091 on Build GRCh38
rs748936687 Chr.12: 106682067-106682067 on Build GRCh38
rs550963925 Chr.12: 106682125-106682125 on Build GRCh38
rs772848510 Chr.12: 106682241-106682241 on Build GRCh38
rs761930747 Chr.12: 106681962-106681962 on Build GRCh38
rs919569122 Chr.12: 106682289-106682289 on Build GRCh38
rs767393065 Chr.12: 106682108-106682108 on Build GRCh38
rs1053871139 Chr.12: 106682312-106682312 on Build GRCh38
rs973722691 Chr.12: 106682247-106682247 on Build GRCh38
rs943890066 Chr.12: 106682354-106682354 on Build GRCh38
rs367624650 Chr.12: 106681960-106681960 on Build GRCh38
rs556016715 Chr.12: 106682357-106682357 on Build GRCh38
rs767944893 Chr.12: 106682028-106682028 on Build GRCh38
rs200119049 Chr.12: 106682007-106682007 on Build GRCh38
rs200667832 Chr.12: 106682096-106682096 on Build GRCh38
rs951038856 Chr.12: 106682316-106682316 on Build GRCh38
rs755473453 Chr.12: 106682025-106682025 on Build GRCh38
rs377144638 Chr.12: 106682088-106682088 on Build GRCh38
rs755570948 Chr.12: 106682063-106682063 on Build GRCh38
rs774193631 Chr.12: 106681952-106681952 on Build GRCh38
rs769967438 Chr.12: 106682087-106682087 on Build GRCh38
rs768034498 Chr.12: 106682105-106682105 on Build GRCh38

View Full Product Details