Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 107734222-107734653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM4
Entrez Gene ID
11108
Gene Name
PR domain 4
Gene Aliases
PFM1
UniGene
Hs.506655
Gene Chromosome Location
Chr.12: 107732866-107761137 on Build GRCh38
Transcripts
NM_012406.3 XM_005268593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5778696 c.2279C>T p.S760L Chr.12: 107734337-107734337 on Build GRCh38
4168536 c.2296G>C p.E766Q Chr.12: 107734320-107734320 on Build GRCh38
6755165 c.2099G>A p.R700H Chr.12: 107734517-107734517 on Build GRCh38
246765 c.2235C>T p.L745L Chr.12: 107734381-107734381 on Build GRCh38
3368637 c.2387C>A p.S796Y Chr.12: 107734229-107734229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776117303 Chr.12: 107734281-107734281 on Build GRCh38
rs766003588 Chr.12: 107734536-107734536 on Build GRCh38
rs907032492 Chr.12: 107734325-107734325 on Build GRCh38
rs371787197 Chr.12: 107734517-107734517 on Build GRCh38
rs755842750 Chr.12: 107734467-107734467 on Build GRCh38
rs778972230 Chr.12: 107734353-107734353 on Build GRCh38
rs912408873 Chr.12: 107734483-107734483 on Build GRCh38
rs374059727 Chr.12: 107734450-107734450 on Build GRCh38
rs908853873 Chr.12: 107734583-107734583 on Build GRCh38
rs762457553 Chr.12: 107734552-107734552 on Build GRCh38
rs999630885 Chr.12: 107734607-107734607 on Build GRCh38
rs145325057 Chr.12: 107734436-107734436 on Build GRCh38
rs200101622 Chr.12: 107734434-107734434 on Build GRCh38
rs764511908 Chr.12: 107734287-107734287 on Build GRCh38
rs762075424 Chr.12: 107734393-107734393 on Build GRCh38
rs371800658 Chr.12: 107734346-107734346 on Build GRCh38
rs757811918 Chr.12: 107734322-107734322 on Build GRCh38
rs769699258 Chr.12: 107734350-107734350 on Build GRCh38
rs746150723 Chr.12: 107734355-107734355 on Build GRCh38
rs1052570018 Chr.12: 107734616-107734616 on Build GRCh38
rs73189508 Chr.12: 107734594-107734594 on Build GRCh38
rs771073556 Chr.12: 107734246-107734246 on Build GRCh38
rs369142733 Chr.12: 107734372-107734372 on Build GRCh38
rs746037087 Chr.12: 107734499-107734499 on Build GRCh38
rs950491554 Chr.12: 107734527-107734527 on Build GRCh38
rs777242739 Chr.12: 107734533-107734533 on Build GRCh38
rs761194964 Chr.12: 107734377-107734377 on Build GRCh38
rs772341325 Chr.12: 107734513-107734513 on Build GRCh38
rs982554814 Chr.12: 107734468-107734468 on Build GRCh38
rs1022417918 Chr.12: 107734307-107734307 on Build GRCh38
rs148832135 Chr.12: 107734543-107734543 on Build GRCh38
rs972840363 Chr.12: 107734487-107734487 on Build GRCh38
rs766005446 Chr.12: 107734569-107734569 on Build GRCh38
rs762701897 Chr.12: 107734427-107734427 on Build GRCh38
rs575003510 Chr.12: 107734604-107734604 on Build GRCh38
rs148191921 Chr.12: 107734340-107734340 on Build GRCh38
rs908893359 Chr.12: 107734285-107734285 on Build GRCh38
rs749998394 Chr.12: 107734410-107734410 on Build GRCh38
rs756873434 Chr.12: 107734294-107734294 on Build GRCh38
rs896656989 Chr.12: 107734631-107734631 on Build GRCh38
rs747926890 Chr.12: 107734344-107734344 on Build GRCh38
rs756739443 Chr.12: 107734582-107734585 on Build GRCh38
rs759530984 Chr.12: 107734277-107734277 on Build GRCh38
rs761286616 Chr.12: 107734286-107734286 on Build GRCh38
rs751281315 Chr.12: 107734570-107734570 on Build GRCh38
rs889822751 Chr.12: 107734381-107734381 on Build GRCh38
rs764476667 Chr.12: 107734282-107734283 on Build GRCh38
rs762609067 Chr.12: 107734343-107734344 on Build GRCh38
rs759018249 Chr.12: 107734573-107734573 on Build GRCh38
rs765670500 Chr.12: 107734399-107734399 on Build GRCh38
rs200596155 Chr.12: 107734358-107734358 on Build GRCh38
rs759640445 Chr.12: 107734348-107734348 on Build GRCh38
rs777645035 Chr.12: 107734486-107734486 on Build GRCh38
rs747069656 Chr.12: 107734518-107734518 on Build GRCh38
rs929064026 Chr.12: 107734255-107734255 on Build GRCh38
rs143824903 Chr.12: 107734291-107734291 on Build GRCh38
rs1052948567 Chr.12: 107734352-107734352 on Build GRCh38
rs553464967 Chr.12: 107734598-107734598 on Build GRCh38
rs570856934 Chr.12: 107734592-107734592 on Build GRCh38
rs768921616 Chr.12: 107734521-107734521 on Build GRCh38
rs1044188816 Chr.12: 107734590-107734590 on Build GRCh38
rs754135018 Chr.12: 107734316-107734318 on Build GRCh38
rs951199302 Chr.12: 107734457-107734457 on Build GRCh38
rs779566334 Chr.12: 107734332-107734332 on Build GRCh38
rs769297620 Chr.12: 107734383-107734383 on Build GRCh38
rs777332636 Chr.12: 107734386-107734386 on Build GRCh38
rs751098686 Chr.12: 107734567-107734569 on Build GRCh38
rs765810913 Chr.12: 107734431-107734431 on Build GRCh38
rs767675481 Chr.12: 107734280-107734280 on Build GRCh38
rs764780238 Chr.12: 107734295-107734295 on Build GRCh38
rs201810021 Chr.12: 107734339-107734339 on Build GRCh38
rs980419252 Chr.12: 107734599-107734599 on Build GRCh38
rs774693123 Chr.12: 107734269-107734269 on Build GRCh38
rs750152850 Chr.12: 107734311-107734311 on Build GRCh38
rs770018153 Chr.12: 107734537-107734537 on Build GRCh38
rs761041073 Chr.12: 107734279-107734280 on Build GRCh38
rs751087036 Chr.12: 107734433-107734433 on Build GRCh38

View Full Product Details