Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109793447-109793950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPV4
Entrez Gene ID
59341
Gene Name
transient receptor potential cation channel subfamily V member 4
Gene Aliases
BCYM3, CMT2C, HMSN2C, OTRPC4, SMAL, SPSMA, SSQTL1, TRP12, VRL2, VROAC
UniGene
Hs.506713
Gene Chromosome Location
Chr.12: 109783087-109833407 on Build GRCh38
Transcripts
NM_001177428.1 NM_001177431.1 NM_001177433.1 NM_021625.4 NM_147204.2 XM_005253918.1 XM_011538630.2 XM_011538631.2 XM_011538632.2 XM_011538633.2 XM_011538634.2 XM_011538635.2 XM_011538636.2 XM_017019774.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
934865 c.1593C>A p.D531E Chr.12: 109793592-109793592 on Build GRCh38
1744633 c.1572C>G p.F524L Chr.12: 109793942-109793942 on Build GRCh38
7395367 c.1612C>T p.P538S Chr.12: 109793573-109793573 on Build GRCh38
4949621 c.1619T>C p.V540A Chr.12: 109793566-109793566 on Build GRCh38
6810005 c.1584C>T p.N528N Chr.12: 109793930-109793930 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs199712027 Chr.12: 109793917-109793917 on Build GRCh38
rs767801563 Chr.12: 109793622-109793622 on Build GRCh38
rs371623092 Chr.12: 109793505-109793505 on Build GRCh38
rs928767006 Chr.12: 109793774-109793774 on Build GRCh38
rs968089386 Chr.12: 109793808-109793808 on Build GRCh38
rs1056876646 Chr.12: 109793767-109793767 on Build GRCh38
rs527681432 Chr.12: 109793790-109793790 on Build GRCh38
rs976191720 Chr.12: 109793599-109793599 on Build GRCh38
rs779150598 Chr.12: 109793891-109793891 on Build GRCh38
rs892429099 Chr.12: 109793777-109793777 on Build GRCh38
rs749772879 Chr.12: 109793538-109793538 on Build GRCh38
rs564663557 Chr.12: 109793659-109793659 on Build GRCh38
rs746189782 Chr.12: 109793905-109793905 on Build GRCh38
rs745522429 Chr.12: 109793559-109793559 on Build GRCh38
rs567120731 Chr.12: 109793680-109793680 on Build GRCh38
rs1048763492 Chr.12: 109793648-109793648 on Build GRCh38
rs751791805 Chr.12: 109793508-109793508 on Build GRCh38
rs779325064 Chr.12: 109793558-109793558 on Build GRCh38
rs772999764 Chr.12: 109793619-109793619 on Build GRCh38
rs990362875 Chr.12: 109793494-109793494 on Build GRCh38
rs1003791275 Chr.12: 109793746-109793746 on Build GRCh38
rs957922511 Chr.12: 109793894-109793918 on Build GRCh38
rs746473735 Chr.12: 109793588-109793588 on Build GRCh38
rs765519891 Chr.12: 109793821-109793821 on Build GRCh38
rs756714651 Chr.12: 109793522-109793522 on Build GRCh38
rs10850750 Chr.12: 109793912-109793912 on Build GRCh38
rs1037985809 Chr.12: 109793816-109793816 on Build GRCh38
rs752787333 Chr.12: 109793516-109793516 on Build GRCh38
rs913515146 Chr.12: 109793746-109793747 on Build GRCh38
rs756305668 Chr.12: 109793641-109793641 on Build GRCh38
rs748046273 Chr.12: 109793914-109793914 on Build GRCh38
rs545275380 Chr.12: 109793688-109793688 on Build GRCh38
rs768130865 Chr.12: 109793595-109793595 on Build GRCh38
rs1018279960 Chr.12: 109793705-109793705 on Build GRCh38
rs774925241 Chr.12: 109793482-109793482 on Build GRCh38
rs375478213 Chr.12: 109793906-109793906 on Build GRCh38
rs541606391 Chr.12: 109793529-109793529 on Build GRCh38
rs779730296 Chr.12: 109793907-109793907 on Build GRCh38
rs373233900 Chr.12: 109793915-109793915 on Build GRCh38
rs754262716 Chr.12: 109793904-109793904 on Build GRCh38
rs1018810356 Chr.12: 109793909-109793909 on Build GRCh38
rs1055242239 Chr.12: 109793797-109793797 on Build GRCh38
rs1010087108 Chr.12: 109793779-109793779 on Build GRCh38
rs1006724351 Chr.12: 109793908-109793908 on Build GRCh38
rs755277816 Chr.12: 109793813-109793813 on Build GRCh38
rs764230311 Chr.12: 109793646-109793646 on Build GRCh38
rs145701598 Chr.12: 109793718-109793718 on Build GRCh38
rs767628053 Chr.12: 109793893-109793893 on Build GRCh38
rs537691178 Chr.12: 109793826-109793826 on Build GRCh38
rs941481192 Chr.12: 109793666-109793666 on Build GRCh38
rs1038287578 Chr.12: 109793642-109793646 on Build GRCh38
rs771645140 Chr.12: 109793568-109793568 on Build GRCh38
rs770639004 Chr.12: 109793546-109793546 on Build GRCh38
rs774654042 Chr.12: 109793612-109793613 on Build GRCh38
rs757667255 Chr.12: 109793912-109793912 on Build GRCh38
rs771628950 Chr.12: 109793479-109793479 on Build GRCh38
rs373926569 Chr.12: 109793639-109793639 on Build GRCh38
rs778197123 Chr.12: 109793532-109793532 on Build GRCh38
rs775011025 Chr.12: 109793584-109793584 on Build GRCh38
rs56198703 Chr.12: 109793903-109793904 on Build GRCh38
rs387906902 Chr.12: 109793560-109793560 on Build GRCh38
rs746629480 Chr.12: 109793910-109793910 on Build GRCh38
rs375908611 Chr.12: 109793918-109793918 on Build GRCh38
rs376679493 Chr.12: 109793506-109793506 on Build GRCh38
rs760689921 Chr.12: 109793902-109793903 on Build GRCh38
rs761079651 Chr.12: 109793585-109793585 on Build GRCh38
rs574182242 Chr.12: 109793509-109793509 on Build GRCh38
rs549460191 Chr.12: 109793829-109793829 on Build GRCh38
rs761801079 Chr.12: 109793611-109793611 on Build GRCh38
rs897099210 Chr.12: 109793874-109793874 on Build GRCh38
rs988209996 Chr.12: 109793495-109793495 on Build GRCh38
rs760610613 Chr.12: 109793488-109793488 on Build GRCh38
rs759653708 Chr.12: 109793633-109793635 on Build GRCh38
rs754360769 Chr.12: 109793649-109793649 on Build GRCh38
rs367950220 Chr.12: 109793628-109793628 on Build GRCh38
rs957642420 Chr.12: 109793712-109793712 on Build GRCh38
rs1029889570 Chr.12: 109793669-109793669 on Build GRCh38
rs201534072 Chr.12: 109793621-109793621 on Build GRCh38
rs916328443 Chr.12: 109793802-109793802 on Build GRCh38
rs981624958 Chr.12: 109793768-109793768 on Build GRCh38
rs776469697 Chr.12: 109793602-109793602 on Build GRCh38
rs200083069 Chr.12: 109793512-109793512 on Build GRCh38
rs201974634 Chr.12: 109793618-109793618 on Build GRCh38
rs752762344 Chr.12: 109793886-109793888 on Build GRCh38
rs1011607708 Chr.12: 109793912-109793913 on Build GRCh38
rs757630049 Chr.12: 109793551-109793551 on Build GRCh38
rs763977240 Chr.12: 109793501-109793501 on Build GRCh38
rs767651884 Chr.12: 109793649-109793658 on Build GRCh38

View Full Product Details