Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21140422-21140922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAP18
Entrez Gene ID
10284
Gene Name
Sin3A associated protein 18
Gene Aliases
2HOR0202, SAP18P
UniGene
Hs.524899
Gene Chromosome Location
Chr.13: 21140514-21149085 on Build GRCh38
Transcripts
NM_005870.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5980459 c.14C>T p.S5L Chr.13: 21140623-21140623 on Build GRCh38
3981956 c.18C>T p.R6R Chr.13: 21140627-21140627 on Build GRCh38
7254615 c.64C>A p.R22S Chr.13: 21140673-21140673 on Build GRCh38
7283823 c.37A>T p.K13* Chr.13: 21140646-21140646 on Build GRCh38
4957815 c.37A>G p.K13E Chr.13: 21140646-21140646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773074400 Chr.13: 21140880-21140880 on Build GRCh38
rs760246435 Chr.13: 21140538-21140538 on Build GRCh38
rs745900432 Chr.13: 21140860-21140860 on Build GRCh38
rs767398747 Chr.13: 21140830-21140830 on Build GRCh38
rs139949359 Chr.13: 21140610-21140610 on Build GRCh38
rs778665161 Chr.13: 21140558-21140558 on Build GRCh38
rs768613316 Chr.13: 21140527-21140527 on Build GRCh38
rs975446690 Chr.13: 21140445-21140445 on Build GRCh38
rs775068346 Chr.13: 21140564-21140564 on Build GRCh38
rs751686217 Chr.13: 21140816-21140817 on Build GRCh38
rs1042805999 Chr.13: 21140484-21140484 on Build GRCh38
rs373936573 Chr.13: 21140591-21140591 on Build GRCh38
rs774572930 Chr.13: 21140698-21140698 on Build GRCh38
rs146445196 Chr.13: 21140785-21140785 on Build GRCh38
rs552329707 Chr.13: 21140583-21140583 on Build GRCh38
rs748872014 Chr.13: 21140691-21140691 on Build GRCh38
rs373660324 Chr.13: 21140456-21140456 on Build GRCh38
rs774972864 Chr.13: 21140592-21140592 on Build GRCh38
rs973947321 Chr.13: 21140717-21140717 on Build GRCh38
rs755736035 Chr.13: 21140763-21140763 on Build GRCh38
rs908359977 Chr.13: 21140461-21140461 on Build GRCh38
rs753229369 Chr.13: 21140837-21140837 on Build GRCh38
rs112687601 Chr.13: 21140576-21140576 on Build GRCh38
rs750433806 Chr.13: 21140604-21140604 on Build GRCh38
rs934333275 Chr.13: 21140492-21140492 on Build GRCh38
rs758460442 Chr.13: 21140507-21140507 on Build GRCh38
rs962754382 Chr.13: 21140713-21140713 on Build GRCh38
rs752464689 Chr.13: 21140620-21140620 on Build GRCh38
rs772594898 Chr.13: 21140800-21140800 on Build GRCh38
rs1003849675 Chr.13: 21140654-21140654 on Build GRCh38
rs748462649 Chr.13: 21140799-21140799 on Build GRCh38
rs772607672 Chr.13: 21140568-21140568 on Build GRCh38
rs557341838 Chr.13: 21140815-21140815 on Build GRCh38
rs550761091 Chr.13: 21140663-21140663 on Build GRCh38
rs765735715 Chr.13: 21140682-21140683 on Build GRCh38
rs779349398 Chr.13: 21140789-21140789 on Build GRCh38
rs765332394 Chr.13: 21140821-21140821 on Build GRCh38
rs761716479 Chr.13: 21140532-21140532 on Build GRCh38
rs762311999 Chr.13: 21140700-21140700 on Build GRCh38
rs374776695 Chr.13: 21140569-21140569 on Build GRCh38
rs751434418 Chr.13: 21140823-21140823 on Build GRCh38
rs536159279 Chr.13: 21140804-21140804 on Build GRCh38
rs775250865 Chr.13: 21140690-21140690 on Build GRCh38
rs774312518 Chr.13: 21140530-21140530 on Build GRCh38
rs769121125 Chr.13: 21140692-21140692 on Build GRCh38
rs143313767 Chr.13: 21140839-21140839 on Build GRCh38
rs754273316 Chr.13: 21140540-21140540 on Build GRCh38
rs1017859131 Chr.13: 21140534-21140534 on Build GRCh38
rs978956587 Chr.13: 21140554-21140554 on Build GRCh38
rs769582269 Chr.13: 21140518-21140518 on Build GRCh38
rs964979702 Chr.13: 21140627-21140627 on Build GRCh38
rs761615849 Chr.13: 21140502-21140502 on Build GRCh38
rs372783758 Chr.13: 21140709-21140709 on Build GRCh38
rs183417048 Chr.13: 21140769-21140769 on Build GRCh38
rs1029317634 Chr.13: 21140548-21140548 on Build GRCh38
rs949723620 Chr.13: 21140760-21140760 on Build GRCh38
rs746060471 Chr.13: 21140553-21140553 on Build GRCh38
rs775933057 Chr.13: 21140572-21140572 on Build GRCh38
rs201454186 Chr.13: 21140531-21140531 on Build GRCh38
rs904288486 Chr.13: 21140484-21140485 on Build GRCh38
rs745523301 Chr.13: 21140584-21140584 on Build GRCh38
rs757445435 Chr.13: 21140545-21140545 on Build GRCh38
rs575712951 Chr.13: 21140452-21140452 on Build GRCh38
rs773744887 Chr.13: 21140801-21140801 on Build GRCh38
rs769516459 Chr.13: 21140586-21140586 on Build GRCh38
rs760702593 Chr.13: 21140711-21140711 on Build GRCh38
rs1015648061 Chr.13: 21140881-21140881 on Build GRCh38
rs376020479 Chr.13: 21140666-21140666 on Build GRCh38
rs970024079 Chr.13: 21140727-21140727 on Build GRCh38
rs747763373 Chr.13: 21140840-21140840 on Build GRCh38
rs1029371490 Chr.13: 21140602-21140602 on Build GRCh38
rs765768187 Chr.13: 21140571-21140571 on Build GRCh38
rs998156028 Chr.13: 21140598-21140598 on Build GRCh38
rs540281720 Chr.13: 21140858-21140858 on Build GRCh38
rs370333573 Chr.13: 21140567-21140567 on Build GRCh38
rs750841197 Chr.13: 21140684-21140685 on Build GRCh38
rs772931538 Chr.13: 21140541-21140541 on Build GRCh38
rs761740714 Chr.13: 21140828-21140828 on Build GRCh38
rs575819023 Chr.13: 21140834-21140834 on Build GRCh38
rs759431578 Chr.13: 21140529-21140531 on Build GRCh38
rs375651525 Chr.13: 21140582-21140582 on Build GRCh38
rs76010291 Chr.13: 21140838-21140838 on Build GRCh38
rs772015507 Chr.13: 21140531-21140535 on Build GRCh38
rs774921169 Chr.13: 21140537-21140538 on Build GRCh38
rs777722330 Chr.13: 21140543-21140543 on Build GRCh38
rs749806992 Chr.13: 21140517-21140517 on Build GRCh38
rs777404407 Chr.13: 21140625-21140625 on Build GRCh38
rs199958048 Chr.13: 21140542-21140542 on Build GRCh38
rs981316221 Chr.13: 21140731-21140731 on Build GRCh38
rs896940106 Chr.13: 21140869-21140869 on Build GRCh38
rs747038252 Chr.13: 21140639-21140639 on Build GRCh38
rs113930453 Chr.13: 21140883-21140883 on Build GRCh38
rs548145674 Chr.13: 21140575-21140575 on Build GRCh38
rs200936744 Chr.13: 21140539-21140539 on Build GRCh38
rs759676201 Chr.13: 21140719-21140719 on Build GRCh38
rs753584617 Chr.13: 21140680-21140681 on Build GRCh38
rs752852568 Chr.13: 21140506-21140506 on Build GRCh38
rs781351427 Chr.13: 21140549-21140549 on Build GRCh38
rs763255860 Chr.13: 21140869-21140869 on Build GRCh38
rs772472003 Chr.13: 21140844-21140844 on Build GRCh38
rs754135538 Chr.13: 21140712-21140712 on Build GRCh38
rs758919465 Chr.13: 21140686-21140687 on Build GRCh38
rs781236337 Chr.13: 21140755-21140755 on Build GRCh38
rs762835068 Chr.13: 21140577-21140577 on Build GRCh38
rs369212638 Chr.13: 21140855-21140855 on Build GRCh38
rs751259780 Chr.13: 21140730-21140730 on Build GRCh38
rs563914498 Chr.13: 21140444-21140444 on Build GRCh38
rs767488618 Chr.13: 21140879-21140879 on Build GRCh38
rs764700763 Chr.13: 21140894-21140894 on Build GRCh38
rs941311914 Chr.13: 21140464-21140464 on Build GRCh38
rs758288241 Chr.13: 21140579-21140579 on Build GRCh38
rs368803461 Chr.13: 21140702-21140702 on Build GRCh38
rs764614475 Chr.13: 21140573-21140573 on Build GRCh38
rs763338973 Chr.13: 21140589-21140589 on Build GRCh38
rs750446218 Chr.13: 21140833-21140833 on Build GRCh38
rs760672805 Chr.13: 21140882-21140882 on Build GRCh38
rs572997131 Chr.13: 21140845-21140845 on Build GRCh38
rs540223029 Chr.13: 21140519-21140519 on Build GRCh38
rs112253706 Chr.13: 21140682-21140682 on Build GRCh38
rs759820243 Chr.13: 21140819-21140819 on Build GRCh38
rs751601385 Chr.13: 21140544-21140544 on Build GRCh38
rs908412190 Chr.13: 21140566-21140566 on Build GRCh38
rs769888253 Chr.13: 21140862-21140862 on Build GRCh38
rs200122454 Chr.13: 21140515-21140515 on Build GRCh38
rs113022305 Chr.13: 21140683-21140683 on Build GRCh38
rs771098354 Chr.13: 21140812-21140812 on Build GRCh38
rs758249818 Chr.13: 21140624-21140624 on Build GRCh38
rs763957224 Chr.13: 21140721-21140721 on Build GRCh38
rs530592109 Chr.13: 21140578-21140578 on Build GRCh38
rs564217124 Chr.13: 21140500-21140500 on Build GRCh38
rs922366222 Chr.13: 21140446-21140446 on Build GRCh38
rs777899487 Chr.13: 21140511-21140511 on Build GRCh38
rs893298516 Chr.13: 21140520-21140520 on Build GRCh38
rs1038281216 Chr.13: 21140497-21140497 on Build GRCh38
rs745325674 Chr.13: 21140664-21140664 on Build GRCh38
rs757286290 Chr.13: 21140742-21140742 on Build GRCh38
rs763649081 Chr.13: 21140599-21140599 on Build GRCh38
rs1053219185 Chr.13: 21140494-21140494 on Build GRCh38
rs762060747 Chr.13: 21140681-21140682 on Build GRCh38
rs771292591 Chr.13: 21140537-21140537 on Build GRCh38
rs780712267 Chr.13: 21140718-21140718 on Build GRCh38
rs751622316 Chr.13: 21140574-21140574 on Build GRCh38
rs570548873 Chr.13: 21140599-21140599 on Build GRCh38
rs955130207 Chr.13: 21140551-21140551 on Build GRCh38
rs754483393 Chr.13: 21140888-21140888 on Build GRCh38
rs202209747 Chr.13: 21140864-21140864 on Build GRCh38
rs368287644 Chr.13: 21140570-21140570 on Build GRCh38
rs982643722 Chr.13: 21140460-21140460 on Build GRCh38
rs190705962 Chr.13: 21140442-21140442 on Build GRCh38
rs2281914 Chr.13: 21140781-21140781 on Build GRCh38
rs767650009 Chr.13: 21140601-21140601 on Build GRCh38
rs757198379 Chr.13: 21140660-21140660 on Build GRCh38

View Full Product Details