Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12206649-12207151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF1B
Entrez Gene ID
7133
Gene Name
TNF receptor superfamily member 1B
Gene Aliases
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR
UniGene
Hs.256278
Gene Chromosome Location
Chr.1: 12166948-12209222 on Build GRCh38
Transcripts
NM_001066.2 XM_011542060.2 XM_011542063.2 XM_017002214.1 XM_017002215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4406007 c.1270T>G p.S424A Chr.1: 12206904-12206904 on Build GRCh38
6019437 c.1379C>G p.P460R Chr.1: 12207013-12207013 on Build GRCh38
1687012 c.1215G>A p.M405I Chr.1: 12206849-12206849 on Build GRCh38
4021362 c.1382G>A p.S461N Chr.1: 12207016-12207016 on Build GRCh38
1268215 c.1150_1164del15 p.I384_C388delIVNVC Chr.1: 12206784-12206798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564369569 Chr.1: 12206721-12206721 on Build GRCh38
rs776056858 Chr.1: 12206867-12206867 on Build GRCh38
rs781203144 Chr.1: 12206810-12206810 on Build GRCh38
rs149512902 Chr.1: 12206909-12206909 on Build GRCh38
rs371676482 Chr.1: 12207045-12207045 on Build GRCh38
rs773470753 Chr.1: 12206966-12206966 on Build GRCh38
rs774465726 Chr.1: 12206943-12206943 on Build GRCh38
rs55925819 Chr.1: 12206758-12206758 on Build GRCh38
rs771452615 Chr.1: 12206848-12206848 on Build GRCh38
rs896289977 Chr.1: 12207107-12207107 on Build GRCh38
rs531788500 Chr.1: 12206881-12206881 on Build GRCh38
rs555457691 Chr.1: 12206717-12206717 on Build GRCh38
rs575469521 Chr.1: 12206675-12206675 on Build GRCh38
rs148178396 Chr.1: 12206706-12206706 on Build GRCh38
rs746610371 Chr.1: 12207026-12207026 on Build GRCh38
rs773764020 Chr.1: 12206702-12206702 on Build GRCh38
rs780895721 Chr.1: 12206821-12206821 on Build GRCh38
rs902150853 Chr.1: 12206891-12206891 on Build GRCh38
rs368365494 Chr.1: 12207027-12207027 on Build GRCh38
rs750229037 Chr.1: 12206882-12206882 on Build GRCh38
rs917000076 Chr.1: 12206885-12206885 on Build GRCh38
rs758171297 Chr.1: 12206922-12206922 on Build GRCh38
rs763056084 Chr.1: 12206795-12206795 on Build GRCh38
rs767165937 Chr.1: 12207022-12207022 on Build GRCh38
rs752767671 Chr.1: 12206792-12206792 on Build GRCh38
rs760912981 Chr.1: 12207067-12207067 on Build GRCh38
rs753644221 Chr.1: 12207108-12207108 on Build GRCh38
rs754913078 Chr.1: 12206981-12206981 on Build GRCh38
rs150675043 Chr.1: 12207042-12207042 on Build GRCh38
rs753751204 Chr.1: 12206893-12206893 on Build GRCh38
rs770243493 Chr.1: 12206866-12206866 on Build GRCh38
rs975718434 Chr.1: 12206764-12206764 on Build GRCh38
rs955614166 Chr.1: 12207037-12207037 on Build GRCh38
rs373722040 Chr.1: 12206926-12206926 on Build GRCh38
rs555160047 Chr.1: 12207069-12207069 on Build GRCh38
rs765379693 Chr.1: 12206786-12206786 on Build GRCh38
rs777283533 Chr.1: 12206783-12206783 on Build GRCh38
rs748689571 Chr.1: 12206857-12206857 on Build GRCh38
rs371983819 Chr.1: 12207028-12207028 on Build GRCh38
rs375627878 Chr.1: 12207006-12207006 on Build GRCh38
rs370414826 Chr.1: 12206818-12206818 on Build GRCh38
rs1008701992 Chr.1: 12206720-12206720 on Build GRCh38
rs781160620 Chr.1: 12206748-12206748 on Build GRCh38
rs764108902 Chr.1: 12207081-12207081 on Build GRCh38
rs747823545 Chr.1: 12206921-12206921 on Build GRCh38
rs201385323 Chr.1: 12207059-12207059 on Build GRCh38
rs754383725 Chr.1: 12206707-12206707 on Build GRCh38
rs575457978 Chr.1: 12207087-12207087 on Build GRCh38
rs1013427348 Chr.1: 12206884-12206884 on Build GRCh38
rs754452683 Chr.1: 12206894-12206894 on Build GRCh38
rs753752076 Chr.1: 12206979-12206979 on Build GRCh38
rs755064457 Chr.1: 12206763-12206763 on Build GRCh38
rs780013560 Chr.1: 12206830-12206830 on Build GRCh38
rs866383062 Chr.1: 12206746-12206746 on Build GRCh38
rs919987446 Chr.1: 12207075-12207075 on Build GRCh38
rs142002010 Chr.1: 12206726-12206726 on Build GRCh38
rs1002434839 Chr.1: 12206874-12206874 on Build GRCh38
rs745744593 Chr.1: 12206749-12206749 on Build GRCh38
rs560260854 Chr.1: 12206750-12206750 on Build GRCh38
rs780321022 Chr.1: 12206696-12206696 on Build GRCh38
rs761978081 Chr.1: 12206873-12206873 on Build GRCh38
rs774062294 Chr.1: 12206941-12206941 on Build GRCh38
rs767669838 Chr.1: 12207054-12207054 on Build GRCh38
rs549171418 Chr.1: 12206780-12206780 on Build GRCh38
rs767949672 Chr.1: 12206697-12206697 on Build GRCh38
rs773618391 Chr.1: 12206872-12206872 on Build GRCh38
rs199689616 Chr.1: 12207040-12207040 on Build GRCh38
rs1042284268 Chr.1: 12207127-12207127 on Build GRCh38
rs374945013 Chr.1: 12207062-12207062 on Build GRCh38
rs755122734 Chr.1: 12206710-12206710 on Build GRCh38
rs781014897 Chr.1: 12206747-12206747 on Build GRCh38
rs200600328 Chr.1: 12206955-12206955 on Build GRCh38
rs770178506 Chr.1: 12206935-12206935 on Build GRCh38
rs750384859 Chr.1: 12206825-12206825 on Build GRCh38
rs370292777 Chr.1: 12206902-12206902 on Build GRCh38
rs376576479 Chr.1: 12206836-12206836 on Build GRCh38
rs765241635 Chr.1: 12206985-12206985 on Build GRCh38
rs571590743 Chr.1: 12206925-12206925 on Build GRCh38
rs150855008 Chr.1: 12206888-12206888 on Build GRCh38
rs757038561 Chr.1: 12206744-12206744 on Build GRCh38
rs368107139 Chr.1: 12207043-12207043 on Build GRCh38
rs759919319 Chr.1: 12206784-12206784 on Build GRCh38
rs775871058 Chr.1: 12206940-12206940 on Build GRCh38
rs1055074788 Chr.1: 12206875-12206875 on Build GRCh38
rs75299314 Chr.1: 12206743-12206743 on Build GRCh38
rs200538351 Chr.1: 12206989-12206989 on Build GRCh38
rs747764180 Chr.1: 12207084-12207085 on Build GRCh38
rs748817002 Chr.1: 12206831-12206831 on Build GRCh38
rs561824368 Chr.1: 12206673-12206673 on Build GRCh38
rs766286313 Chr.1: 12206889-12206889 on Build GRCh38
rs139665575 Chr.1: 12206870-12206870 on Build GRCh38
rs1030189043 Chr.1: 12206676-12206676 on Build GRCh38
rs759922521 Chr.1: 12206859-12206859 on Build GRCh38
rs529293509 Chr.1: 12206768-12206768 on Build GRCh38
rs762477735 Chr.1: 12206690-12206690 on Build GRCh38
rs371439340 Chr.1: 12206970-12206970 on Build GRCh38
rs966448361 Chr.1: 12206718-12206718 on Build GRCh38
rs762997433 Chr.1: 12206869-12206869 on Build GRCh38
rs773310858 Chr.1: 12207060-12207060 on Build GRCh38
rs767798835 Chr.1: 12206965-12206965 on Build GRCh38
rs778032363 Chr.1: 12206725-12206725 on Build GRCh38
rs778723682 Chr.1: 12206839-12206839 on Build GRCh38
rs777085409 Chr.1: 12206858-12206858 on Build GRCh38
rs768236405 Chr.1: 12206766-12206766 on Build GRCh38
rs142879753 Chr.1: 12206826-12206826 on Build GRCh38
rs370508215 Chr.1: 12207082-12207082 on Build GRCh38
rs752294850 Chr.1: 12206994-12206994 on Build GRCh38
rs764362275 Chr.1: 12206799-12206799 on Build GRCh38
rs938086908 Chr.1: 12207086-12207086 on Build GRCh38
rs368698604 Chr.1: 12206942-12206942 on Build GRCh38
rs1055380816 Chr.1: 12207106-12207106 on Build GRCh38

View Full Product Details