Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 13583667-13584162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDPN
Entrez Gene ID
10630
Gene Name
podoplanin
Gene Aliases
AGGRUS, GP36, GP40, Gp38, HT1A-1, OTS8, PA2.26, T1A, T1A-2, T1A2, TI1A
UniGene
Hs.468675
Gene Chromosome Location
Chr.1: 13583757-13617957 on Build GRCh38
Transcripts
NM_006474.4 NM_198389.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7201550 c.47G>T p.R16L Chr.1: 13583852-13583852 on Build GRCh38
7205578 c.170T>C p.F57S Chr.1: 13583975-13583975 on Build GRCh38
4713401 c.112_113insC p.P41fs*67 Chr.1: 13583917-13583918 on Build GRCh38
6743433 c.126G>A p.S42S Chr.1: 13583931-13583931 on Build GRCh38
4805536 c.205G>T p.E69* Chr.1: 13584010-13584010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777064736 Chr.1: 13583770-13583770 on Build GRCh38
rs367829245 Chr.1: 13584105-13584105 on Build GRCh38
rs140760055 Chr.1: 13584068-13584068 on Build GRCh38
rs779072490 Chr.1: 13583918-13583918 on Build GRCh38
rs755588997 Chr.1: 13583807-13583807 on Build GRCh38
rs529805848 Chr.1: 13583911-13583911 on Build GRCh38
rs141040955 Chr.1: 13584033-13584033 on Build GRCh38
rs748693505 Chr.1: 13583756-13583756 on Build GRCh38
rs763889776 Chr.1: 13583786-13583786 on Build GRCh38
rs201766470 Chr.1: 13583910-13583910 on Build GRCh38
rs111795535 Chr.1: 13584052-13584052 on Build GRCh38
rs752160983 Chr.1: 13583850-13583850 on Build GRCh38
rs771063439 Chr.1: 13583766-13583766 on Build GRCh38
rs370833657 Chr.1: 13583934-13583934 on Build GRCh38
rs758533099 Chr.1: 13583972-13583972 on Build GRCh38
rs777666469 Chr.1: 13583974-13583974 on Build GRCh38
rs781546601 Chr.1: 13583822-13583822 on Build GRCh38
rs909745893 Chr.1: 13583859-13583859 on Build GRCh38
rs753292713 Chr.1: 13583844-13583844 on Build GRCh38
rs1035742770 Chr.1: 13583715-13583715 on Build GRCh38
rs750365640 Chr.1: 13584128-13584128 on Build GRCh38
rs368134560 Chr.1: 13583958-13583958 on Build GRCh38
rs770812249 Chr.1: 13583923-13583923 on Build GRCh38
rs761020351 Chr.1: 13584093-13584093 on Build GRCh38
rs975219481 Chr.1: 13583705-13583705 on Build GRCh38
rs750124928 Chr.1: 13583803-13583803 on Build GRCh38
rs771804597 Chr.1: 13583872-13583872 on Build GRCh38
rs773006448 Chr.1: 13583873-13583873 on Build GRCh38
rs772443538 Chr.1: 13583760-13583760 on Build GRCh38
rs202164928 Chr.1: 13583847-13583847 on Build GRCh38
rs771483531 Chr.1: 13583821-13583821 on Build GRCh38
rs769957523 Chr.1: 13583824-13583824 on Build GRCh38
rs373560383 Chr.1: 13583874-13583874 on Build GRCh38
rs199557731 Chr.1: 13583937-13583937 on Build GRCh38
rs761839708 Chr.1: 13583870-13583870 on Build GRCh38
rs141888769 Chr.1: 13584091-13584091 on Build GRCh38
rs200437395 Chr.1: 13584116-13584116 on Build GRCh38
rs954876027 Chr.1: 13583750-13583750 on Build GRCh38
rs765459650 Chr.1: 13583776-13583776 on Build GRCh38
rs534158698 Chr.1: 13583951-13583951 on Build GRCh38
rs890584138 Chr.1: 13584097-13584097 on Build GRCh38
rs772685858 Chr.1: 13583987-13583988 on Build GRCh38
rs767340937 Chr.1: 13583838-13583838 on Build GRCh38
rs142084255 Chr.1: 13583924-13583924 on Build GRCh38
rs549422402 Chr.1: 13583868-13583868 on Build GRCh38
rs1018449481 Chr.1: 13583921-13583921 on Build GRCh38
rs958170856 Chr.1: 13583689-13583689 on Build GRCh38
rs751311332 Chr.1: 13583792-13583792 on Build GRCh38
rs187344388 Chr.1: 13583858-13583858 on Build GRCh38
rs764847742 Chr.1: 13583885-13583885 on Build GRCh38
rs747068653 Chr.1: 13584141-13584141 on Build GRCh38
rs996168189 Chr.1: 13583889-13583889 on Build GRCh38
rs751453337 Chr.1: 13584065-13584065 on Build GRCh38
rs765819037 Chr.1: 13583841-13583841 on Build GRCh38
rs745456102 Chr.1: 13583985-13583985 on Build GRCh38
rs774826187 Chr.1: 13583917-13583918 on Build GRCh38
rs1025334377 Chr.1: 13583927-13583927 on Build GRCh38
rs753957633 Chr.1: 13583953-13583953 on Build GRCh38
rs962601555 Chr.1: 13583772-13583772 on Build GRCh38
rs752879040 Chr.1: 13584113-13584113 on Build GRCh38
rs765486157 Chr.1: 13584050-13584050 on Build GRCh38
rs781716183 Chr.1: 13583853-13583853 on Build GRCh38
rs987598118 Chr.1: 13583754-13583754 on Build GRCh38
rs758445061 Chr.1: 13584055-13584055 on Build GRCh38
rs750036508 Chr.1: 13583839-13583839 on Build GRCh38
rs78133275 Chr.1: 13584026-13584026 on Build GRCh38
rs532055309 Chr.1: 13583901-13583901 on Build GRCh38
rs868794296 Chr.1: 13583936-13583936 on Build GRCh38
rs768828741 Chr.1: 13583864-13583864 on Build GRCh38
rs769478918 Chr.1: 13583986-13583986 on Build GRCh38
rs949134122 Chr.1: 13584013-13584013 on Build GRCh38
rs355025 Chr.1: 13583922-13583922 on Build GRCh38
rs951119460 Chr.1: 13584142-13584142 on Build GRCh38
rs980612745 Chr.1: 13583997-13583997 on Build GRCh38
rs112177943 Chr.1: 13583708-13583708 on Build GRCh38
rs768229516 Chr.1: 13584082-13584082 on Build GRCh38
rs762435358 Chr.1: 13583941-13583941 on Build GRCh38
rs560374422 Chr.1: 13584113-13584114 on Build GRCh38
rs753201185 Chr.1: 13583816-13583816 on Build GRCh38
rs767924535 Chr.1: 13583894-13583894 on Build GRCh38
rs1046236900 Chr.1: 13584018-13584018 on Build GRCh38
rs779243954 Chr.1: 13583902-13583902 on Build GRCh38
rs375753913 Chr.1: 13583856-13583856 on Build GRCh38
rs762817453 Chr.1: 13583783-13583783 on Build GRCh38
rs746026348 Chr.1: 13583854-13583854 on Build GRCh38
rs768469466 Chr.1: 13583829-13583829 on Build GRCh38
rs756514325 Chr.1: 13583940-13583940 on Build GRCh38
rs149456565 Chr.1: 13583919-13583919 on Build GRCh38
rs779818780 Chr.1: 13584081-13584081 on Build GRCh38
rs758915594 Chr.1: 13584010-13584010 on Build GRCh38
rs759884770 Chr.1: 13584046-13584046 on Build GRCh38
rs773813176 Chr.1: 13584017-13584017 on Build GRCh38
rs777864670 Chr.1: 13584060-13584060 on Build GRCh38
rs548210721 Chr.1: 13584021-13584021 on Build GRCh38
rs757864690 Chr.1: 13583893-13583893 on Build GRCh38
rs1029052634 Chr.1: 13583722-13583722 on Build GRCh38
rs151117818 Chr.1: 13583820-13583820 on Build GRCh38
rs777062623 Chr.1: 13584103-13584103 on Build GRCh38
rs774667116 Chr.1: 13584049-13584049 on Build GRCh38
rs141254869 Chr.1: 13583883-13583883 on Build GRCh38
rs34662476 Chr.1: 13584123-13584124 on Build GRCh38
rs201233313 Chr.1: 13584000-13584000 on Build GRCh38
rs758844371 Chr.1: 13583817-13583817 on Build GRCh38
rs577807103 Chr.1: 13583840-13583840 on Build GRCh38
rs113350533 Chr.1: 13583851-13583851 on Build GRCh38
rs759679360 Chr.1: 13583774-13583774 on Build GRCh38
rs376115475 Chr.1: 13583886-13583886 on Build GRCh38
rs778134240 Chr.1: 13583818-13583818 on Build GRCh38
rs767142794 Chr.1: 13583800-13583800 on Build GRCh38
rs1002956020 Chr.1: 13583702-13583702 on Build GRCh38
rs761731257 Chr.1: 13583793-13583793 on Build GRCh38
rs192071055 Chr.1: 13584123-13584123 on Build GRCh38
rs969361545 Chr.1: 13583785-13583785 on Build GRCh38
rs775336326 Chr.1: 13583991-13583991 on Build GRCh38
rs757335369 Chr.1: 13584124-13584124 on Build GRCh38
rs985708917 Chr.1: 13583791-13583791 on Build GRCh38
rs773474024 Chr.1: 13583765-13583765 on Build GRCh38
rs776516469 Chr.1: 13583980-13583980 on Build GRCh38
rs757235677 Chr.1: 13584071-13584071 on Build GRCh38
rs775540618 Chr.1: 13583782-13583782 on Build GRCh38
rs972466333 Chr.1: 13583778-13583778 on Build GRCh38
rs745457951 Chr.1: 13583837-13583838 on Build GRCh38
rs776403428 Chr.1: 13583875-13583875 on Build GRCh38
rs564395708 Chr.1: 13583823-13583823 on Build GRCh38
rs754970758 Chr.1: 13583912-13583912 on Build GRCh38
rs1054817819 Chr.1: 13583795-13583795 on Build GRCh38
rs147939246 Chr.1: 13584045-13584045 on Build GRCh38
rs746992405 Chr.1: 13583976-13583976 on Build GRCh38
rs529372840 Chr.1: 13583862-13583862 on Build GRCh38
rs781054465 Chr.1: 13584075-13584075 on Build GRCh38
rs781163541 Chr.1: 13584127-13584127 on Build GRCh38
rs765632676 Chr.1: 13584110-13584110 on Build GRCh38
rs367551277 Chr.1: 13584133-13584133 on Build GRCh38
rs770593402 Chr.1: 13583978-13583978 on Build GRCh38
rs755241481 Chr.1: 13583954-13583954 on Build GRCh38
rs774401177 Chr.1: 13583833-13583833 on Build GRCh38
rs761490614 Chr.1: 13583837-13583837 on Build GRCh38
rs745658026 Chr.1: 13584080-13584080 on Build GRCh38
rs369019664 Chr.1: 13583825-13583825 on Build GRCh38
rs779570316 Chr.1: 13583808-13583808 on Build GRCh38
rs769355813 Chr.1: 13583903-13583911 on Build GRCh38
rs764107863 Chr.1: 13584115-13584115 on Build GRCh38
rs771466218 Chr.1: 13584095-13584095 on Build GRCh38
rs748221735 Chr.1: 13583960-13583960 on Build GRCh38
rs745375098 Chr.1: 13583755-13583755 on Build GRCh38
rs780881002 Chr.1: 13583838-13583839 on Build GRCh38

View Full Product Details