Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30917391-30917937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEDAG
Entrez Gene ID
84935
Gene Name
mesenteric estrogen dependent adipogenesis
Gene Aliases
AWMS3, C13orf33, MEDA-4, MEDA4, hAWMS3
UniGene
Hs.147880
Gene Chromosome Location
Chr.13: 30906175-30925577 on Build GRCh38
Transcripts
NM_032849.3 XM_017020801.1 NR_038287.1

Gene Symbol
TEX26-AS1
Entrez Gene ID
100507064
Gene Name
TEX26 antisense RNA 1
Gene Aliases
LINC00447
UniGene
Hs.654878
Gene Chromosome Location
Chr.13: 30882835-30932608 on Build GRCh38
Transcripts
NM_032849.3 XM_017020801.1 NR_038287.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
342819 c.330A>T p.I110I Chr.13: 30917454-30917454 on Build GRCh38
5184754 c.320G>T p.R107M Chr.13: 30917444-30917444 on Build GRCh38
5677121 c.388+1G>T p.? Chr.13: 30917513-30917513 on Build GRCh38
6015292 c.279-5C>T p.? Chr.13: 30917398-30917398 on Build GRCh38
6413594 c.364C>A p.Q122K Chr.13: 30917488-30917488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571140586 Chr.13: 30917892-30917892 on Build GRCh38
rs138323245 Chr.13: 30917723-30917723 on Build GRCh38
rs955170989 Chr.13: 30917730-30917730 on Build GRCh38
rs749004022 Chr.13: 30917424-30917424 on Build GRCh38
rs117860465 Chr.13: 30917701-30917701 on Build GRCh38
rs7982266 Chr.13: 30917707-30917707 on Build GRCh38
rs186294247 Chr.13: 30917603-30917603 on Build GRCh38
rs1003103616 Chr.13: 30917690-30917690 on Build GRCh38
rs778636280 Chr.13: 30917619-30917619 on Build GRCh38
rs757064712 Chr.13: 30917608-30917608 on Build GRCh38
rs777204638 Chr.13: 30917515-30917515 on Build GRCh38
rs758117009 Chr.13: 30917648-30917648 on Build GRCh38
rs142736520 Chr.13: 30917729-30917729 on Build GRCh38
rs150419121 Chr.13: 30917450-30917450 on Build GRCh38
rs956113546 Chr.13: 30917793-30917793 on Build GRCh38
rs376316063 Chr.13: 30917451-30917451 on Build GRCh38
rs768974181 Chr.13: 30917489-30917489 on Build GRCh38
rs1047850669 Chr.13: 30917906-30917906 on Build GRCh38
rs1016695809 Chr.13: 30917513-30917513 on Build GRCh38
rs886695921 Chr.13: 30917911-30917911 on Build GRCh38
rs188329433 Chr.13: 30917842-30917842 on Build GRCh38
rs372770078 Chr.13: 30917556-30917556 on Build GRCh38
rs986605788 Chr.13: 30917752-30917752 on Build GRCh38
rs866228837 Chr.13: 30917670-30917670 on Build GRCh38
rs561192906 Chr.13: 30917673-30917673 on Build GRCh38
rs768204734 Chr.13: 30917444-30917444 on Build GRCh38
rs951032321 Chr.13: 30917464-30917464 on Build GRCh38
rs200213117 Chr.13: 30917454-30917454 on Build GRCh38
rs1000087172 Chr.13: 30917593-30917593 on Build GRCh38
rs747405518 Chr.13: 30917487-30917487 on Build GRCh38
rs955306237 Chr.13: 30917604-30917604 on Build GRCh38
rs975274042 Chr.13: 30917565-30917565 on Build GRCh38
rs773922479 Chr.13: 30917437-30917437 on Build GRCh38
rs1022765497 Chr.13: 30917535-30917535 on Build GRCh38
rs1031602330 Chr.13: 30917787-30917787 on Build GRCh38
rs542537001 Chr.13: 30917636-30917636 on Build GRCh38
rs761335981 Chr.13: 30917441-30917441 on Build GRCh38
rs774830096 Chr.13: 30917902-30917902 on Build GRCh38
rs891493466 Chr.13: 30917740-30917740 on Build GRCh38
rs59319457 Chr.13: 30917905-30917905 on Build GRCh38
rs754103414 Chr.13: 30917465-30917465 on Build GRCh38
rs992851725 Chr.13: 30917908-30917908 on Build GRCh38
rs764020070 Chr.13: 30917572-30917572 on Build GRCh38
rs940122276 Chr.13: 30917521-30917521 on Build GRCh38
rs556772294 Chr.13: 30917526-30917526 on Build GRCh38
rs1008564113 Chr.13: 30917741-30917741 on Build GRCh38
rs1012754210 Chr.13: 30917525-30917525 on Build GRCh38
rs371697036 Chr.13: 30917425-30917425 on Build GRCh38
rs1047985871 Chr.13: 30917697-30917697 on Build GRCh38
rs548908896 Chr.13: 30917815-30917815 on Build GRCh38
rs758463427 Chr.13: 30917523-30917523 on Build GRCh38
rs752890351 Chr.13: 30917505-30917505 on Build GRCh38
rs940028764 Chr.13: 30917700-30917700 on Build GRCh38
rs563094866 Chr.13: 30917794-30917794 on Build GRCh38
rs1004262664 Chr.13: 30917504-30917504 on Build GRCh38
rs768354654 Chr.13: 30917557-30917557 on Build GRCh38
rs765309979 Chr.13: 30917492-30917492 on Build GRCh38
rs1018990209 Chr.13: 30917791-30917791 on Build GRCh38
rs948860695 Chr.13: 30917848-30917848 on Build GRCh38
rs887091902 Chr.13: 30917517-30917517 on Build GRCh38
rs779917597 Chr.13: 30917542-30917542 on Build GRCh38
rs139007786 Chr.13: 30917870-30917870 on Build GRCh38
rs140189386 Chr.13: 30917452-30917452 on Build GRCh38
rs546604227 Chr.13: 30917887-30917887 on Build GRCh38
rs565110706 Chr.13: 30917718-30917718 on Build GRCh38
rs1016970877 Chr.13: 30917714-30917714 on Build GRCh38
rs952868708 Chr.13: 30917827-30917827 on Build GRCh38
rs755243659 Chr.13: 30917475-30917475 on Build GRCh38
rs181982642 Chr.13: 30917524-30917524 on Build GRCh38
rs779559542 Chr.13: 30917672-30917672 on Build GRCh38
rs535224579 Chr.13: 30917835-30917835 on Build GRCh38
rs908565536 Chr.13: 30917699-30917699 on Build GRCh38
rs760472929 Chr.13: 30917638-30917638 on Build GRCh38
rs145005980 Chr.13: 30917808-30917808 on Build GRCh38
rs756008683 Chr.13: 30917540-30917540 on Build GRCh38
rs904449425 Chr.13: 30917590-30917590 on Build GRCh38
rs867307756 Chr.13: 30917490-30917490 on Build GRCh38
rs907368228 Chr.13: 30917689-30917689 on Build GRCh38
rs771534513 Chr.13: 30917807-30917807 on Build GRCh38
rs749774149 Chr.13: 30917769-30917769 on Build GRCh38

View Full Product Details