Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 35770082-35770595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLK1
Entrez Gene ID
9201
Gene Name
doublecortin like kinase 1
Gene Aliases
CL1, CLICK1, DCAMKL1, DCDC3A, DCLK
UniGene
Hs.507755
Gene Chromosome Location
Chr.13: 35768652-36132020 on Build GRCh38
Transcripts
NM_001195415.1 NM_001195416.1 NM_004734.4 XM_005266592.2 XM_006719893.2 XM_017020847.1

SNP Detail

SNP ID Chromosome Location
rs996502000 Chr.13: 35770519-35770519 on Build GRCh38
rs922363349 Chr.13: 35770225-35770225 on Build GRCh38
rs1045176915 Chr.13: 35770135-35770135 on Build GRCh38
rs934090832 Chr.13: 35770209-35770209 on Build GRCh38
rs887284760 Chr.13: 35770472-35770472 on Build GRCh38
rs9574542 Chr.13: 35770297-35770297 on Build GRCh38
rs146094094 Chr.13: 35770246-35770247 on Build GRCh38
rs547010943 Chr.13: 35770512-35770512 on Build GRCh38
rs1045083252 Chr.13: 35770417-35770417 on Build GRCh38
rs975008872 Chr.13: 35770196-35770196 on Build GRCh38
rs766111324 Chr.13: 35770200-35770200 on Build GRCh38
rs1029281919 Chr.13: 35770545-35770545 on Build GRCh38
rs946973517 Chr.13: 35770137-35770137 on Build GRCh38
rs915469280 Chr.13: 35770137-35770139 on Build GRCh38
rs906583652 Chr.13: 35770138-35770138 on Build GRCh38
rs533987730 Chr.13: 35770361-35770361 on Build GRCh38
rs116097087 Chr.13: 35770107-35770107 on Build GRCh38
rs906657300 Chr.13: 35770430-35770430 on Build GRCh38
rs142793851 Chr.13: 35770125-35770125 on Build GRCh38
rs146087439 Chr.13: 35770496-35770496 on Build GRCh38
rs1034754224 Chr.13: 35770412-35770412 on Build GRCh38
rs561655891 Chr.13: 35770422-35770422 on Build GRCh38
rs757420107 Chr.13: 35770231-35770231 on Build GRCh38
rs759123233 Chr.13: 35770270-35770270 on Build GRCh38
rs9531012 Chr.13: 35770290-35770290 on Build GRCh38
rs934183462 Chr.13: 35770558-35770558 on Build GRCh38
rs899046570 Chr.13: 35770320-35770320 on Build GRCh38
rs757879463 Chr.13: 35770463-35770463 on Build GRCh38
rs543656778 Chr.13: 35770392-35770392 on Build GRCh38
rs182972714 Chr.13: 35770312-35770312 on Build GRCh38
rs1023463218 Chr.13: 35770351-35770351 on Build GRCh38
rs1004020981 Chr.13: 35770444-35770444 on Build GRCh38
rs776169533 Chr.13: 35770185-35770185 on Build GRCh38
rs1006202255 Chr.13: 35770162-35770162 on Build GRCh38
rs192665343 Chr.13: 35770186-35770186 on Build GRCh38
rs987739403 Chr.13: 35770441-35770444 on Build GRCh38
rs751460905 Chr.13: 35770499-35770499 on Build GRCh38
rs9574543 Chr.13: 35770389-35770389 on Build GRCh38
rs1003750245 Chr.13: 35770143-35770143 on Build GRCh38
rs1005693198 Chr.13: 35770478-35770478 on Build GRCh38
rs201142068 Chr.13: 35770569-35770570 on Build GRCh38
rs955116181 Chr.13: 35770229-35770229 on Build GRCh38
rs951688496 Chr.13: 35770151-35770151 on Build GRCh38
rs9545304 Chr.13: 35770349-35770349 on Build GRCh38
rs112448374 Chr.13: 35770464-35770464 on Build GRCh38
rs1026233296 Chr.13: 35770149-35770149 on Build GRCh38
rs572603106 Chr.13: 35770314-35770314 on Build GRCh38
rs368889831 Chr.13: 35770447-35770447 on Build GRCh38
rs1039840834 Chr.13: 35770175-35770175 on Build GRCh38

View Full Product Details