Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40555837-40556337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXO1
Entrez Gene ID
2308
Gene Name
forkhead box O1
Gene Aliases
FKH1, FKHR, FOXO1A
UniGene
Hs.370666
Gene Chromosome Location
Chr.13: 40555664-40666597 on Build GRCh38
Transcripts
NM_002015.3 XM_011535008.2 XM_011535010.2

SNP Detail

SNP ID Chromosome Location
rs530412976 Chr.13: 40555954-40555954 on Build GRCh38
rs536362400 Chr.13: 40556151-40556151 on Build GRCh38
rs766433687 Chr.13: 40556205-40556205 on Build GRCh38
rs1031434039 Chr.13: 40555864-40555864 on Build GRCh38
rs907585055 Chr.13: 40555965-40555965 on Build GRCh38
rs567201093 Chr.13: 40556083-40556083 on Build GRCh38
rs763945786 Chr.13: 40556024-40556024 on Build GRCh38
rs965532131 Chr.13: 40555889-40555889 on Build GRCh38
rs1043974474 Chr.13: 40556071-40556071 on Build GRCh38
rs907720873 Chr.13: 40556281-40556281 on Build GRCh38
rs987294749 Chr.13: 40556270-40556270 on Build GRCh38
rs551681464 Chr.13: 40555931-40555931 on Build GRCh38
rs182616961 Chr.13: 40555930-40555930 on Build GRCh38
rs1031778671 Chr.13: 40556185-40556185 on Build GRCh38
rs369371669 Chr.13: 40555964-40555964 on Build GRCh38
rs144573709 Chr.13: 40555908-40555908 on Build GRCh38
rs565604388 Chr.13: 40555941-40555941 on Build GRCh38
rs940528830 Chr.13: 40556285-40556285 on Build GRCh38
rs550595286 Chr.13: 40555994-40555994 on Build GRCh38
rs998379920 Chr.13: 40556209-40556209 on Build GRCh38
rs536960434 Chr.13: 40556271-40556271 on Build GRCh38
rs973138152 Chr.13: 40556303-40556304 on Build GRCh38
rs920377434 Chr.13: 40556308-40556308 on Build GRCh38
rs1055211631 Chr.13: 40556048-40556048 on Build GRCh38
rs902745873 Chr.13: 40556090-40556090 on Build GRCh38
rs984048530 Chr.13: 40555938-40555938 on Build GRCh38
rs940019537 Chr.13: 40555974-40555974 on Build GRCh38
rs1036280547 Chr.13: 40556079-40556079 on Build GRCh38
rs953853381 Chr.13: 40555929-40555929 on Build GRCh38
rs1021690969 Chr.13: 40556199-40556199 on Build GRCh38
rs1037348084 Chr.13: 40555978-40555978 on Build GRCh38
rs754257786 Chr.13: 40555949-40555949 on Build GRCh38
rs983929628 Chr.13: 40556264-40556264 on Build GRCh38
rs1010996325 Chr.13: 40556070-40556070 on Build GRCh38
rs895352138 Chr.13: 40556065-40556065 on Build GRCh38
rs909759110 Chr.13: 40555959-40555959 on Build GRCh38
rs940436969 Chr.13: 40555972-40555972 on Build GRCh38
rs113132653 Chr.13: 40555863-40555863 on Build GRCh38
rs189937695 Chr.13: 40555920-40555920 on Build GRCh38
rs898815194 Chr.13: 40555989-40555989 on Build GRCh38
rs756343090 Chr.13: 40556092-40556092 on Build GRCh38
rs999770958 Chr.13: 40556174-40556174 on Build GRCh38
rs968712792 Chr.13: 40555888-40555888 on Build GRCh38
rs953865158 Chr.13: 40556232-40556232 on Build GRCh38
rs186612860 Chr.13: 40555902-40555902 on Build GRCh38
rs1010256872 Chr.13: 40556118-40556118 on Build GRCh38
rs546782697 Chr.13: 40556210-40556210 on Build GRCh38
rs927972457 Chr.13: 40555979-40555979 on Build GRCh38
rs574124830 Chr.13: 40555874-40555874 on Build GRCh38
rs1037409398 Chr.13: 40555977-40555977 on Build GRCh38
rs947661784 Chr.13: 40556309-40556309 on Build GRCh38

View Full Product Details