Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40559422-40559927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXO1
Entrez Gene ID
2308
Gene Name
forkhead box O1
Gene Aliases
FKH1, FKHR, FOXO1A
UniGene
Hs.370666
Gene Chromosome Location
Chr.13: 40555664-40666597 on Build GRCh38
Transcripts
NM_002015.3 XM_011535008.2 XM_011535010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6666460 c.1961C>G p.S654* Chr.13: 40559530-40559530 on Build GRCh38
356746 c.1649C>G p.S550W Chr.13: 40559842-40559842 on Build GRCh38
6980949 c.1897G>C p.D633H Chr.13: 40559594-40559594 on Build GRCh38
374141 c.1770G>T p.M590I Chr.13: 40559721-40559721 on Build GRCh38
4685447 c.1609C>T p.R537C Chr.13: 40559882-40559882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757736365 Chr.13: 40559625-40559625 on Build GRCh38
rs752155939 Chr.13: 40559613-40559613 on Build GRCh38
rs976584585 Chr.13: 40559457-40559457 on Build GRCh38
rs372059765 Chr.13: 40559902-40559902 on Build GRCh38
rs765387753 Chr.13: 40559561-40559561 on Build GRCh38
rs201186715 Chr.13: 40559803-40559803 on Build GRCh38
rs745385825 Chr.13: 40559635-40559635 on Build GRCh38
rs771921917 Chr.13: 40559685-40559685 on Build GRCh38
rs372159853 Chr.13: 40559709-40559709 on Build GRCh38
rs540866852 Chr.13: 40559714-40559714 on Build GRCh38
rs766830396 Chr.13: 40559865-40559865 on Build GRCh38
rs562138604 Chr.13: 40559459-40559459 on Build GRCh38
rs1018227132 Chr.13: 40559449-40559449 on Build GRCh38
rs148440850 Chr.13: 40559873-40559873 on Build GRCh38
rs1008631029 Chr.13: 40559872-40559872 on Build GRCh38
rs759124635 Chr.13: 40559448-40559448 on Build GRCh38
rs893465781 Chr.13: 40559837-40559837 on Build GRCh38
rs769354575 Chr.13: 40559534-40559534 on Build GRCh38
rs766520390 Chr.13: 40559551-40559551 on Build GRCh38
rs555894254 Chr.13: 40559452-40559452 on Build GRCh38
rs758892888 Chr.13: 40559590-40559590 on Build GRCh38
rs145453868 Chr.13: 40559866-40559866 on Build GRCh38
rs149675484 Chr.13: 40559842-40559842 on Build GRCh38
rs773141065 Chr.13: 40559700-40559700 on Build GRCh38
rs1009738135 Chr.13: 40559470-40559470 on Build GRCh38
rs867122605 Chr.13: 40559871-40559871 on Build GRCh38
rs367918340 Chr.13: 40559651-40559651 on Build GRCh38
rs144016210 Chr.13: 40559881-40559881 on Build GRCh38
rs987273613 Chr.13: 40559535-40559535 on Build GRCh38
rs778492835 Chr.13: 40559485-40559485 on Build GRCh38
rs950234021 Chr.13: 40559788-40559788 on Build GRCh38
rs778538205 Chr.13: 40559874-40559874 on Build GRCh38
rs781392626 Chr.13: 40559458-40559458 on Build GRCh38
rs768488321 Chr.13: 40559666-40559666 on Build GRCh38
rs920623940 Chr.13: 40559580-40559580 on Build GRCh38
rs139398811 Chr.13: 40559830-40559830 on Build GRCh38
rs755827604 Chr.13: 40559750-40559750 on Build GRCh38
rs770037212 Chr.13: 40559805-40559805 on Build GRCh38
rs778263954 Chr.13: 40559609-40559609 on Build GRCh38
rs781689181 Chr.13: 40559634-40559634 on Build GRCh38
rs771019295 Chr.13: 40559885-40559885 on Build GRCh38
rs752387735 Chr.13: 40559878-40559878 on Build GRCh38
rs929292024 Chr.13: 40559679-40559679 on Build GRCh38
rs747509005 Chr.13: 40559493-40559493 on Build GRCh38
rs749877266 Chr.13: 40559510-40559510 on Build GRCh38
rs1020662472 Chr.13: 40559504-40559504 on Build GRCh38
rs771830343 Chr.13: 40559761-40559761 on Build GRCh38
rs755586255 Chr.13: 40559640-40559640 on Build GRCh38
rs752897790 Chr.13: 40559567-40559567 on Build GRCh38
rs541544048 Chr.13: 40559559-40559559 on Build GRCh38
rs373830610 Chr.13: 40559785-40559785 on Build GRCh38
rs548058902 Chr.13: 40559886-40559886 on Build GRCh38
rs747930943 Chr.13: 40559673-40559673 on Build GRCh38
rs775906686 Chr.13: 40559712-40559712 on Build GRCh38
rs374930087 Chr.13: 40559506-40559506 on Build GRCh38
rs991177035 Chr.13: 40559894-40559894 on Build GRCh38
rs767915535 Chr.13: 40559734-40559734 on Build GRCh38
rs770526416 Chr.13: 40559494-40559494 on Build GRCh38
rs146488369 Chr.13: 40559882-40559882 on Build GRCh38
rs762743966 Chr.13: 40559541-40559541 on Build GRCh38
rs554193370 Chr.13: 40559560-40559560 on Build GRCh38
rs150896957 Chr.13: 40559755-40559755 on Build GRCh38
rs757585400 Chr.13: 40559731-40559731 on Build GRCh38
rs751994432 Chr.13: 40559728-40559728 on Build GRCh38
rs767908809 Chr.13: 40559831-40559831 on Build GRCh38
rs867332663 Chr.13: 40559699-40559699 on Build GRCh38
rs747905007 Chr.13: 40559758-40559758 on Build GRCh38
rs768433269 Chr.13: 40559544-40559544 on Build GRCh38
rs112682206 Chr.13: 40559900-40559900 on Build GRCh38
rs775747228 Chr.13: 40559807-40559807 on Build GRCh38
rs768233462 Chr.13: 40559883-40559883 on Build GRCh38
rs763078745 Chr.13: 40559823-40559823 on Build GRCh38
rs773087473 Chr.13: 40559777-40559777 on Build GRCh38
rs957725137 Chr.13: 40559463-40559463 on Build GRCh38
rs761714739 Chr.13: 40559547-40559547 on Build GRCh38
rs890236166 Chr.13: 40559867-40559867 on Build GRCh38
rs148727582 Chr.13: 40559735-40559735 on Build GRCh38
rs775066641 Chr.13: 40559538-40559538 on Build GRCh38
rs368042937 Chr.13: 40559841-40559841 on Build GRCh38
rs748924681 Chr.13: 40559665-40559665 on Build GRCh38
rs564402015 Chr.13: 40559715-40559715 on Build GRCh38
rs746945626 Chr.13: 40559792-40559792 on Build GRCh38
rs750150806 Chr.13: 40559710-40559710 on Build GRCh38
rs954719653 Chr.13: 40559510-40559510 on Build GRCh38
rs199953664 Chr.13: 40559672-40559672 on Build GRCh38
rs377056137 Chr.13: 40559751-40559751 on Build GRCh38
rs45491494 Chr.13: 40559754-40559754 on Build GRCh38
rs745343501 Chr.13: 40559515-40559515 on Build GRCh38
rs906022358 Chr.13: 40559810-40559810 on Build GRCh38

View Full Product Details