Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43836432-43836982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC122
Entrez Gene ID
160857
Gene Name
coiled-coil domain containing 122
Gene Aliases
-
UniGene
Hs.170849
Gene Chromosome Location
Chr.13: 43821959-43879690 on Build GRCh38
Transcripts
NM_144974.3 XM_017020397.1 XM_017020399.1

SNP Detail

SNP ID Chromosome Location
rs575025294 Chr.13: 43836616-43836616 on Build GRCh38
rs796606524 Chr.13: 43836878-43836878 on Build GRCh38
rs537468209 Chr.13: 43836472-43836472 on Build GRCh38
rs1002502949 Chr.13: 43836621-43836621 on Build GRCh38
rs535190719 Chr.13: 43836552-43836552 on Build GRCh38
rs933128963 Chr.13: 43836766-43836766 on Build GRCh38
rs888787515 Chr.13: 43836573-43836573 on Build GRCh38
rs187054485 Chr.13: 43836458-43836458 on Build GRCh38
rs957946618 Chr.13: 43836722-43836722 on Build GRCh38
rs965307663 Chr.13: 43836856-43836856 on Build GRCh38
rs1041438949 Chr.13: 43836527-43836527 on Build GRCh38
rs913731785 Chr.13: 43836730-43836730 on Build GRCh38
rs1012895120 Chr.13: 43836669-43836670 on Build GRCh38
rs910327942 Chr.13: 43836956-43836956 on Build GRCh38
rs887278123 Chr.13: 43836820-43836820 on Build GRCh38
rs564597628 Chr.13: 43836848-43836848 on Build GRCh38
rs9533646 Chr.13: 43836533-43836533 on Build GRCh38
rs553295637 Chr.13: 43836572-43836572 on Build GRCh38
rs1014096668 Chr.13: 43836646-43836646 on Build GRCh38
rs71099857 Chr.13: 43836877-43836878 on Build GRCh38
rs575611277 Chr.13: 43836807-43836807 on Build GRCh38
rs887235776 Chr.13: 43836636-43836636 on Build GRCh38
rs1019628177 Chr.13: 43836637-43836637 on Build GRCh38
rs911198392 Chr.13: 43836901-43836901 on Build GRCh38
rs987290066 Chr.13: 43836942-43836942 on Build GRCh38
rs933056800 Chr.13: 43836534-43836534 on Build GRCh38
rs1024990655 Chr.13: 43836666-43836666 on Build GRCh38
rs373599099 Chr.13: 43836911-43836911 on Build GRCh38
rs371931213 Chr.13: 43836910-43836910 on Build GRCh38
rs1039993594 Chr.13: 43836611-43836611 on Build GRCh38
rs1025041658 Chr.13: 43836843-43836843 on Build GRCh38
rs1055886626 Chr.13: 43836632-43836632 on Build GRCh38
rs988465203 Chr.13: 43836933-43836933 on Build GRCh38
rs2084086 Chr.13: 43836826-43836826 on Build GRCh38
rs192341917 Chr.13: 43836516-43836516 on Build GRCh38
rs2084085 Chr.13: 43836923-43836923 on Build GRCh38
rs1041534330 Chr.13: 43836943-43836943 on Build GRCh38
rs11420172 Chr.13: 43836856-43836857 on Build GRCh38
rs1047277742 Chr.13: 43836812-43836812 on Build GRCh38
rs942994011 Chr.13: 43836588-43836588 on Build GRCh38
rs1004841804 Chr.13: 43836625-43836625 on Build GRCh38
rs931551634 Chr.13: 43836806-43836806 on Build GRCh38
rs1037352961 Chr.13: 43836796-43836796 on Build GRCh38
rs980724304 Chr.13: 43836707-43836707 on Build GRCh38
rs910275757 Chr.13: 43836777-43836777 on Build GRCh38
rs936535585 Chr.13: 43836481-43836481 on Build GRCh38
rs1026233203 Chr.13: 43836635-43836635 on Build GRCh38
rs969371847 Chr.13: 43836692-43836692 on Build GRCh38
rs547227161 Chr.13: 43836712-43836712 on Build GRCh38
rs1051489072 Chr.13: 43836536-43836536 on Build GRCh38
rs894170341 Chr.13: 43836660-43836660 on Build GRCh38
rs1052677035 Chr.13: 43836517-43836517 on Build GRCh38
rs994346727 Chr.13: 43836762-43836762 on Build GRCh38
rs905266547 Chr.13: 43836860-43836860 on Build GRCh38
rs576532443 Chr.13: 43836899-43836899 on Build GRCh38
rs200031536 Chr.13: 43836589-43836589 on Build GRCh38
rs925170885 Chr.13: 43836476-43836476 on Build GRCh38
rs988026461 Chr.13: 43836729-43836729 on Build GRCh38
rs1020847398 Chr.13: 43836936-43836936 on Build GRCh38
rs868820925 Chr.13: 43836940-43836940 on Build GRCh38
rs2084087 Chr.13: 43836775-43836775 on Build GRCh38
rs943044980 Chr.13: 43836781-43836781 on Build GRCh38
rs9567279 Chr.13: 43836622-43836622 on Build GRCh38
rs898790666 Chr.13: 43836803-43836803 on Build GRCh38

View Full Product Details