Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49308888-49309391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAB39L
Entrez Gene ID
81617
Gene Name
calcium binding protein 39 like
Gene Aliases
MO25-BETA, MO2L, bA103J18.3
UniGene
Hs.742971
Gene Chromosome Location
Chr.13: 49308628-49444346 on Build GRCh38
Transcripts
NM_001079670.2 NM_001287337.1 NM_001287338.1 NM_001287339.1 NM_030925.3 XM_011535254.2 XM_011535255.2 XM_011535256.2 XM_017020785.1 XM_017020786.1 XM_017020787.1 XM_017020788.1

SNP Detail

SNP ID Chromosome Location
rs968046394 Chr.13: 49309314-49309314 on Build GRCh38
rs980846924 Chr.13: 49309319-49309319 on Build GRCh38
rs947249638 Chr.13: 49309180-49309180 on Build GRCh38
rs1018989253 Chr.13: 49309199-49309199 on Build GRCh38
rs908717887 Chr.13: 49309368-49309368 on Build GRCh38
rs952195841 Chr.13: 49309256-49309256 on Build GRCh38
rs898280134 Chr.13: 49309038-49309038 on Build GRCh38
rs980853569 Chr.13: 49309173-49309173 on Build GRCh38
rs981653861 Chr.13: 49309367-49309367 on Build GRCh38
rs972134142 Chr.13: 49309076-49309076 on Build GRCh38
rs1008130343 Chr.13: 49309197-49309197 on Build GRCh38
rs7330440 Chr.13: 49309169-49309169 on Build GRCh38
rs952249245 Chr.13: 49309095-49309095 on Build GRCh38
rs764719404 Chr.13: 49309179-49309179 on Build GRCh38
rs553569391 Chr.13: 49309286-49309286 on Build GRCh38
rs572404827 Chr.13: 49309262-49309262 on Build GRCh38
rs956901930 Chr.13: 49309138-49309138 on Build GRCh38
rs905377318 Chr.13: 49308934-49308934 on Build GRCh38
rs571460799 Chr.13: 49309162-49309162 on Build GRCh38
rs375939647 Chr.13: 49309115-49309115 on Build GRCh38
rs926768907 Chr.13: 49309328-49309328 on Build GRCh38
rs550402195 Chr.13: 49309141-49309141 on Build GRCh38
rs755873372 Chr.13: 49309142-49309142 on Build GRCh38
rs1031993044 Chr.13: 49309280-49309280 on Build GRCh38
rs934394613 Chr.13: 49308947-49308947 on Build GRCh38
rs527663119 Chr.13: 49308923-49308923 on Build GRCh38
rs889668626 Chr.13: 49308991-49308991 on Build GRCh38
rs890950290 Chr.13: 49309249-49309249 on Build GRCh38
rs993458437 Chr.13: 49309248-49309248 on Build GRCh38
rs1018933936 Chr.13: 49309062-49309062 on Build GRCh38
rs538307095 Chr.13: 49309022-49309022 on Build GRCh38
rs1026406074 Chr.13: 49309255-49309255 on Build GRCh38
rs567566030 Chr.13: 49308971-49308971 on Build GRCh38
rs990083321 Chr.13: 49309305-49309306 on Build GRCh38
rs777866106 Chr.13: 49309145-49309150 on Build GRCh38
rs1052915233 Chr.13: 49308965-49308965 on Build GRCh38
rs889729523 Chr.13: 49309195-49309195 on Build GRCh38
rs370155171 Chr.13: 49309160-49309160 on Build GRCh38
rs549390848 Chr.13: 49308966-49308966 on Build GRCh38
rs1016623061 Chr.13: 49309015-49309015 on Build GRCh38
rs9535197 Chr.13: 49309308-49309308 on Build GRCh38
rs1011948997 Chr.13: 49308968-49308968 on Build GRCh38
rs554088923 Chr.13: 49309185-49309185 on Build GRCh38
rs181014819 Chr.13: 49308918-49308918 on Build GRCh38
rs1026938475 Chr.13: 49309094-49309094 on Build GRCh38
rs9535196 Chr.13: 49309178-49309178 on Build GRCh38
rs780874682 Chr.13: 49309116-49309116 on Build GRCh38
rs868137418 Chr.13: 49309347-49309347 on Build GRCh38
rs777402753 Chr.13: 49309156-49309156 on Build GRCh38
rs574311314 Chr.13: 49308915-49308915 on Build GRCh38
rs184102238 Chr.13: 49309102-49309102 on Build GRCh38
rs996514916 Chr.13: 49309200-49309200 on Build GRCh38
rs1049997575 Chr.13: 49309191-49309191 on Build GRCh38
rs1007579179 Chr.13: 49309026-49309026 on Build GRCh38
rs372499585 Chr.13: 49309065-49309065 on Build GRCh38
rs1006406331 Chr.13: 49309300-49309300 on Build GRCh38
rs901840597 Chr.13: 49309245-49309245 on Build GRCh38
rs1004161881 Chr.13: 49308952-49308952 on Build GRCh38

View Full Product Details