Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 75284605-75285101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D4
Entrez Gene ID
9882
Gene Name
TBC1 domain family member 4
Gene Aliases
AS160, NIDDM5
UniGene
Hs.210891
Gene Chromosome Location
Chr.13: 75284664-75482646 on Build GRCh38
Transcripts
NM_001286658.2 NM_001286659.2 NM_014832.4 XM_005266603.1 XM_005266605.2 XM_006719903.3 XM_011535331.1 XM_017020882.1 XM_017020883.1 XM_017020884.1

SNP Detail

SNP ID Chromosome Location
rs150881540 Chr.13: 75285010-75285010 on Build GRCh38
rs15167 Chr.13: 75284973-75284973 on Build GRCh38
rs1022147497 Chr.13: 75284678-75284678 on Build GRCh38
rs778167183 Chr.13: 75284889-75284889 on Build GRCh38
rs906664822 Chr.13: 75285001-75285001 on Build GRCh38
rs1010513748 Chr.13: 75285034-75285034 on Build GRCh38
rs1058830 Chr.13: 75285009-75285009 on Build GRCh38
rs141538982 Chr.13: 75284738-75284738 on Build GRCh38
rs1029279465 Chr.13: 75284822-75284822 on Build GRCh38
rs1058845 Chr.13: 75284737-75284737 on Build GRCh38
rs920671771 Chr.13: 75285028-75285028 on Build GRCh38
rs1058841 Chr.13: 75284758-75284758 on Build GRCh38
rs1046373545 Chr.13: 75284715-75284715 on Build GRCh38
rs916757469 Chr.13: 75284976-75284976 on Build GRCh38
rs529077533 Chr.13: 75285078-75285078 on Build GRCh38
rs559955426 Chr.13: 75284843-75284843 on Build GRCh38
rs948302311 Chr.13: 75284963-75284963 on Build GRCh38
rs892237229 Chr.13: 75285016-75285016 on Build GRCh38
rs187788782 Chr.13: 75284748-75284748 on Build GRCh38
rs1036400803 Chr.13: 75285024-75285024 on Build GRCh38
rs751081393 Chr.13: 75284650-75284650 on Build GRCh38
rs948332774 Chr.13: 75284682-75284682 on Build GRCh38
rs537028401 Chr.13: 75284668-75284668 on Build GRCh38
rs898957640 Chr.13: 75284755-75284755 on Build GRCh38
rs542549548 Chr.13: 75284880-75284880 on Build GRCh38
rs1009255250 Chr.13: 75284862-75284862 on Build GRCh38
rs139004273 Chr.13: 75284666-75284666 on Build GRCh38
rs546338395 Chr.13: 75285029-75285029 on Build GRCh38
rs920835552 Chr.13: 75284962-75284962 on Build GRCh38
rs756718385 Chr.13: 75284879-75284879 on Build GRCh38
rs377111156 Chr.13: 75284661-75284661 on Build GRCh38
rs11566 Chr.13: 75284931-75284931 on Build GRCh38
rs75054979 Chr.13: 75284667-75284667 on Build GRCh38
rs987537960 Chr.13: 75284799-75284799 on Build GRCh38
rs545829869 Chr.13: 75284992-75284992 on Build GRCh38
rs564043763 Chr.13: 75285002-75285002 on Build GRCh38
rs940670279 Chr.13: 75284925-75284925 on Build GRCh38
rs575339926 Chr.13: 75284984-75284984 on Build GRCh38
rs749664326 Chr.13: 75284911-75284911 on Build GRCh38
rs574894168 Chr.13: 75284770-75284770 on Build GRCh38
rs534764836 Chr.13: 75284744-75284744 on Build GRCh38
rs561960973 Chr.13: 75285037-75285037 on Build GRCh38

View Full Product Details