Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 75306221-75306725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D4
Entrez Gene ID
9882
Gene Name
TBC1 domain family member 4
Gene Aliases
AS160, NIDDM5
UniGene
Hs.210891
Gene Chromosome Location
Chr.13: 75284664-75482646 on Build GRCh38
Transcripts
NM_001286658.2 NM_001286659.2 NM_014832.4 XM_005266603.1 XM_005266605.2 XM_006719903.3 XM_011535331.1 XM_017020882.1 XM_017020883.1 XM_017020884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6568782 c.2594-6C>T p.? Chr.13: 75306477-75306477 on Build GRCh38
1639377 c.2710delA p.I904fs*53 Chr.13: 75306355-75306355 on Build GRCh38
4453235 c.2686A>G p.K896E Chr.13: 75306379-75306379 on Build GRCh38
5049488 c.2653T>C p.C885R Chr.13: 75306412-75306412 on Build GRCh38
3814040 c.2632G>C p.D878H Chr.13: 75306433-75306433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs33992075 Chr.13: 75306249-75306249 on Build GRCh38
rs202064304 Chr.13: 75306402-75306402 on Build GRCh38
rs573386893 Chr.13: 75306661-75306661 on Build GRCh38
rs760242565 Chr.13: 75306366-75306366 on Build GRCh38
rs200791184 Chr.13: 75306248-75306248 on Build GRCh38
rs546573144 Chr.13: 75306251-75306252 on Build GRCh38
rs751714544 Chr.13: 75306413-75306413 on Build GRCh38
rs764657106 Chr.13: 75306411-75306411 on Build GRCh38
rs577975603 Chr.13: 75306603-75306603 on Build GRCh38
rs891559134 Chr.13: 75306586-75306586 on Build GRCh38
rs562358706 Chr.13: 75306492-75306492 on Build GRCh38
rs779699251 Chr.13: 75306332-75306332 on Build GRCh38
rs376620394 Chr.13: 75306502-75306502 on Build GRCh38
rs959629061 Chr.13: 75306614-75306614 on Build GRCh38
rs765695310 Chr.13: 75306378-75306378 on Build GRCh38
rs868504167 Chr.13: 75306450-75306450 on Build GRCh38
rs756002956 Chr.13: 75306330-75306330 on Build GRCh38
rs779935663 Chr.13: 75306466-75306466 on Build GRCh38
rs747604452 Chr.13: 75306351-75306351 on Build GRCh38
rs150157071 Chr.13: 75306656-75306656 on Build GRCh38
rs1041297195 Chr.13: 75306478-75306478 on Build GRCh38
rs373004177 Chr.13: 75306447-75306447 on Build GRCh38
rs771879786 Chr.13: 75306250-75306250 on Build GRCh38
rs772857668 Chr.13: 75306355-75306355 on Build GRCh38
rs1039008892 Chr.13: 75306533-75306536 on Build GRCh38
rs374228481 Chr.13: 75306292-75306292 on Build GRCh38
rs767489753 Chr.13: 75306429-75306429 on Build GRCh38
rs952057666 Chr.13: 75306532-75306532 on Build GRCh38
rs772587234 Chr.13: 75306518-75306518 on Build GRCh38
rs17064121 Chr.13: 75306545-75306545 on Build GRCh38
rs2297209 Chr.13: 75306531-75306531 on Build GRCh38
rs549414313 Chr.13: 75306251-75306251 on Build GRCh38
rs1033889527 Chr.13: 75306579-75306579 on Build GRCh38
rs869075043 Chr.13: 75306258-75306259 on Build GRCh38
rs765669248 Chr.13: 75306267-75306267 on Build GRCh38
rs919364362 Chr.13: 75306477-75306478 on Build GRCh38
rs398023464 Chr.13: 75306260-75306260 on Build GRCh38
rs748992763 Chr.13: 75306486-75306486 on Build GRCh38
rs58430556 Chr.13: 75306249-75306250 on Build GRCh38
rs1034369534 Chr.13: 75306678-75306678 on Build GRCh38
rs757338666 Chr.13: 75306419-75306419 on Build GRCh38
rs933010892 Chr.13: 75306578-75306578 on Build GRCh38
rs968276363 Chr.13: 75306312-75306312 on Build GRCh38
rs751659576 Chr.13: 75306299-75306299 on Build GRCh38
rs757991747 Chr.13: 75306485-75306485 on Build GRCh38
rs563251195 Chr.13: 75306259-75306260 on Build GRCh38
rs761565683 Chr.13: 75306595-75306595 on Build GRCh38
rs369089434 Chr.13: 75306382-75306382 on Build GRCh38
rs140884591 Chr.13: 75306646-75306646 on Build GRCh38
rs771640992 Chr.13: 75306506-75306506 on Build GRCh38
rs763595626 Chr.13: 75306398-75306398 on Build GRCh38
rs747665369 Chr.13: 75306503-75306503 on Build GRCh38
rs778762841 Chr.13: 75306493-75306493 on Build GRCh38
rs201045831 Chr.13: 75306260-75306261 on Build GRCh38
rs945132030 Chr.13: 75306252-75306252 on Build GRCh38
rs1052845994 Chr.13: 75306583-75306583 on Build GRCh38
rs781517791 Chr.13: 75306307-75306307 on Build GRCh38
rs541317362 Chr.13: 75306249-75306251 on Build GRCh38
rs745653179 Chr.13: 75306316-75306316 on Build GRCh38
rs768545937 Chr.13: 75306333-75306333 on Build GRCh38
rs375892188 Chr.13: 75306350-75306350 on Build GRCh38
rs778049006 Chr.13: 75306295-75306295 on Build GRCh38
rs904062858 Chr.13: 75306649-75306649 on Build GRCh38
rs182577521 Chr.13: 75306276-75306276 on Build GRCh38
rs1042042355 Chr.13: 75306286-75306286 on Build GRCh38
rs758964780 Chr.13: 75306291-75306291 on Build GRCh38
rs757586535 Chr.13: 75306304-75306304 on Build GRCh38
rs551937789 Chr.13: 75306250-75306251 on Build GRCh38
rs529510908 Chr.13: 75306530-75306530 on Build GRCh38

View Full Product Details