Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 78601887-78602425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POU4F1
Entrez Gene ID
5457
Gene Name
POU class 4 homeobox 1
Gene Aliases
BRN3A, Oct-T1, RDC-1, brn-3A
UniGene
Hs.654522
Gene Chromosome Location
Chr.13: 78599092-78603560 on Build GRCh38
Transcripts
NM_006237.3 NR_047001.1

Gene Symbol
RNF219-AS1
Entrez Gene ID
100874222
Gene Name
RNF219 antisense RNA 1
Gene Aliases
POU4F1-AS1
UniGene
Hs.493062
Gene Chromosome Location
Chr.13: 78054855-78617325 on Build GRCh38
Transcripts
NM_006237.3 NR_047001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7209157 c.298_300delCAC p.H108delH Chr.13: 78602375-78602377 on Build GRCh38
4410228 c.368C>T p.P123L Chr.13: 78602307-78602307 on Build GRCh38
948744 c.334G>A p.E112K Chr.13: 78602341-78602341 on Build GRCh38
6537990 c.726G>C p.A242A Chr.13: 78601949-78601949 on Build GRCh38
948745 c.284C>T p.T95M Chr.13: 78602391-78602391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780413396 Chr.13: 78602247-78602282 on Build GRCh38
rs1008460368 Chr.13: 78602112-78602112 on Build GRCh38
rs1013723680 Chr.13: 78602258-78602258 on Build GRCh38
rs962919584 Chr.13: 78602291-78602291 on Build GRCh38
rs866960156 Chr.13: 78601982-78601982 on Build GRCh38
rs149245376 Chr.13: 78602392-78602392 on Build GRCh38
rs552672805 Chr.13: 78602188-78602189 on Build GRCh38
rs762014663 Chr.13: 78602349-78602351 on Build GRCh38
rs995759174 Chr.13: 78602294-78602294 on Build GRCh38
rs368562136 Chr.13: 78602393-78602393 on Build GRCh38
rs925126425 Chr.13: 78602391-78602391 on Build GRCh38
rs1004422908 Chr.13: 78602273-78602273 on Build GRCh38
rs750756634 Chr.13: 78601922-78601922 on Build GRCh38
rs763743520 Chr.13: 78601921-78601921 on Build GRCh38
rs866440488 Chr.13: 78602135-78602135 on Build GRCh38
rs746799445 Chr.13: 78602342-78602342 on Build GRCh38
rs1004858148 Chr.13: 78602000-78602000 on Build GRCh38
rs748551009 Chr.13: 78602349-78602354 on Build GRCh38
rs866826021 Chr.13: 78602231-78602231 on Build GRCh38
rs546818659 Chr.13: 78602345-78602345 on Build GRCh38
rs1042322522 Chr.13: 78601931-78601931 on Build GRCh38
rs757980850 Chr.13: 78601931-78601932 on Build GRCh38
rs1042680630 Chr.13: 78602103-78602103 on Build GRCh38
rs752771035 Chr.13: 78602324-78602324 on Build GRCh38
rs113980525 Chr.13: 78601914-78601914 on Build GRCh38
rs1024870460 Chr.13: 78601977-78601988 on Build GRCh38
rs773356981 Chr.13: 78602349-78602360 on Build GRCh38
rs966943701 Chr.13: 78602380-78602380 on Build GRCh38
rs570767813 Chr.13: 78602297-78602297 on Build GRCh38
rs539817405 Chr.13: 78602344-78602344 on Build GRCh38
rs375560814 Chr.13: 78602378-78602378 on Build GRCh38
rs116563781 Chr.13: 78602390-78602390 on Build GRCh38
rs757052426 Chr.13: 78602371-78602371 on Build GRCh38
rs372609451 Chr.13: 78602321-78602321 on Build GRCh38
rs925243460 Chr.13: 78602131-78602131 on Build GRCh38
rs1054684368 Chr.13: 78602230-78602230 on Build GRCh38
rs998792501 Chr.13: 78602247-78602267 on Build GRCh38
rs754365572 Chr.13: 78601969-78601969 on Build GRCh38
rs1015869966 Chr.13: 78602001-78602001 on Build GRCh38
rs762580667 Chr.13: 78601920-78601920 on Build GRCh38
rs867739817 Chr.13: 78602039-78602039 on Build GRCh38
rs758556486 Chr.13: 78602333-78602333 on Build GRCh38
rs550817835 Chr.13: 78602268-78602268 on Build GRCh38
rs986924172 Chr.13: 78602075-78602075 on Build GRCh38
rs1013296836 Chr.13: 78601973-78601973 on Build GRCh38
rs912359047 Chr.13: 78602078-78602078 on Build GRCh38
rs886976374 Chr.13: 78602046-78602046 on Build GRCh38
rs190268005 Chr.13: 78602134-78602134 on Build GRCh38
rs747434243 Chr.13: 78602310-78602312 on Build GRCh38
rs745797845 Chr.13: 78602379-78602379 on Build GRCh38
rs769945525 Chr.13: 78602349-78602369 on Build GRCh38
rs903517905 Chr.13: 78601940-78601940 on Build GRCh38
rs768216181 Chr.13: 78602403-78602403 on Build GRCh38
rs1015844835 Chr.13: 78602288-78602288 on Build GRCh38
rs1054800175 Chr.13: 78601958-78601958 on Build GRCh38
rs761052903 Chr.13: 78601923-78601923 on Build GRCh38
rs921271989 Chr.13: 78602047-78602047 on Build GRCh38
rs781744436 Chr.13: 78602348-78602349 on Build GRCh38
rs987277524 Chr.13: 78602349-78602357 on Build GRCh38
rs1000511431 Chr.13: 78601953-78601953 on Build GRCh38
rs1046175374 Chr.13: 78602264-78602264 on Build GRCh38
rs766846140 Chr.13: 78601942-78601942 on Build GRCh38
rs962699546 Chr.13: 78602032-78602032 on Build GRCh38
rs755175983 Chr.13: 78602080-78602080 on Build GRCh38
rs778019885 Chr.13: 78602338-78602338 on Build GRCh38
rs954069424 Chr.13: 78602067-78602067 on Build GRCh38
rs530848977 Chr.13: 78602267-78602267 on Build GRCh38
rs1047242679 Chr.13: 78602023-78602023 on Build GRCh38
rs907345569 Chr.13: 78602269-78602277 on Build GRCh38
rs568393161 Chr.13: 78601948-78601948 on Build GRCh38

View Full Product Details